Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73176397-73176910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEO1
Entrez Gene ID
4756
Gene Name
neogenin 1
Gene Aliases
IGDCC2, NGN, NTN1R2
UniGene
Hs.388613
Gene Chromosome Location
Chr.15: 73051715-73305206 on Build GRCh38
Transcripts
NM_001172623.1 NM_001172624.1 NM_002499.3 XM_005254408.1 XM_011521628.2 XM_011521629.2 XM_011521630.2 XM_011521632.2 XM_011521633.2 XM_017022231.1 XM_017022232.1 XM_017022233.1 XM_017022234.1 XM_017022235.1 XM_017022236.1 XM_017022237.1 XM_017022238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6724675 c.1060G>A p.E354K Chr.15: 73176447-73176447 on Build GRCh38
389162 c.1082A>T p.E361V Chr.15: 73176469-73176469 on Build GRCh38
5953894 c.1092G>A p.V364V Chr.15: 73176479-73176479 on Build GRCh38
3503741 c.1109C>T p.P370L Chr.15: 73176496-73176496 on Build GRCh38
1937562 c.1150A>C p.S384R Chr.15: 73176537-73176537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908457365 Chr.15: 73176758-73176758 on Build GRCh38
rs762810167 Chr.15: 73176604-73176604 on Build GRCh38
rs963054569 Chr.15: 73176816-73176816 on Build GRCh38
rs960683931 Chr.15: 73176737-73176737 on Build GRCh38
rs1053337420 Chr.15: 73176884-73176884 on Build GRCh38
rs762327984 Chr.15: 73176571-73176571 on Build GRCh38
rs764969367 Chr.15: 73176598-73176598 on Build GRCh38
rs377655879 Chr.15: 73176581-73176581 on Build GRCh38
rs181530230 Chr.15: 73176642-73176642 on Build GRCh38
rs568544532 Chr.15: 73176676-73176676 on Build GRCh38
rs536380681 Chr.15: 73176732-73176732 on Build GRCh38
rs769061384 Chr.15: 73176507-73176507 on Build GRCh38
rs996751043 Chr.15: 73176653-73176653 on Build GRCh38
rs1053815629 Chr.15: 73176553-73176553 on Build GRCh38
rs921325688 Chr.15: 73176864-73176865 on Build GRCh38
rs757174587 Chr.15: 73176453-73176453 on Build GRCh38
rs199626965 Chr.15: 73176596-73176596 on Build GRCh38
rs907616413 Chr.15: 73176435-73176435 on Build GRCh38
rs1014628697 Chr.15: 73176797-73176797 on Build GRCh38
rs983530000 Chr.15: 73176429-73176429 on Build GRCh38
rs747007237 Chr.15: 73176447-73176447 on Build GRCh38
rs571077961 Chr.15: 73176482-73176482 on Build GRCh38
rs902586195 Chr.15: 73176636-73176636 on Build GRCh38
rs766302441 Chr.15: 73176713-73176713 on Build GRCh38
rs895732110 Chr.15: 73176606-73176606 on Build GRCh38
rs962539938 Chr.15: 73176782-73176782 on Build GRCh38
rs149313472 Chr.15: 73176499-73176499 on Build GRCh38
rs138589758 Chr.15: 73176498-73176498 on Build GRCh38
rs372456375 Chr.15: 73176791-73176791 on Build GRCh38
rs554676236 Chr.15: 73176761-73176761 on Build GRCh38
rs912557540 Chr.15: 73176880-73176880 on Build GRCh38
rs1030548027 Chr.15: 73176666-73176666 on Build GRCh38
rs955163461 Chr.15: 73176691-73176691 on Build GRCh38
rs758233986 Chr.15: 73176670-73176670 on Build GRCh38
rs1034675987 Chr.15: 73176650-73176650 on Build GRCh38
rs937159148 Chr.15: 73176877-73176877 on Build GRCh38
rs752444138 Chr.15: 73176599-73176599 on Build GRCh38
rs927083632 Chr.15: 73176485-73176485 on Build GRCh38
rs927034604 Chr.15: 73176871-73176871 on Build GRCh38
rs1011756341 Chr.15: 73176583-73176583 on Build GRCh38
rs969142748 Chr.15: 73176623-73176623 on Build GRCh38
rs769938083 Chr.15: 73176497-73176497 on Build GRCh38
rs111282698 Chr.15: 73176613-73176613 on Build GRCh38
rs949407601 Chr.15: 73176622-73176622 on Build GRCh38
rs200345773 Chr.15: 73176588-73176588 on Build GRCh38
rs777548596 Chr.15: 73176446-73176446 on Build GRCh38
rs201453469 Chr.15: 73176428-73176428 on Build GRCh38
rs982716297 Chr.15: 73176743-73176743 on Build GRCh38
rs1000931441 Chr.15: 73176641-73176641 on Build GRCh38
rs900655886 Chr.15: 73176644-73176644 on Build GRCh38
rs893304662 Chr.15: 73176557-73176560 on Build GRCh38
rs781284375 Chr.15: 73176455-73176455 on Build GRCh38
rs934699142 Chr.15: 73176883-73176883 on Build GRCh38
rs774693358 Chr.15: 73176560-73176560 on Build GRCh38
rs973889622 Chr.15: 73176811-73176811 on Build GRCh38
rs973126630 Chr.15: 73176861-73176861 on Build GRCh38
rs370874079 Chr.15: 73176585-73176585 on Build GRCh38
rs1042820423 Chr.15: 73176627-73176627 on Build GRCh38

View Full Product Details