Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82648035-82648528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB1-AS1
Entrez Gene ID
283692
Gene Name
CPEB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.578667
Gene Chromosome Location
Chr.15: 82647770-82692820 on Build GRCh38
Transcripts
NR_046096.1

SNP Detail

SNP ID Chromosome Location
rs189993435 Chr.15: 82648229-82648229 on Build GRCh38
rs937838723 Chr.15: 82648350-82648350 on Build GRCh38
rs767806290 Chr.15: 82648456-82648456 on Build GRCh38
rs587726835 Chr.15: 82648503-82648503 on Build GRCh38
rs587610067 Chr.15: 82648188-82648188 on Build GRCh38
rs1039691739 Chr.15: 82648410-82648410 on Build GRCh38
rs1046326634 Chr.15: 82648144-82648144 on Build GRCh38
rs993544174 Chr.15: 82648197-82648197 on Build GRCh38
rs956213044 Chr.15: 82648431-82648431 on Build GRCh38
rs925059002 Chr.15: 82648108-82648108 on Build GRCh38
rs949102395 Chr.15: 82648319-82648319 on Build GRCh38
rs926283642 Chr.15: 82648479-82648479 on Build GRCh38
rs28592023 Chr.15: 82648088-82648088 on Build GRCh38
rs587749332 Chr.15: 82648135-82648135 on Build GRCh38
rs923962786 Chr.15: 82648125-82648125 on Build GRCh38
rs1018167062 Chr.15: 82648064-82648064 on Build GRCh38
rs981838075 Chr.15: 82648478-82648478 on Build GRCh38
rs978993407 Chr.15: 82648275-82648275 on Build GRCh38
rs978601876 Chr.15: 82648301-82648301 on Build GRCh38
rs587717724 Chr.15: 82648367-82648367 on Build GRCh38
rs587612594 Chr.15: 82648477-82648477 on Build GRCh38
rs1008934101 Chr.15: 82648391-82648391 on Build GRCh38
rs587714278 Chr.15: 82648385-82648385 on Build GRCh38
rs764103990 Chr.15: 82648137-82648137 on Build GRCh38
rs957743703 Chr.15: 82648284-82648284 on Build GRCh38
rs978074525 Chr.15: 82648124-82648124 on Build GRCh38
rs587761438 Chr.15: 82648178-82648178 on Build GRCh38
rs943763619 Chr.15: 82648419-82648419 on Build GRCh38
rs890474749 Chr.15: 82648228-82648228 on Build GRCh38
rs975556011 Chr.15: 82648100-82648100 on Build GRCh38
rs954919359 Chr.15: 82648057-82648057 on Build GRCh38
rs913580573 Chr.15: 82648118-82648118 on Build GRCh38
rs896829927 Chr.15: 82648183-82648184 on Build GRCh38
rs970789924 Chr.15: 82648469-82648469 on Build GRCh38
rs974418776 Chr.15: 82648096-82648096 on Build GRCh38
rs780983406 Chr.15: 82648257-82648257 on Build GRCh38
rs866128025 Chr.15: 82648352-82648352 on Build GRCh38
rs1027324638 Chr.15: 82648230-82648230 on Build GRCh38
rs987904620 Chr.15: 82648293-82648293 on Build GRCh38
rs587761043 Chr.15: 82648286-82648286 on Build GRCh38
rs900715748 Chr.15: 82648415-82648415 on Build GRCh38
rs937887222 Chr.15: 82648159-82648159 on Build GRCh38
rs587771625 Chr.15: 82648210-82648210 on Build GRCh38
rs936675949 Chr.15: 82648343-82648343 on Build GRCh38
rs1015240254 Chr.15: 82648183-82648183 on Build GRCh38
rs587717162 Chr.15: 82648201-82648201 on Build GRCh38
rs932156779 Chr.15: 82648151-82648151 on Build GRCh38
rs949300542 Chr.15: 82648127-82648127 on Build GRCh38
rs890646495 Chr.15: 82648387-82648387 on Build GRCh38
rs1033152429 Chr.15: 82648426-82648426 on Build GRCh38
rs112454974 Chr.15: 82648378-82648378 on Build GRCh38
rs900637249 Chr.15: 82648150-82648150 on Build GRCh38
rs183372420 Chr.15: 82648244-82648244 on Build GRCh38
rs587669363 Chr.15: 82648114-82648114 on Build GRCh38
rs926592602 Chr.15: 82648342-82648342 on Build GRCh38
rs1000458738 Chr.15: 82648422-82648422 on Build GRCh38
rs587634639 Chr.15: 82648078-82648078 on Build GRCh38
rs989711492 Chr.15: 82648356-82648356 on Build GRCh38
rs1048097478 Chr.15: 82648451-82648451 on Build GRCh38
rs587601112 Chr.15: 82648369-82648369 on Build GRCh38
rs905004178 Chr.15: 82648145-82648145 on Build GRCh38
rs899011462 Chr.15: 82648358-82648358 on Build GRCh38
rs1037457820 Chr.15: 82648357-82648357 on Build GRCh38
rs1026238634 Chr.15: 82648209-82648209 on Build GRCh38
rs932271711 Chr.15: 82648372-82648372 on Build GRCh38
rs780243252 Chr.15: 82648446-82648446 on Build GRCh38
rs1033183398 Chr.15: 82648277-82648277 on Build GRCh38
rs899294849 Chr.15: 82648195-82648195 on Build GRCh38
rs186702267 Chr.15: 82648177-82648177 on Build GRCh38
rs113359247 Chr.15: 82648360-82648360 on Build GRCh38
rs587625711 Chr.15: 82648168-82648168 on Build GRCh38
rs763746041 Chr.15: 82648298-82648298 on Build GRCh38
rs150015470 Chr.15: 82648376-82648376 on Build GRCh38
rs1009300027 Chr.15: 82648445-82648445 on Build GRCh38
rs965236247 Chr.15: 82648270-82648270 on Build GRCh38
rs959206299 Chr.15: 82648103-82648103 on Build GRCh38
rs936321928 Chr.15: 82648109-82648109 on Build GRCh38
rs587669421 Chr.15: 82648502-82648502 on Build GRCh38
rs8038497 Chr.15: 82648330-82648330 on Build GRCh38
rs987748564 Chr.15: 82648060-82648060 on Build GRCh38

View Full Product Details