Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11377197-11377741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPOR
Entrez Gene ID
2057
Gene Name
erythropoietin receptor
Gene Aliases
EPO-R
UniGene
Hs.631624
Gene Chromosome Location
Chr.19: 11377205-11384342 on Build GRCh38
Transcripts
NM_000121.3 NR_033663.1

SNP Detail

SNP ID Chromosome Location
rs974040661 Chr.19: 11377629-11377629 on Build GRCh38
rs756216403 Chr.19: 11377409-11377409 on Build GRCh38
rs942574587 Chr.19: 11377675-11377675 on Build GRCh38
rs756134980 Chr.19: 11377255-11377255 on Build GRCh38
rs1021924996 Chr.19: 11377420-11377420 on Build GRCh38
rs150535617 Chr.19: 11377480-11377480 on Build GRCh38
rs931013548 Chr.19: 11377331-11377331 on Build GRCh38
rs781493978 Chr.19: 11377329-11377329 on Build GRCh38
rs745615457 Chr.19: 11377369-11377369 on Build GRCh38
rs975291752 Chr.19: 11377680-11377680 on Build GRCh38
rs1045852426 Chr.19: 11377670-11377670 on Build GRCh38
rs868528174 Chr.19: 11377564-11377564 on Build GRCh38
rs986677292 Chr.19: 11377262-11377262 on Build GRCh38
rs191481987 Chr.19: 11377707-11377707 on Build GRCh38
rs886884768 Chr.19: 11377373-11377373 on Build GRCh38
rs934445087 Chr.19: 11377697-11377697 on Build GRCh38
rs955153071 Chr.19: 11377520-11377520 on Build GRCh38
rs185249501 Chr.19: 11377587-11377587 on Build GRCh38
rs775302463 Chr.19: 11377348-11377348 on Build GRCh38
rs948456670 Chr.19: 11377274-11377274 on Build GRCh38
rs558924254 Chr.19: 11377698-11377698 on Build GRCh38
rs764896894 Chr.19: 11377631-11377631 on Build GRCh38
rs912447647 Chr.19: 11377297-11377297 on Build GRCh38
rs772920916 Chr.19: 11377640-11377640 on Build GRCh38
rs757491897 Chr.19: 11377324-11377324 on Build GRCh38
rs982622373 Chr.19: 11377529-11377529 on Build GRCh38
rs542954029 Chr.19: 11377300-11377300 on Build GRCh38
rs987002796 Chr.19: 11377646-11377646 on Build GRCh38
rs915620216 Chr.19: 11377268-11377268 on Build GRCh38
rs1035876227 Chr.19: 11377378-11377378 on Build GRCh38
rs749474213 Chr.19: 11377312-11377312 on Build GRCh38
rs893380239 Chr.19: 11377364-11377364 on Build GRCh38
rs763511227 Chr.19: 11377453-11377453 on Build GRCh38
rs576181733 Chr.19: 11377352-11377352 on Build GRCh38
rs140441486 Chr.19: 11377559-11377559 on Build GRCh38
rs771124995 Chr.19: 11377314-11377314 on Build GRCh38
rs780044495 Chr.19: 11377469-11377469 on Build GRCh38
rs752004469 Chr.19: 11377246-11377246 on Build GRCh38
rs1011820847 Chr.19: 11377367-11377367 on Build GRCh38
rs751952585 Chr.19: 11377689-11377689 on Build GRCh38
rs768450038 Chr.19: 11377357-11377359 on Build GRCh38
rs1003429180 Chr.19: 11377376-11377376 on Build GRCh38
rs746137508 Chr.19: 11377386-11377386 on Build GRCh38
rs753197605 Chr.19: 11377712-11377712 on Build GRCh38
rs894685727 Chr.19: 11377406-11377406 on Build GRCh38
rs376736196 Chr.19: 11377307-11377307 on Build GRCh38
rs912397802 Chr.19: 11377656-11377656 on Build GRCh38
rs924755481 Chr.19: 11377245-11377245 on Build GRCh38
rs774307582 Chr.19: 11377323-11377323 on Build GRCh38
rs530287783 Chr.19: 11377700-11377700 on Build GRCh38
rs772172210 Chr.19: 11377377-11377377 on Build GRCh38
rs760294447 Chr.19: 11377436-11377436 on Build GRCh38
rs886054191 Chr.19: 11377365-11377365 on Build GRCh38
rs143402071 Chr.19: 11377247-11377247 on Build GRCh38
rs148340466 Chr.19: 11377321-11377321 on Build GRCh38
rs1047237655 Chr.19: 11377366-11377366 on Build GRCh38
rs908410630 Chr.19: 11377554-11377554 on Build GRCh38
rs141524122 Chr.19: 11377407-11377407 on Build GRCh38

View Full Product Details