Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77713295-77713794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT6L
Entrez Gene ID
246175
Gene Name
CCR4-NOT transcription complex subunit 6 like
Gene Aliases
CCR4b
UniGene
Hs.592519
Gene Chromosome Location
Chr.4: 77713387-77820238 on Build GRCh38
Transcripts
NM_001286790.1 NM_144571.2 XM_011531806.2 XM_011531807.2 XM_011531808.2 XM_011531809.1 XM_011531810.1 XM_017007956.1 XM_017007957.1

SNP Detail

SNP ID Chromosome Location
rs1012730784 Chr.4: 77713705-77713705 on Build GRCh38
rs3199402 Chr.4: 77713593-77713593 on Build GRCh38
rs1040836404 Chr.4: 77713668-77713668 on Build GRCh38
rs994296288 Chr.4: 77713756-77713756 on Build GRCh38
rs752449542 Chr.4: 77713716-77713716 on Build GRCh38
rs532898424 Chr.4: 77713490-77713490 on Build GRCh38
rs759168376 Chr.4: 77713664-77713664 on Build GRCh38
rs565433926 Chr.4: 77713445-77713445 on Build GRCh38
rs992304120 Chr.4: 77713494-77713494 on Build GRCh38
rs548465115 Chr.4: 77713339-77713340 on Build GRCh38
rs1003512667 Chr.4: 77713633-77713633 on Build GRCh38
rs943111671 Chr.4: 77713355-77713355 on Build GRCh38
rs565643354 Chr.4: 77713735-77713735 on Build GRCh38
rs917769309 Chr.4: 77713546-77713546 on Build GRCh38
rs766125306 Chr.4: 77713570-77713570 on Build GRCh38
rs895171271 Chr.4: 77713693-77713693 on Build GRCh38
rs904144357 Chr.4: 77713742-77713742 on Build GRCh38
rs150175318 Chr.4: 77713621-77713622 on Build GRCh38
rs987549735 Chr.4: 77713324-77713324 on Build GRCh38
rs1017032421 Chr.4: 77713713-77713713 on Build GRCh38
rs767220148 Chr.4: 77713671-77713671 on Build GRCh38
rs528128108 Chr.4: 77713323-77713323 on Build GRCh38
rs535075715 Chr.4: 77713645-77713645 on Build GRCh38
rs569336318 Chr.4: 77713626-77713626 on Build GRCh38
rs751155967 Chr.4: 77713610-77713610 on Build GRCh38
rs528742883 Chr.4: 77713340-77713340 on Build GRCh38
rs1030669713 Chr.4: 77713434-77713434 on Build GRCh38
rs548915412 Chr.4: 77713724-77713724 on Build GRCh38
rs549818175 Chr.4: 77713548-77713548 on Build GRCh38
rs946547835 Chr.4: 77713515-77713515 on Build GRCh38
rs980714839 Chr.4: 77713581-77713581 on Build GRCh38
rs996715190 Chr.4: 77713684-77713684 on Build GRCh38
rs931229306 Chr.4: 77713621-77713621 on Build GRCh38
rs886430767 Chr.4: 77713657-77713657 on Build GRCh38
rs1029153347 Chr.4: 77713732-77713732 on Build GRCh38
rs774912685 Chr.4: 77713550-77713550 on Build GRCh38
rs996460807 Chr.4: 77713322-77713322 on Build GRCh38
rs979125224 Chr.4: 77713385-77713385 on Build GRCh38
rs145157794 Chr.4: 77713400-77713400 on Build GRCh38
rs1038807423 Chr.4: 77713435-77713436 on Build GRCh38
rs911594387 Chr.4: 77713344-77713344 on Build GRCh38
rs967715535 Chr.4: 77713360-77713360 on Build GRCh38
rs1025816111 Chr.4: 77713757-77713758 on Build GRCh38
rs956466571 Chr.4: 77713439-77713439 on Build GRCh38
rs962827694 Chr.4: 77713736-77713736 on Build GRCh38

View Full Product Details