Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107067301-107067820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND3
Entrez Gene ID
57673
Gene Name
BEN domain containing 3
Gene Aliases
KIAA1553
UniGene
Hs.418045
Gene Chromosome Location
Chr.6: 107065180-107115187 on Build GRCh38
Transcripts
NM_001080450.2 XM_005267079.3 XM_005267080.3 XM_011536005.2 XM_017011138.1

SNP Detail

SNP ID Chromosome Location
rs924283287 Chr.6: 107067493-107067493 on Build GRCh38
rs1027588108 Chr.6: 107067407-107067407 on Build GRCh38
rs73763707 Chr.6: 107067739-107067739 on Build GRCh38
rs574246721 Chr.6: 107067461-107067461 on Build GRCh38
rs965816606 Chr.6: 107067471-107067471 on Build GRCh38
rs541406128 Chr.6: 107067615-107067615 on Build GRCh38
rs183165166 Chr.6: 107067565-107067565 on Build GRCh38
rs1003919552 Chr.6: 107067360-107067360 on Build GRCh38
rs985967599 Chr.6: 107067439-107067439 on Build GRCh38
rs1018786937 Chr.6: 107067751-107067751 on Build GRCh38
rs538108129 Chr.6: 107067395-107067395 on Build GRCh38
rs782801727 Chr.6: 107067574-107067574 on Build GRCh38
rs1014916353 Chr.6: 107067388-107067388 on Build GRCh38
rs115612104 Chr.6: 107067472-107067472 on Build GRCh38
rs782809630 Chr.6: 107067368-107067368 on Build GRCh38
rs906629807 Chr.6: 107067357-107067357 on Build GRCh38
rs150002280 Chr.6: 107067581-107067581 on Build GRCh38
rs576573600 Chr.6: 107067518-107067518 on Build GRCh38
rs966296305 Chr.6: 107067773-107067773 on Build GRCh38
rs540047124 Chr.6: 107067462-107067462 on Build GRCh38
rs995267065 Chr.6: 107067639-107067639 on Build GRCh38
rs939447082 Chr.6: 107067616-107067616 on Build GRCh38
rs782501475 Chr.6: 107067526-107067526 on Build GRCh38
rs114085529 Chr.6: 107067332-107067332 on Build GRCh38
rs55817462 Chr.6: 107067392-107067392 on Build GRCh38
rs531232299 Chr.6: 107067609-107067609 on Build GRCh38
rs948184012 Chr.6: 107067576-107067576 on Build GRCh38
rs935694867 Chr.6: 107067505-107067505 on Build GRCh38
rs545457849 Chr.6: 107067580-107067580 on Build GRCh38
rs188110314 Chr.6: 107067647-107067647 on Build GRCh38
rs897810396 Chr.6: 107067636-107067636 on Build GRCh38
rs1036877067 Chr.6: 107067619-107067619 on Build GRCh38
rs78361125 Chr.6: 107067419-107067419 on Build GRCh38
rs35740762 Chr.6: 107067607-107067608 on Build GRCh38
rs994744828 Chr.6: 107067397-107067397 on Build GRCh38
rs782432716 Chr.6: 107067661-107067661 on Build GRCh38
rs781842044 Chr.6: 107067482-107067482 on Build GRCh38
rs889047645 Chr.6: 107067727-107067727 on Build GRCh38

View Full Product Details