Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11423255-11423751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167A
Entrez Gene ID
83648
Gene Name
family with sequence similarity 167 member A
Gene Aliases
C8orf13, D8S265
UniGene
Hs.124299
Gene Chromosome Location
Chr.8: 11421464-11475904 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Gene Symbol
FAM167A-AS1
Entrez Gene ID
83656
Gene Name
FAM167A antisense RNA 1
Gene Aliases
C8orf12
UniGene
Hs.583896
Gene Chromosome Location
Chr.8: 11368402-11438657 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

SNP Detail

SNP ID Chromosome Location
rs906880206 Chr.8: 11423332-11423332 on Build GRCh38
rs1022918884 Chr.8: 11423727-11423727 on Build GRCh38
rs924699766 Chr.8: 11423650-11423650 on Build GRCh38
rs540523402 Chr.8: 11423501-11423501 on Build GRCh38
rs1047874119 Chr.8: 11423707-11423707 on Build GRCh38
rs370530900 Chr.8: 11423684-11423684 on Build GRCh38
rs887562145 Chr.8: 11423525-11423525 on Build GRCh38
rs935838193 Chr.8: 11423302-11423302 on Build GRCh38
rs887523447 Chr.8: 11423716-11423716 on Build GRCh38
rs138656309 Chr.8: 11423311-11423311 on Build GRCh38
rs1011968735 Chr.8: 11423540-11423542 on Build GRCh38
rs544250662 Chr.8: 11423574-11423574 on Build GRCh38
rs113102639 Chr.8: 11423617-11423617 on Build GRCh38
rs1017183450 Chr.8: 11423380-11423380 on Build GRCh38
rs889354982 Chr.8: 11423442-11423442 on Build GRCh38
rs961159451 Chr.8: 11423588-11423588 on Build GRCh38
rs552593188 Chr.8: 11423291-11423291 on Build GRCh38
rs948589551 Chr.8: 11423463-11423463 on Build GRCh38
rs889298229 Chr.8: 11423294-11423294 on Build GRCh38
rs542601808 Chr.8: 11423713-11423715 on Build GRCh38
rs1012567840 Chr.8: 11423313-11423313 on Build GRCh38
rs192810285 Chr.8: 11423535-11423535 on Build GRCh38
rs551609103 Chr.8: 11423706-11423706 on Build GRCh38
rs756986362 Chr.8: 11423469-11423469 on Build GRCh38
rs767124984 Chr.8: 11423499-11423499 on Build GRCh38
rs868659255 Chr.8: 11423718-11423718 on Build GRCh38
rs931016505 Chr.8: 11423639-11423639 on Build GRCh38
rs951543985 Chr.8: 11423361-11423361 on Build GRCh38
rs756551730 Chr.8: 11423606-11423606 on Build GRCh38
rs1024003714 Chr.8: 11423323-11423323 on Build GRCh38
rs144841188 Chr.8: 11423280-11423280 on Build GRCh38
rs957714342 Chr.8: 11423652-11423652 on Build GRCh38
rs901969721 Chr.8: 11423476-11423476 on Build GRCh38
rs902020541 Chr.8: 11423682-11423682 on Build GRCh38
rs182541253 Chr.8: 11423544-11423544 on Build GRCh38
rs573220448 Chr.8: 11423485-11423485 on Build GRCh38
rs997033131 Chr.8: 11423552-11423552 on Build GRCh38
rs1040610578 Chr.8: 11423663-11423663 on Build GRCh38
rs915815736 Chr.8: 11423444-11423444 on Build GRCh38
rs966107102 Chr.8: 11423308-11423308 on Build GRCh38
rs546054648 Chr.8: 11423386-11423386 on Build GRCh38
rs1029874939 Chr.8: 11423600-11423600 on Build GRCh38
rs922578769 Chr.8: 11423397-11423397 on Build GRCh38
rs755069469 Chr.8: 11423518-11423518 on Build GRCh38
rs902995158 Chr.8: 11423293-11423293 on Build GRCh38
rs910364576 Chr.8: 11423427-11423427 on Build GRCh38
rs149343752 Chr.8: 11423312-11423312 on Build GRCh38
rs367774419 Chr.8: 11423439-11423439 on Build GRCh38
rs990091063 Chr.8: 11423336-11423336 on Build GRCh38
rs532802583 Chr.8: 11423391-11423391 on Build GRCh38
rs754374644 Chr.8: 11423438-11423438 on Build GRCh38
rs1007706756 Chr.8: 11423296-11423296 on Build GRCh38
rs1045643237 Chr.8: 11423694-11423694 on Build GRCh38
rs867457138 Chr.8: 11423483-11423483 on Build GRCh38
rs910843746 Chr.8: 11423653-11423653 on Build GRCh38
rs866661663 Chr.8: 11423683-11423683 on Build GRCh38
rs534220898 Chr.8: 11423288-11423288 on Build GRCh38
rs187798729 Chr.8: 11423342-11423342 on Build GRCh38
rs3021511 Chr.8: 11423376-11423376 on Build GRCh38
rs1019065041 Chr.8: 11423305-11423305 on Build GRCh38
rs990261232 Chr.8: 11423656-11423656 on Build GRCh38
rs977861910 Chr.8: 11423619-11423619 on Build GRCh38
rs1031669925 Chr.8: 11423310-11423310 on Build GRCh38
rs866635379 Chr.8: 11423295-11423295 on Build GRCh38
rs943601077 Chr.8: 11423662-11423662 on Build GRCh38
rs998891818 Chr.8: 11423309-11423309 on Build GRCh38
rs915723859 Chr.8: 11423676-11423676 on Build GRCh38
rs1045695298 Chr.8: 11423475-11423475 on Build GRCh38
rs981278728 Chr.8: 11423555-11423555 on Build GRCh38
rs964195175 Chr.8: 11423538-11423538 on Build GRCh38
rs1022910098 Chr.8: 11423541-11423541 on Build GRCh38
rs561793124 Chr.8: 11423526-11423526 on Build GRCh38
rs1040557438 Chr.8: 11423466-11423466 on Build GRCh38
rs924700239 Chr.8: 11423416-11423416 on Build GRCh38
rs751187068 Chr.8: 11423381-11423381 on Build GRCh38
rs552292728 Chr.8: 11423399-11423399 on Build GRCh38
rs977497235 Chr.8: 11423411-11423411 on Build GRCh38
rs975373267 Chr.8: 11423395-11423395 on Build GRCh38
rs1026485066 Chr.8: 11423347-11423347 on Build GRCh38
rs533234851 Chr.8: 11423705-11423705 on Build GRCh38
rs557961512 Chr.8: 11423408-11423408 on Build GRCh38
rs919643914 Chr.8: 11423616-11423616 on Build GRCh38
rs955393956 Chr.8: 11423401-11423401 on Build GRCh38

View Full Product Details