Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17086624-17087140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1 XM_017013343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5108269 c.796delA p.N268fs*47 Chr.8: 17086982-17086982 on Build GRCh38
4560075 c.829G>A p.E277K Chr.8: 17087015-17087015 on Build GRCh38
4840459 c.835A>G p.K279E Chr.8: 17087021-17087021 on Build GRCh38
6307904 c.836A>T p.K279M Chr.8: 17087022-17087022 on Build GRCh38
3834345 c.839G>T p.R280L Chr.8: 17087025-17087025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578221124 Chr.8: 17086996-17086996 on Build GRCh38
rs953522603 Chr.8: 17086744-17086744 on Build GRCh38
rs374232012 Chr.8: 17086951-17086951 on Build GRCh38
rs144959956 Chr.8: 17087030-17087030 on Build GRCh38
rs187173663 Chr.8: 17087109-17087109 on Build GRCh38
rs558181366 Chr.8: 17086933-17086933 on Build GRCh38
rs778472385 Chr.8: 17086909-17086909 on Build GRCh38
rs770801887 Chr.8: 17086742-17086742 on Build GRCh38
rs933716212 Chr.8: 17086670-17086670 on Build GRCh38
rs199759760 Chr.8: 17086982-17086982 on Build GRCh38
rs368379761 Chr.8: 17087055-17087055 on Build GRCh38
rs113805370 Chr.8: 17086820-17086820 on Build GRCh38
rs1003609429 Chr.8: 17086990-17086990 on Build GRCh38
rs867237206 Chr.8: 17087034-17087034 on Build GRCh38
rs763620157 Chr.8: 17087081-17087081 on Build GRCh38
rs918279279 Chr.8: 17086891-17086891 on Build GRCh38
rs947835957 Chr.8: 17086930-17086930 on Build GRCh38
rs761019871 Chr.8: 17086934-17086934 on Build GRCh38
rs73666270 Chr.8: 17086655-17086655 on Build GRCh38
rs554387006 Chr.8: 17087068-17087068 on Build GRCh38
rs770324407 Chr.8: 17087070-17087070 on Build GRCh38
rs1048320115 Chr.8: 17086911-17086911 on Build GRCh38
rs773839721 Chr.8: 17087071-17087071 on Build GRCh38
rs943815956 Chr.8: 17086870-17086870 on Build GRCh38
rs780177617 Chr.8: 17087048-17087048 on Build GRCh38
rs752287110 Chr.8: 17087015-17087015 on Build GRCh38
rs377656354 Chr.8: 17087085-17087085 on Build GRCh38
rs112253967 Chr.8: 17086975-17086975 on Build GRCh38
rs773741785 Chr.8: 17086992-17086992 on Build GRCh38
rs370647732 Chr.8: 17087025-17087025 on Build GRCh38
rs372387819 Chr.8: 17087062-17087062 on Build GRCh38
rs1000042305 Chr.8: 17087028-17087028 on Build GRCh38
rs372712442 Chr.8: 17087045-17087045 on Build GRCh38
rs745976675 Chr.8: 17086919-17086919 on Build GRCh38
rs373176297 Chr.8: 17087110-17087110 on Build GRCh38
rs62502369 Chr.8: 17087100-17087100 on Build GRCh38
rs796544195 Chr.8: 17086799-17086799 on Build GRCh38
rs912278531 Chr.8: 17086834-17086834 on Build GRCh38
rs751018914 Chr.8: 17087024-17087024 on Build GRCh38
rs908352661 Chr.8: 17086749-17086749 on Build GRCh38
rs1046694910 Chr.8: 17087001-17087001 on Build GRCh38
rs115795420 Chr.8: 17086896-17086896 on Build GRCh38
rs568872415 Chr.8: 17086794-17086794 on Build GRCh38
rs1029211891 Chr.8: 17086669-17086669 on Build GRCh38
rs199676059 Chr.8: 17086920-17086920 on Build GRCh38
rs113897407 Chr.8: 17087041-17087041 on Build GRCh38
rs898455299 Chr.8: 17086829-17086829 on Build GRCh38
rs762169541 Chr.8: 17086982-17086982 on Build GRCh38
rs968101296 Chr.8: 17086649-17086649 on Build GRCh38
rs778912281 Chr.8: 17086915-17086915 on Build GRCh38
rs777059323 Chr.8: 17086976-17086976 on Build GRCh38
rs770910666 Chr.8: 17086914-17086914 on Build GRCh38
rs542717464 Chr.8: 17087098-17087098 on Build GRCh38
rs747138536 Chr.8: 17087053-17087053 on Build GRCh38
rs199936197 Chr.8: 17087027-17087027 on Build GRCh38
rs772182770 Chr.8: 17086922-17086922 on Build GRCh38
rs972868639 Chr.8: 17087104-17087104 on Build GRCh38
rs751076440 Chr.8: 17086984-17086984 on Build GRCh38
rs141246387 Chr.8: 17086983-17086983 on Build GRCh38
rs745857178 Chr.8: 17087036-17087036 on Build GRCh38
rs573759344 Chr.8: 17087080-17087080 on Build GRCh38
rs767192572 Chr.8: 17087011-17087011 on Build GRCh38
rs774456316 Chr.8: 17086859-17086859 on Build GRCh38
rs972422316 Chr.8: 17086881-17086881 on Build GRCh38
rs145792572 Chr.8: 17086756-17086756 on Build GRCh38
rs768299402 Chr.8: 17086991-17086995 on Build GRCh38
rs768902648 Chr.8: 17086969-17086969 on Build GRCh38

View Full Product Details