Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27459986-27460493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

SNP Detail

SNP ID Chromosome Location
rs376282591 Chr.8: 27460226-27460226 on Build GRCh38
rs907677233 Chr.8: 27460134-27460134 on Build GRCh38
rs907644861 Chr.8: 27460351-27460351 on Build GRCh38
rs369380309 Chr.8: 27460388-27460388 on Build GRCh38
rs963168961 Chr.8: 27460362-27460362 on Build GRCh38
rs535057169 Chr.8: 27460026-27460026 on Build GRCh38
rs530957765 Chr.8: 27460295-27460295 on Build GRCh38
rs917654803 Chr.8: 27460109-27460109 on Build GRCh38
rs545789115 Chr.8: 27460070-27460070 on Build GRCh38
rs149731616 Chr.8: 27460405-27460405 on Build GRCh38
rs575088206 Chr.8: 27460052-27460053 on Build GRCh38
rs1010290683 Chr.8: 27460229-27460229 on Build GRCh38
rs970488322 Chr.8: 27460128-27460128 on Build GRCh38
rs959235347 Chr.8: 27460268-27460268 on Build GRCh38
rs977095543 Chr.8: 27460323-27460323 on Build GRCh38
rs959039907 Chr.8: 27460049-27460049 on Build GRCh38
rs1055850626 Chr.8: 27460155-27460155 on Build GRCh38
rs886062841 Chr.8: 27460234-27460234 on Build GRCh38
rs1035031878 Chr.8: 27460028-27460028 on Build GRCh38
rs2280376 Chr.8: 27460245-27460245 on Build GRCh38
rs1037596781 Chr.8: 27460164-27460164 on Build GRCh38
rs1000782647 Chr.8: 27460255-27460255 on Build GRCh38
rs866036899 Chr.8: 27460124-27460124 on Build GRCh38
rs936223018 Chr.8: 27460328-27460328 on Build GRCh38
rs569707665 Chr.8: 27460302-27460302 on Build GRCh38
rs970372160 Chr.8: 27460311-27460311 on Build GRCh38
rs1051692583 Chr.8: 27460406-27460406 on Build GRCh38
rs28697263 Chr.8: 27460183-27460183 on Build GRCh38
rs113471592 Chr.8: 27460149-27460149 on Build GRCh38
rs199764226 Chr.8: 27460343-27460343 on Build GRCh38
rs1046259890 Chr.8: 27460218-27460218 on Build GRCh38
rs1000413072 Chr.8: 27460027-27460027 on Build GRCh38
rs561812612 Chr.8: 27460175-27460175 on Build GRCh38
rs1034592541 Chr.8: 27460264-27460264 on Build GRCh38
rs1560344 Chr.8: 27460470-27460470 on Build GRCh38
rs1018076614 Chr.8: 27460271-27460271 on Build GRCh38
rs911061640 Chr.8: 27460034-27460034 on Build GRCh38
rs556174072 Chr.8: 27460093-27460093 on Build GRCh38
rs576142259 Chr.8: 27460133-27460133 on Build GRCh38
rs983141566 Chr.8: 27460319-27460319 on Build GRCh38
rs1055991241 Chr.8: 27460335-27460335 on Build GRCh38
rs752765192 Chr.8: 27460280-27460280 on Build GRCh38
rs990919493 Chr.8: 27460104-27460104 on Build GRCh38
rs1014317597 Chr.8: 27460197-27460197 on Build GRCh38
rs911855421 Chr.8: 27460469-27460469 on Build GRCh38
rs931984634 Chr.8: 27460172-27460172 on Build GRCh38
rs564765220 Chr.8: 27460237-27460237 on Build GRCh38
rs973667975 Chr.8: 27460364-27460364 on Build GRCh38
rs1041938876 Chr.8: 27460095-27460095 on Build GRCh38
rs900098264 Chr.8: 27460346-27460346 on Build GRCh38
rs897813564 Chr.8: 27460224-27460224 on Build GRCh38
rs925623920 Chr.8: 27460324-27460324 on Build GRCh38
rs922267676 Chr.8: 27460091-27460091 on Build GRCh38
rs866927631 Chr.8: 27460385-27460385 on Build GRCh38
rs921874446 Chr.8: 27460165-27460165 on Build GRCh38
rs976919710 Chr.8: 27460067-27460067 on Build GRCh38
rs922371929 Chr.8: 27460313-27460313 on Build GRCh38

View Full Product Details