Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27469244-27469736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436455 c.324C>T p.N108N Chr.8: 27469350-27469350 on Build GRCh38
4600299 c.320C>G p.T107S Chr.8: 27469354-27469354 on Build GRCh38
4453506 c.304A>C p.N102H Chr.8: 27469370-27469370 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139680977 Chr.8: 27469322-27469323 on Build GRCh38
rs982512969 Chr.8: 27469619-27469619 on Build GRCh38
rs766272313 Chr.8: 27469470-27469470 on Build GRCh38
rs375118890 Chr.8: 27469574-27469574 on Build GRCh38
rs1026955886 Chr.8: 27469541-27469541 on Build GRCh38
rs116459673 Chr.8: 27469661-27469661 on Build GRCh38
rs995433802 Chr.8: 27469526-27469526 on Build GRCh38
rs537861239 Chr.8: 27469640-27469640 on Build GRCh38
rs1002474650 Chr.8: 27469441-27469441 on Build GRCh38
rs913563840 Chr.8: 27469352-27469352 on Build GRCh38
rs1050980684 Chr.8: 27469669-27469669 on Build GRCh38
rs944982830 Chr.8: 27469384-27469384 on Build GRCh38
rs749083602 Chr.8: 27469705-27469705 on Build GRCh38
rs909706680 Chr.8: 27469457-27469457 on Build GRCh38
rs1046858418 Chr.8: 27469420-27469420 on Build GRCh38
rs746630071 Chr.8: 27469325-27469325 on Build GRCh38
rs547236414 Chr.8: 27469558-27469558 on Build GRCh38
rs530277867 Chr.8: 27469499-27469499 on Build GRCh38
rs374208928 Chr.8: 27469711-27469711 on Build GRCh38
rs550601339 Chr.8: 27469465-27469465 on Build GRCh38
rs1033858902 Chr.8: 27469568-27469568 on Build GRCh38
rs371304320 Chr.8: 27469665-27469665 on Build GRCh38
rs910397272 Chr.8: 27469312-27469312 on Build GRCh38
rs552736727 Chr.8: 27469664-27469664 on Build GRCh38
rs776047724 Chr.8: 27469385-27469386 on Build GRCh38
rs190255317 Chr.8: 27469706-27469706 on Build GRCh38
rs995570511 Chr.8: 27469702-27469702 on Build GRCh38
rs748560508 Chr.8: 27469336-27469336 on Build GRCh38
rs200926431 Chr.8: 27469296-27469296 on Build GRCh38
rs1009578556 Chr.8: 27469548-27469548 on Build GRCh38
rs560827547 Chr.8: 27469482-27469482 on Build GRCh38
rs552944382 Chr.8: 27469607-27469607 on Build GRCh38
rs140159249 Chr.8: 27469343-27469343 on Build GRCh38
rs564532686 Chr.8: 27469351-27469351 on Build GRCh38
rs770464377 Chr.8: 27469321-27469322 on Build GRCh38
rs985900885 Chr.8: 27469283-27469283 on Build GRCh38
rs747401096 Chr.8: 27469390-27469390 on Build GRCh38
rs886044444 Chr.8: 27469353-27469353 on Build GRCh38
rs367739392 Chr.8: 27469586-27469586 on Build GRCh38
rs998122801 Chr.8: 27469495-27469495 on Build GRCh38
rs992440390 Chr.8: 27469604-27469604 on Build GRCh38
rs544902258 Chr.8: 27469689-27469689 on Build GRCh38
rs768490793 Chr.8: 27469327-27469327 on Build GRCh38
rs749863419 Chr.8: 27469297-27469297 on Build GRCh38
rs774451576 Chr.8: 27469418-27469418 on Build GRCh38
rs757951570 Chr.8: 27469303-27469303 on Build GRCh38
rs796052293 Chr.8: 27469326-27469327 on Build GRCh38
rs1034654767 Chr.8: 27469439-27469439 on Build GRCh38
rs796052295 Chr.8: 27469373-27469373 on Build GRCh38
rs975942177 Chr.8: 27469618-27469618 on Build GRCh38
rs369707163 Chr.8: 27469391-27469391 on Build GRCh38
rs147191290 Chr.8: 27469612-27469612 on Build GRCh38
rs770131239 Chr.8: 27469341-27469341 on Build GRCh38
rs544939077 Chr.8: 27469350-27469350 on Build GRCh38
rs185419685 Chr.8: 27469645-27469645 on Build GRCh38
rs567096949 Chr.8: 27469563-27469563 on Build GRCh38
rs745538264 Chr.8: 27469549-27469549 on Build GRCh38
rs926906905 Chr.8: 27469288-27469288 on Build GRCh38
rs796052296 Chr.8: 27469367-27469367 on Build GRCh38
rs745364666 Chr.8: 27469387-27469387 on Build GRCh38
rs757075915 Chr.8: 27469294-27469294 on Build GRCh38
rs892447964 Chr.8: 27469677-27469677 on Build GRCh38
rs892485601 Chr.8: 27469529-27469529 on Build GRCh38
rs568484025 Chr.8: 27469648-27469648 on Build GRCh38
rs899783588 Chr.8: 27469494-27469494 on Build GRCh38
rs916888102 Chr.8: 27469605-27469605 on Build GRCh38
rs756788513 Chr.8: 27469282-27469282 on Build GRCh38
rs896674589 Chr.8: 27469484-27469484 on Build GRCh38
rs1002818189 Chr.8: 27469564-27469564 on Build GRCh38
rs945418539 Chr.8: 27469681-27469681 on Build GRCh38
rs779763586 Chr.8: 27469304-27469304 on Build GRCh38

View Full Product Details