Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27470908-27471419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5543800 c.22T>C p.F8L Chr.8: 27471037-27471037 on Build GRCh38
310100 c.11C>A p.S4Y Chr.8: 27471048-27471048 on Build GRCh38
5596499 c.57C>T p.L19L Chr.8: 27471002-27471002 on Build GRCh38
7230454 c.65C>T p.T22I Chr.8: 27470994-27470994 on Build GRCh38
5942192 c.16C>T p.P6S Chr.8: 27471043-27471043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753445848 Chr.8: 27471231-27471231 on Build GRCh38
rs1039353056 Chr.8: 27471300-27471300 on Build GRCh38
rs61507083 Chr.8: 27471383-27471383 on Build GRCh38
rs372291035 Chr.8: 27471266-27471266 on Build GRCh38
rs886062847 Chr.8: 27471208-27471208 on Build GRCh38
rs768151492 Chr.8: 27471008-27471008 on Build GRCh38
rs772876723 Chr.8: 27470949-27470949 on Build GRCh38
rs1019044128 Chr.8: 27471322-27471322 on Build GRCh38
rs116683540 Chr.8: 27471287-27471287 on Build GRCh38
rs1004789936 Chr.8: 27471244-27471244 on Build GRCh38
rs772665352 Chr.8: 27471097-27471097 on Build GRCh38
rs923597649 Chr.8: 27471172-27471172 on Build GRCh38
rs764266472 Chr.8: 27471107-27471107 on Build GRCh38
rs967573472 Chr.8: 27471185-27471185 on Build GRCh38
rs916351748 Chr.8: 27471208-27471208 on Build GRCh38
rs2472553 Chr.8: 27470994-27470994 on Build GRCh38
rs764360561 Chr.8: 27471030-27471030 on Build GRCh38
rs867626422 Chr.8: 27471189-27471189 on Build GRCh38
rs947981612 Chr.8: 27470942-27470942 on Build GRCh38
rs1029818667 Chr.8: 27471169-27471169 on Build GRCh38
rs774842184 Chr.8: 27470983-27470983 on Build GRCh38
rs748886161 Chr.8: 27471075-27471075 on Build GRCh38
rs538048618 Chr.8: 27471040-27471040 on Build GRCh38
rs754810220 Chr.8: 27471044-27471044 on Build GRCh38
rs149934219 Chr.8: 27471014-27471014 on Build GRCh38
rs752046709 Chr.8: 27471151-27471151 on Build GRCh38
rs549463792 Chr.8: 27471123-27471123 on Build GRCh38
rs774517643 Chr.8: 27471086-27471086 on Build GRCh38
rs769373551 Chr.8: 27470979-27470982 on Build GRCh38
rs201177648 Chr.8: 27470972-27470972 on Build GRCh38
rs769273921 Chr.8: 27471065-27471065 on Build GRCh38
rs761883301 Chr.8: 27471108-27471108 on Build GRCh38
rs886062845 Chr.8: 27471136-27471136 on Build GRCh38
rs1005141006 Chr.8: 27471117-27471117 on Build GRCh38
rs747665660 Chr.8: 27470944-27470944 on Build GRCh38
rs757567429 Chr.8: 27471035-27471035 on Build GRCh38
rs759948980 Chr.8: 27471096-27471096 on Build GRCh38
rs776149538 Chr.8: 27470996-27471005 on Build GRCh38
rs987563818 Chr.8: 27471120-27471120 on Build GRCh38
rs967580541 Chr.8: 27471138-27471138 on Build GRCh38
rs906408607 Chr.8: 27470956-27470956 on Build GRCh38
rs1024493056 Chr.8: 27471137-27471157 on Build GRCh38
rs765846499 Chr.8: 27471197-27471197 on Build GRCh38
rs753978091 Chr.8: 27471031-27471031 on Build GRCh38
rs770174446 Chr.8: 27471153-27471153 on Build GRCh38
rs912066664 Chr.8: 27471136-27471136 on Build GRCh38
rs998993050 Chr.8: 27471398-27471398 on Build GRCh38
rs553788971 Chr.8: 27471123-27471123 on Build GRCh38
rs12114756 Chr.8: 27471103-27471103 on Build GRCh38
rs777302995 Chr.8: 27471071-27471071 on Build GRCh38
rs967550486 Chr.8: 27471327-27471327 on Build GRCh38
rs1008957047 Chr.8: 27471305-27471305 on Build GRCh38
rs375740892 Chr.8: 27471052-27471052 on Build GRCh38
rs886062846 Chr.8: 27471191-27471191 on Build GRCh38
rs936357844 Chr.8: 27471173-27471173 on Build GRCh38
rs747612221 Chr.8: 27471064-27471064 on Build GRCh38
rs762713890 Chr.8: 27470964-27470964 on Build GRCh38
rs776066363 Chr.8: 27470936-27470936 on Build GRCh38
rs947838271 Chr.8: 27471213-27471213 on Build GRCh38
rs755685493 Chr.8: 27471190-27471190 on Build GRCh38
rs763333246 Chr.8: 27471050-27471050 on Build GRCh38
rs982470004 Chr.8: 27471261-27471261 on Build GRCh38
rs534111778 Chr.8: 27471112-27471112 on Build GRCh38
rs1033057388 Chr.8: 27471191-27471191 on Build GRCh38
rs770671362 Chr.8: 27471077-27471077 on Build GRCh38
rs1029001075 Chr.8: 27471294-27471294 on Build GRCh38
rs542421071 Chr.8: 27471387-27471387 on Build GRCh38
rs182483939 Chr.8: 27471365-27471365 on Build GRCh38
rs1036302797 Chr.8: 27471260-27471260 on Build GRCh38
rs746835048 Chr.8: 27471043-27471043 on Build GRCh38
rs369385134 Chr.8: 27470970-27470970 on Build GRCh38
rs560688759 Chr.8: 27470965-27470965 on Build GRCh38
rs906377558 Chr.8: 27471241-27471241 on Build GRCh38
rs55954528 Chr.8: 27471271-27471271 on Build GRCh38
rs1029528073 Chr.8: 27471085-27471085 on Build GRCh38
rs953549932 Chr.8: 27471295-27471295 on Build GRCh38
rs769400359 Chr.8: 27470945-27470945 on Build GRCh38
rs372197535 Chr.8: 27471388-27471388 on Build GRCh38
rs963737055 Chr.8: 27471051-27471051 on Build GRCh38
rs568386814 Chr.8: 27471104-27471104 on Build GRCh38
rs1046239250 Chr.8: 27471227-27471227 on Build GRCh38

View Full Product Details