Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133397735-133398256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STKLD1
Entrez Gene ID
169436
Gene Name
serine/threonine kinase like domain containing 1
Gene Aliases
C9orf96, SgK071, Sk521
UniGene
Hs.720195
Gene Chromosome Location
Chr.9: 133376413-133406096 on Build GRCh38
Transcripts
NM_153710.4 NR_103997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129231 c.1057C>T p.L353L Chr.9: 133398031-133398031 on Build GRCh38
1598137 c.1072G>T p.D358Y Chr.9: 133398046-133398046 on Build GRCh38
3213424 c.1003A>G p.M335V Chr.9: 133397977-133397977 on Build GRCh38
3847968 c.1050G>C p.K350N Chr.9: 133398024-133398024 on Build GRCh38
3655266 c.1081G>A p.G361S Chr.9: 133398055-133398055 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371132192 Chr.9: 133398114-133398114 on Build GRCh38
rs587737205 Chr.9: 133397930-133397930 on Build GRCh38
rs587675401 Chr.9: 133397776-133397776 on Build GRCh38
rs1012821026 Chr.9: 133397871-133397871 on Build GRCh38
rs781978865 Chr.9: 133397944-133397944 on Build GRCh38
rs921417568 Chr.9: 133397762-133397762 on Build GRCh38
rs369488383 Chr.9: 133397865-133397865 on Build GRCh38
rs1056193634 Chr.9: 133398185-133398185 on Build GRCh38
rs782002663 Chr.9: 133398076-133398076 on Build GRCh38
rs782587127 Chr.9: 133398057-133398057 on Build GRCh38
rs782530208 Chr.9: 133397780-133397780 on Build GRCh38
rs749892606 Chr.9: 133397906-133397906 on Build GRCh38
rs781911176 Chr.9: 133398105-133398105 on Build GRCh38
rs756738886 Chr.9: 133398059-133398059 on Build GRCh38
rs782741611 Chr.9: 133397990-133397993 on Build GRCh38
rs782582708 Chr.9: 133397933-133397933 on Build GRCh38
rs587715648 Chr.9: 133398147-133398147 on Build GRCh38
rs934296177 Chr.9: 133397841-133397841 on Build GRCh38
rs587661304 Chr.9: 133397824-133397824 on Build GRCh38
rs782799576 Chr.9: 133398100-133398100 on Build GRCh38
rs199573986 Chr.9: 133397926-133397926 on Build GRCh38
rs782306438 Chr.9: 133398061-133398061 on Build GRCh38
rs587638358 Chr.9: 133398115-133398115 on Build GRCh38
rs782148007 Chr.9: 133398073-133398073 on Build GRCh38
rs188153526 Chr.9: 133398186-133398186 on Build GRCh38
rs782107197 Chr.9: 133398068-133398069 on Build GRCh38
rs975636407 Chr.9: 133398094-133398094 on Build GRCh38
rs909103925 Chr.9: 133397849-133397849 on Build GRCh38
rs587653469 Chr.9: 133398231-133398231 on Build GRCh38
rs782573376 Chr.9: 133398015-133398015 on Build GRCh38
rs587758933 Chr.9: 133398020-133398020 on Build GRCh38
rs80050299 Chr.9: 133397850-133397850 on Build GRCh38
rs587648831 Chr.9: 133398177-133398177 on Build GRCh38
rs782696656 Chr.9: 133398049-133398049 on Build GRCh38
rs930439966 Chr.9: 133398204-133398204 on Build GRCh38
rs893369308 Chr.9: 133397950-133397950 on Build GRCh38
rs781825758 Chr.9: 133398045-133398046 on Build GRCh38
rs587606641 Chr.9: 133398211-133398211 on Build GRCh38
rs934411302 Chr.9: 133398158-133398158 on Build GRCh38
rs782386671 Chr.9: 133398056-133398056 on Build GRCh38
rs4962047 Chr.9: 133397789-133397789 on Build GRCh38
rs587597896 Chr.9: 133397765-133397765 on Build GRCh38
rs919065578 Chr.9: 133398137-133398137 on Build GRCh38
rs147959001 Chr.9: 133398001-133398001 on Build GRCh38
rs1041920615 Chr.9: 133398217-133398218 on Build GRCh38
rs781883892 Chr.9: 133397978-133397978 on Build GRCh38
rs780698304 Chr.9: 133398087-133398087 on Build GRCh38
rs144772739 Chr.9: 133397981-133397981 on Build GRCh38
rs782053086 Chr.9: 133397949-133397949 on Build GRCh38
rs782424706 Chr.9: 133397943-133397943 on Build GRCh38
rs201703214 Chr.9: 133397964-133397964 on Build GRCh38
rs782414662 Chr.9: 133398074-133398074 on Build GRCh38
rs782189457 Chr.9: 133398122-133398122 on Build GRCh38
rs782248502 Chr.9: 133398055-133398055 on Build GRCh38
rs781813794 Chr.9: 133398033-133398033 on Build GRCh38
rs372810649 Chr.9: 133398093-133398093 on Build GRCh38
rs587654154 Chr.9: 133397942-133397942 on Build GRCh38
rs930082189 Chr.9: 133398145-133398145 on Build GRCh38
rs369889107 Chr.9: 133398092-133398092 on Build GRCh38
rs1030428150 Chr.9: 133398026-133398026 on Build GRCh38
rs1056810634 Chr.9: 133397843-133397843 on Build GRCh38
rs587622019 Chr.9: 133397864-133397864 on Build GRCh38
rs782292165 Chr.9: 133397907-133397907 on Build GRCh38
rs782235983 Chr.9: 133397928-133397928 on Build GRCh38
rs181352871 Chr.9: 133398201-133398201 on Build GRCh38
rs185352818 Chr.9: 133398160-133398160 on Build GRCh38
rs782770338 Chr.9: 133397992-133397992 on Build GRCh38
rs587707438 Chr.9: 133397952-133397952 on Build GRCh38
rs965784980 Chr.9: 133398024-133398024 on Build GRCh38
rs1004592776 Chr.9: 133397973-133397973 on Build GRCh38
rs782048842 Chr.9: 133397974-133397974 on Build GRCh38
rs930029809 Chr.9: 133397897-133397897 on Build GRCh38
rs924325970 Chr.9: 133398130-133398130 on Build GRCh38
rs782543232 Chr.9: 133398042-133398042 on Build GRCh38
rs368025940 Chr.9: 133397977-133397977 on Build GRCh38
rs781933023 Chr.9: 133398068-133398068 on Build GRCh38
rs782367513 Chr.9: 133398159-133398159 on Build GRCh38
rs375521277 Chr.9: 133397951-133397951 on Build GRCh38
rs951515857 Chr.9: 133397927-133397927 on Build GRCh38
rs587706092 Chr.9: 133397784-133397784 on Build GRCh38
rs994680225 Chr.9: 133397922-133397922 on Build GRCh38
rs782081902 Chr.9: 133398082-133398082 on Build GRCh38
rs782522978 Chr.9: 133397921-133397921 on Build GRCh38
rs754193271 Chr.9: 133398163-133398163 on Build GRCh38
rs781944972 Chr.9: 133397937-133397940 on Build GRCh38
rs201001539 Chr.9: 133398040-133398040 on Build GRCh38
rs782224193 Chr.9: 133398193-133398193 on Build GRCh38
rs781823944 Chr.9: 133398000-133398000 on Build GRCh38
rs978915905 Chr.9: 133398124-133398124 on Build GRCh38
rs200682924 Chr.9: 133397969-133397969 on Build GRCh38
rs868024760 Chr.9: 133397962-133397962 on Build GRCh38

View Full Product Details