Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135624540-135625083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT6D1
Entrez Gene ID
360203
Gene Name
glycosyltransferase 6 domain containing 1
Gene Aliases
GLTDC1, GT6M7
UniGene
Hs.522491
Gene Chromosome Location
Chr.9: 135623656-135641216 on Build GRCh38
Transcripts
NM_182974.2 XM_011518633.1 XM_011518634.1 XM_011518635.1 XM_011518636.1 XM_011518637.1 XM_011518638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7171494 c.330C>G p.I110M Chr.9: 135624598-135624598 on Build GRCh38
1106662 c.323G>A p.R108Q Chr.9: 135624605-135624605 on Build GRCh38
1158357 c.356T>C p.F119S Chr.9: 135624572-135624572 on Build GRCh38
3665352 c.329T>A p.I110N Chr.9: 135624599-135624599 on Build GRCh38
5440300 c.295G>A p.A99T Chr.9: 135624633-135624633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869224482 Chr.9: 135624913-135624914 on Build GRCh38
rs560477208 Chr.9: 135624577-135624577 on Build GRCh38
rs777686591 Chr.9: 135624692-135624692 on Build GRCh38
rs371842118 Chr.9: 135624715-135624715 on Build GRCh38
rs749044795 Chr.9: 135624720-135624721 on Build GRCh38
rs749206975 Chr.9: 135624593-135624593 on Build GRCh38
rs199672710 Chr.9: 135624708-135624709 on Build GRCh38
rs750971060 Chr.9: 135624898-135624899 on Build GRCh38
rs754957583 Chr.9: 135624681-135624681 on Build GRCh38
rs141866218 Chr.9: 135624928-135624928 on Build GRCh38
rs754546707 Chr.9: 135624580-135624580 on Build GRCh38
rs564771016 Chr.9: 135624747-135624747 on Build GRCh38
rs746697657 Chr.9: 135624715-135624715 on Build GRCh38
rs184745121 Chr.9: 135624830-135624830 on Build GRCh38
rs200835106 Chr.9: 135624648-135624648 on Build GRCh38
rs981866121 Chr.9: 135624650-135624650 on Build GRCh38
rs984647370 Chr.9: 135624882-135624882 on Build GRCh38
rs770447988 Chr.9: 135624720-135624720 on Build GRCh38
rs376699462 Chr.9: 135624720-135624724 on Build GRCh38
rs531444908 Chr.9: 135624725-135624737 on Build GRCh38
rs778745771 Chr.9: 135624605-135624605 on Build GRCh38
rs774619535 Chr.9: 135624888-135624888 on Build GRCh38
rs184333688 Chr.9: 135625011-135625011 on Build GRCh38
rs749501630 Chr.9: 135624878-135624898 on Build GRCh38
rs567374802 Chr.9: 135624760-135624760 on Build GRCh38
rs537059795 Chr.9: 135624735-135624735 on Build GRCh38
rs562523459 Chr.9: 135624876-135624876 on Build GRCh38
rs751057655 Chr.9: 135624715-135624716 on Build GRCh38
rs199518158 Chr.9: 135624653-135624653 on Build GRCh38
rs539639259 Chr.9: 135624855-135624855 on Build GRCh38
rs72471205 Chr.9: 135624725-135624725 on Build GRCh38
rs775971210 Chr.9: 135624638-135624638 on Build GRCh38
rs760116747 Chr.9: 135624645-135624645 on Build GRCh38
rs920325509 Chr.9: 135624727-135624728 on Build GRCh38
rs771846959 Chr.9: 135624610-135624610 on Build GRCh38
rs940337401 Chr.9: 135624709-135624709 on Build GRCh38
rs267602167 Chr.9: 135624634-135624634 on Build GRCh38
rs189611142 Chr.9: 135625041-135625041 on Build GRCh38
rs909955927 Chr.9: 135624916-135624916 on Build GRCh38
rs911573509 Chr.9: 135624688-135624688 on Build GRCh38
rs200651021 Chr.9: 135624728-135624728 on Build GRCh38
rs572331175 Chr.9: 135624854-135624854 on Build GRCh38
rs937433179 Chr.9: 135624963-135624963 on Build GRCh38
rs28399734 Chr.9: 135624917-135624917 on Build GRCh38
rs534377547 Chr.9: 135624769-135624769 on Build GRCh38
rs930381782 Chr.9: 135624729-135624729 on Build GRCh38
rs192109365 Chr.9: 135624579-135624579 on Build GRCh38
rs374362715 Chr.9: 135624724-135624724 on Build GRCh38
rs369326923 Chr.9: 135624723-135624724 on Build GRCh38
rs1020341901 Chr.9: 135624641-135624641 on Build GRCh38
rs765756238 Chr.9: 135624706-135624707 on Build GRCh38
rs543823262 Chr.9: 135624880-135624880 on Build GRCh38
rs750422819 Chr.9: 135624658-135624658 on Build GRCh38
rs200428393 Chr.9: 135624574-135624574 on Build GRCh38
rs763072064 Chr.9: 135624897-135624899 on Build GRCh38
rs750406947 Chr.9: 135624718-135624720 on Build GRCh38
rs1036073257 Chr.9: 135624721-135624727 on Build GRCh38
rs867423837 Chr.9: 135624724-135624724 on Build GRCh38
rs760098918 Chr.9: 135624635-135624636 on Build GRCh38
rs951410608 Chr.9: 135624875-135624875 on Build GRCh38
rs753115294 Chr.9: 135624647-135624647 on Build GRCh38
rs548653567 Chr.9: 135624737-135624737 on Build GRCh38
rs7466817 Chr.9: 135624685-135624685 on Build GRCh38
rs199882249 Chr.9: 135624606-135624606 on Build GRCh38
rs772933599 Chr.9: 135624619-135624619 on Build GRCh38
rs375548287 Chr.9: 135624720-135624720 on Build GRCh38
rs1032186325 Chr.9: 135624613-135624613 on Build GRCh38
rs1055849272 Chr.9: 135624996-135624996 on Build GRCh38
rs192967058 Chr.9: 135624955-135624955 on Build GRCh38
rs757991946 Chr.9: 135624711-135624715 on Build GRCh38
rs547628590 Chr.9: 135624633-135624633 on Build GRCh38
rs563602247 Chr.9: 135625040-135625040 on Build GRCh38
rs1961127 Chr.9: 135624997-135624997 on Build GRCh38
rs377522378 Chr.9: 135624587-135624587 on Build GRCh38
rs778649111 Chr.9: 135624684-135624684 on Build GRCh38
rs562327882 Chr.9: 135624897-135624898 on Build GRCh38
rs757442491 Chr.9: 135624654-135624654 on Build GRCh38
rs561900630 Chr.9: 135624884-135624884 on Build GRCh38
rs971791389 Chr.9: 135625030-135625030 on Build GRCh38
rs756752900 Chr.9: 135624716-135624717 on Build GRCh38
rs12347700 Chr.9: 135625058-135625058 on Build GRCh38
rs138897981 Chr.9: 135624999-135624999 on Build GRCh38
rs905005995 Chr.9: 135624789-135624789 on Build GRCh38
rs997168574 Chr.9: 135625045-135625045 on Build GRCh38
rs775580075 Chr.9: 135624783-135624783 on Build GRCh38
rs1044943218 Chr.9: 135624731-135624731 on Build GRCh38
rs1815481 Chr.9: 135624856-135624856 on Build GRCh38
rs781365281 Chr.9: 135624656-135624656 on Build GRCh38
rs375210935 Chr.9: 135624867-135624867 on Build GRCh38
rs187917605 Chr.9: 135624886-135624886 on Build GRCh38
rs765609739 Chr.9: 135624646-135624646 on Build GRCh38
rs780652650 Chr.9: 135624717-135624720 on Build GRCh38
rs113688287 Chr.9: 135625000-135625000 on Build GRCh38

View Full Product Details