Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135639185-135639724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT6D1
Entrez Gene ID
360203
Gene Name
glycosyltransferase 6 domain containing 1
Gene Aliases
GLTDC1, GT6M7
UniGene
Hs.522491
Gene Chromosome Location
Chr.9: 135623656-135641216 on Build GRCh38
Transcripts
NM_182974.2 XM_011518633.1 XM_011518634.1 XM_011518635.1 XM_011518636.1 XM_011518637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382607 c.2T>C p.M1T Chr.9: 135639186-135639186 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs899595081 Chr.9: 135639678-135639678 on Build GRCh38
rs147911480 Chr.9: 135639667-135639667 on Build GRCh38
rs1050958101 Chr.9: 135639473-135639473 on Build GRCh38
rs572695264 Chr.9: 135639651-135639651 on Build GRCh38
rs376057690 Chr.9: 135639335-135639335 on Build GRCh38
rs369151117 Chr.9: 135639423-135639423 on Build GRCh38
rs1039510510 Chr.9: 135639309-135639309 on Build GRCh38
rs776428649 Chr.9: 135639499-135639499 on Build GRCh38
rs373138535 Chr.9: 135639444-135639444 on Build GRCh38
rs1004497767 Chr.9: 135639406-135639406 on Build GRCh38
rs74515947 Chr.9: 135639292-135639292 on Build GRCh38
rs980957684 Chr.9: 135639391-135639391 on Build GRCh38
rs1001445223 Chr.9: 135639512-135639512 on Build GRCh38
rs956321468 Chr.9: 135639334-135639334 on Build GRCh38
rs1013884840 Chr.9: 135639641-135639641 on Build GRCh38
rs973374354 Chr.9: 135639427-135639432 on Build GRCh38
rs114331488 Chr.9: 135639302-135639302 on Build GRCh38
rs766796554 Chr.9: 135639220-135639220 on Build GRCh38
rs553090563 Chr.9: 135639424-135639424 on Build GRCh38
rs928268227 Chr.9: 135639265-135639265 on Build GRCh38
rs374644790 Chr.9: 135639208-135639208 on Build GRCh38
rs771045879 Chr.9: 135639489-135639489 on Build GRCh38
rs886907848 Chr.9: 135639698-135639698 on Build GRCh38
rs917624100 Chr.9: 135639374-135639374 on Build GRCh38
rs767124749 Chr.9: 135639212-135639212 on Build GRCh38
rs746491547 Chr.9: 135639443-135639443 on Build GRCh38
rs754101395 Chr.9: 135639230-135639230 on Build GRCh38
rs372571883 Chr.9: 135639596-135639596 on Build GRCh38
rs771736756 Chr.9: 135639284-135639284 on Build GRCh38
rs533509624 Chr.9: 135639277-135639277 on Build GRCh38
rs527507847 Chr.9: 135639419-135639419 on Build GRCh38
rs528488737 Chr.9: 135639442-135639442 on Build GRCh38
rs980522016 Chr.9: 135639666-135639666 on Build GRCh38
rs149804729 Chr.9: 135639647-135639647 on Build GRCh38
rs542694863 Chr.9: 135639392-135639392 on Build GRCh38
rs948263107 Chr.9: 135639236-135639236 on Build GRCh38
rs933703807 Chr.9: 135639691-135639691 on Build GRCh38
rs931156493 Chr.9: 135639407-135639407 on Build GRCh38
rs761485010 Chr.9: 135639210-135639210 on Build GRCh38
rs563896825 Chr.9: 135639354-135639354 on Build GRCh38
rs988766748 Chr.9: 135639532-135639532 on Build GRCh38
rs751017986 Chr.9: 135639216-135639216 on Build GRCh38
rs183738463 Chr.9: 135639490-135639490 on Build GRCh38
rs996264795 Chr.9: 135639464-135639464 on Build GRCh38
rs935600710 Chr.9: 135639513-135639513 on Build GRCh38
rs546648772 Chr.9: 135639463-135639463 on Build GRCh38
rs887032569 Chr.9: 135639440-135639440 on Build GRCh38
rs969055939 Chr.9: 135639652-135639652 on Build GRCh38
rs763332794 Chr.9: 135639209-135639210 on Build GRCh38
rs865803377 Chr.9: 135639553-135639553 on Build GRCh38
rs572806617 Chr.9: 135639264-135639264 on Build GRCh38
rs915491037 Chr.9: 135639549-135639549 on Build GRCh38
rs761055572 Chr.9: 135639217-135639217 on Build GRCh38
rs550657331 Chr.9: 135639657-135639657 on Build GRCh38
rs878888592 Chr.9: 135639259-135639259 on Build GRCh38
rs766448767 Chr.9: 135639559-135639559 on Build GRCh38
rs947783484 Chr.9: 135639375-135639375 on Build GRCh38
rs1029881296 Chr.9: 135639388-135639388 on Build GRCh38
rs1040347973 Chr.9: 135639459-135639459 on Build GRCh38
rs376146043 Chr.9: 135639607-135639607 on Build GRCh38
rs77273125 Chr.9: 135639281-135639281 on Build GRCh38
rs1031226351 Chr.9: 135639554-135639554 on Build GRCh38

View Full Product Details