Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153454299-153454789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS7
Entrez Gene ID
55559
Gene Name
HAUS augmin like complex subunit 7
Gene Aliases
UCHL5IP, UIP1
UniGene
Hs.170835
Gene Chromosome Location
Chr.X: 153447665-153495525 on Build GRCh38
Transcripts
NM_017518.7 NR_073156.1

SNP Detail

SNP ID Chromosome Location
rs199738769 Chr.X: 153454374-153454374 on Build GRCh38
rs1002382247 Chr.X: 153454704-153454704 on Build GRCh38
rs932521059 Chr.X: 153454572-153454572 on Build GRCh38
rs782044463 Chr.X: 153454539-153454539 on Build GRCh38
rs782362578 Chr.X: 153454511-153454511 on Build GRCh38
rs969570085 Chr.X: 153454437-153454437 on Build GRCh38
rs782649644 Chr.X: 153454353-153454353 on Build GRCh38
rs782658603 Chr.X: 153454464-153454464 on Build GRCh38
rs781884449 Chr.X: 153454668-153454668 on Build GRCh38
rs782035878 Chr.X: 153454533-153454533 on Build GRCh38
rs374148539 Chr.X: 153454402-153454402 on Build GRCh38
rs112656480 Chr.X: 153454674-153454674 on Build GRCh38
rs782730661 Chr.X: 153454540-153454540 on Build GRCh38
rs781889161 Chr.X: 153454441-153454441 on Build GRCh38
rs143101778 Chr.X: 153454699-153454699 on Build GRCh38
rs782411411 Chr.X: 153454470-153454470 on Build GRCh38
rs782150765 Chr.X: 153454537-153454537 on Build GRCh38
rs781825980 Chr.X: 153454583-153454583 on Build GRCh38
rs782397002 Chr.X: 153454407-153454407 on Build GRCh38
rs944040783 Chr.X: 153454549-153454549 on Build GRCh38
rs782098607 Chr.X: 153454432-153454432 on Build GRCh38
rs782708171 Chr.X: 153454545-153454545 on Build GRCh38
rs782196464 Chr.X: 153454390-153454390 on Build GRCh38
rs1040950413 Chr.X: 153454342-153454342 on Build GRCh38
rs200356507 Chr.X: 153454351-153454351 on Build GRCh38
rs782306666 Chr.X: 153454324-153454324 on Build GRCh38
rs1013920214 Chr.X: 153454686-153454686 on Build GRCh38
rs782242417 Chr.X: 153454362-153454362 on Build GRCh38
rs140068588 Chr.X: 153454410-153454410 on Build GRCh38
rs782130588 Chr.X: 153454516-153454516 on Build GRCh38
rs782221784 Chr.X: 153454406-153454406 on Build GRCh38
rs143828088 Chr.X: 153454422-153454422 on Build GRCh38
rs782570235 Chr.X: 153454487-153454487 on Build GRCh38
rs782355109 Chr.X: 153454430-153454430 on Build GRCh38
rs1023016378 Chr.X: 153454695-153454695 on Build GRCh38
rs781988163 Chr.X: 153454454-153454454 on Build GRCh38
rs371820008 Chr.X: 153454589-153454589 on Build GRCh38
rs782320708 Chr.X: 153454424-153454424 on Build GRCh38
rs782099275 Chr.X: 153454586-153454586 on Build GRCh38
rs1052858826 Chr.X: 153454611-153454611 on Build GRCh38
rs367988369 Chr.X: 153454455-153454455 on Build GRCh38
rs782774877 Chr.X: 153454587-153454587 on Build GRCh38
rs187474458 Chr.X: 153454745-153454745 on Build GRCh38
rs782103739 Chr.X: 153454550-153454550 on Build GRCh38
rs782624525 Chr.X: 153454404-153454404 on Build GRCh38
rs192467959 Chr.X: 153454764-153454764 on Build GRCh38
rs782104997 Chr.X: 153454527-153454527 on Build GRCh38
rs781877581 Chr.X: 153454536-153454536 on Build GRCh38
rs1042921750 Chr.X: 153454584-153454584 on Build GRCh38
rs781791916 Chr.X: 153454541-153454541 on Build GRCh38
rs908562364 Chr.X: 153454548-153454548 on Build GRCh38
rs998574389 Chr.X: 153454378-153454378 on Build GRCh38
rs782348370 Chr.X: 153454506-153454506 on Build GRCh38
rs782026775 Chr.X: 153454420-153454420 on Build GRCh38
rs781953096 Chr.X: 153454512-153454512 on Build GRCh38
rs782399516 Chr.X: 153454471-153454471 on Build GRCh38
rs782374980 Chr.X: 153454520-153454520 on Build GRCh38
rs782661091 Chr.X: 153454385-153454385 on Build GRCh38
rs782051339 Chr.X: 153454542-153454542 on Build GRCh38
rs782173293 Chr.X: 153454494-153454494 on Build GRCh38
rs782725800 Chr.X: 153454436-153454436 on Build GRCh38
rs377607117 Chr.X: 153454350-153454350 on Build GRCh38
rs921185281 Chr.X: 153454567-153454567 on Build GRCh38

View Full Product Details