Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65114606-65115102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM7SF2
Entrez Gene ID
7108
Gene Name
transmembrane 7 superfamily member 2
Gene Aliases
ANG1, DHCR14A, NET47
UniGene
Hs.31130
Gene Chromosome Location
Chr.11: 65111854-65116235 on Build GRCh38
Transcripts
NM_001277233.1 NM_003273.3 NR_102367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3931398 c.677T>G p.V226G Chr.11: 65114786-65114786 on Build GRCh38
5689714 c.640G>C p.E214Q Chr.11: 65114749-65114749 on Build GRCh38
5619696 c.802C>T p.P268S Chr.11: 65114991-65114991 on Build GRCh38
3791789 c.696C>G p.L232L Chr.11: 65114805-65114805 on Build GRCh38
6802656 c.729C>T p.A243A Chr.11: 65114918-65114918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375674215 Chr.11: 65115001-65115001 on Build GRCh38
rs374920822 Chr.11: 65114781-65114781 on Build GRCh38
rs878878508 Chr.11: 65115053-65115053 on Build GRCh38
rs760734524 Chr.11: 65115082-65115085 on Build GRCh38
rs370899002 Chr.11: 65114910-65114910 on Build GRCh38
rs754602343 Chr.11: 65114845-65114845 on Build GRCh38
rs1013606088 Chr.11: 65114713-65114713 on Build GRCh38
rs555372850 Chr.11: 65114650-65114650 on Build GRCh38
rs1041730451 Chr.11: 65114634-65114634 on Build GRCh38
rs775696515 Chr.11: 65114786-65114786 on Build GRCh38
rs776660302 Chr.11: 65115017-65115017 on Build GRCh38
rs374323627 Chr.11: 65114871-65114871 on Build GRCh38
rs776671000 Chr.11: 65114777-65114777 on Build GRCh38
rs895322845 Chr.11: 65114981-65114981 on Build GRCh38
rs774666568 Chr.11: 65114714-65114716 on Build GRCh38
rs762061332 Chr.11: 65114887-65114887 on Build GRCh38
rs768796457 Chr.11: 65114918-65114918 on Build GRCh38
rs540861703 Chr.11: 65114808-65114808 on Build GRCh38
rs747382183 Chr.11: 65114883-65114883 on Build GRCh38
rs71468691 Chr.11: 65114743-65114743 on Build GRCh38
rs763712793 Chr.11: 65114948-65114948 on Build GRCh38
rs868412324 Chr.11: 65114913-65114913 on Build GRCh38
rs951158490 Chr.11: 65114850-65114850 on Build GRCh38
rs959651671 Chr.11: 65114662-65114662 on Build GRCh38
rs745883751 Chr.11: 65114897-65114897 on Build GRCh38
rs1033886511 Chr.11: 65114783-65114784 on Build GRCh38
rs766464859 Chr.11: 65114832-65114832 on Build GRCh38
rs759674435 Chr.11: 65114780-65114780 on Build GRCh38
rs368969753 Chr.11: 65114849-65114849 on Build GRCh38
rs763739423 Chr.11: 65115045-65115045 on Build GRCh38
rs143577587 Chr.11: 65114949-65114949 on Build GRCh38
rs747583561 Chr.11: 65114760-65114760 on Build GRCh38
rs757769793 Chr.11: 65114888-65114888 on Build GRCh38
rs33981236 Chr.11: 65114969-65114969 on Build GRCh38
rs779642530 Chr.11: 65114715-65114715 on Build GRCh38
rs544170554 Chr.11: 65114698-65114698 on Build GRCh38
rs776714853 Chr.11: 65114932-65114932 on Build GRCh38
rs748516890 Chr.11: 65114737-65114737 on Build GRCh38
rs754653686 Chr.11: 65114734-65114734 on Build GRCh38
rs201108253 Chr.11: 65114768-65114768 on Build GRCh38
rs770862173 Chr.11: 65115006-65115006 on Build GRCh38
rs764871670 Chr.11: 65114942-65114942 on Build GRCh38
rs374474834 Chr.11: 65115071-65115071 on Build GRCh38
rs200355387 Chr.11: 65114934-65114934 on Build GRCh38
rs12270532 Chr.11: 65114691-65114691 on Build GRCh38
rs758049533 Chr.11: 65114943-65114943 on Build GRCh38
rs1013053449 Chr.11: 65115004-65115004 on Build GRCh38
rs751346635 Chr.11: 65114684-65114684 on Build GRCh38
rs781752283 Chr.11: 65114890-65114890 on Build GRCh38
rs778766450 Chr.11: 65114735-65114735 on Build GRCh38
rs774501532 Chr.11: 65114988-65114988 on Build GRCh38
rs187240326 Chr.11: 65115034-65115034 on Build GRCh38
rs760444560 Chr.11: 65114921-65114921 on Build GRCh38
rs769902078 Chr.11: 65115035-65115035 on Build GRCh38
rs762121544 Chr.11: 65115055-65115055 on Build GRCh38
rs748423693 Chr.11: 65114631-65114631 on Build GRCh38
rs867555087 Chr.11: 65114806-65114806 on Build GRCh38
rs749373964 Chr.11: 65114985-65114985 on Build GRCh38
rs912403197 Chr.11: 65114903-65114903 on Build GRCh38
rs760505763 Chr.11: 65114830-65114830 on Build GRCh38
rs753383929 Chr.11: 65114835-65114835 on Build GRCh38
rs922505214 Chr.11: 65114870-65114870 on Build GRCh38
rs775645185 Chr.11: 65114904-65114904 on Build GRCh38
rs750526130 Chr.11: 65115081-65115084 on Build GRCh38
rs765493716 Chr.11: 65114681-65114681 on Build GRCh38
rs761739440 Chr.11: 65114688-65114688 on Build GRCh38
rs938764103 Chr.11: 65114951-65114951 on Build GRCh38
rs758861678 Chr.11: 65114997-65114998 on Build GRCh38
rs560516989 Chr.11: 65114852-65114852 on Build GRCh38
rs148878643 Chr.11: 65114919-65114919 on Build GRCh38
rs183249527 Chr.11: 65114756-65114756 on Build GRCh38
rs761460155 Chr.11: 65114789-65114789 on Build GRCh38
rs772980923 Chr.11: 65114827-65114827 on Build GRCh38
rs751188189 Chr.11: 65115046-65115046 on Build GRCh38
rs768855590 Chr.11: 65114703-65114704 on Build GRCh38
rs755527266 Chr.11: 65114701-65114701 on Build GRCh38
rs551357064 Chr.11: 65114968-65114968 on Build GRCh38
rs762439033 Chr.11: 65115040-65115040 on Build GRCh38
rs766413625 Chr.11: 65114929-65114929 on Build GRCh38
rs767903778 Chr.11: 65115059-65115059 on Build GRCh38
rs760625665 Chr.11: 65114829-65114829 on Build GRCh38
rs902293592 Chr.11: 65114952-65114952 on Build GRCh38
rs993693223 Chr.11: 65115083-65115083 on Build GRCh38
rs1002937925 Chr.11: 65114656-65114656 on Build GRCh38
rs772488065 Chr.11: 65114755-65114755 on Build GRCh38
rs768906444 Chr.11: 65114987-65114987 on Build GRCh38
rs775054516 Chr.11: 65115039-65115039 on Build GRCh38
rs369237248 Chr.11: 65114725-65114725 on Build GRCh38
rs201385088 Chr.11: 65115032-65115032 on Build GRCh38
rs372885636 Chr.11: 65115080-65115080 on Build GRCh38
rs769879468 Chr.11: 65114900-65114900 on Build GRCh38
rs368684139 Chr.11: 65115079-65115079 on Build GRCh38

View Full Product Details