Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66511113-66511586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS1
Entrez Gene ID
582
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
BBS2L2
UniGene
Hs.502915
Gene Chromosome Location
Chr.11: 66510648-66533613 on Build GRCh38
Transcripts
NM_024649.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6487973 c.259G>A p.G87R Chr.11: 66511228-66511228 on Build GRCh38
7357321 c.133G>C p.D45H Chr.11: 66511213-66511213 on Build GRCh38
7357322 c.244G>C p.D82H Chr.11: 66511213-66511213 on Build GRCh38
6487972 c.148G>A p.G50R Chr.11: 66511228-66511228 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs962623435 Chr.11: 66511160-66511160 on Build GRCh38
rs759208852 Chr.11: 66511196-66511196 on Build GRCh38
rs758468091 Chr.11: 66511253-66511253 on Build GRCh38
rs949330163 Chr.11: 66511311-66511311 on Build GRCh38
rs762092708 Chr.11: 66511194-66511194 on Build GRCh38
rs752898124 Chr.11: 66511326-66511326 on Build GRCh38
rs751335279 Chr.11: 66511497-66511497 on Build GRCh38
rs565170162 Chr.11: 66511467-66511467 on Build GRCh38
rs957132327 Chr.11: 66511530-66511530 on Build GRCh38
rs373705671 Chr.11: 66511281-66511281 on Build GRCh38
rs550567096 Chr.11: 66511335-66511335 on Build GRCh38
rs907188596 Chr.11: 66511316-66511316 on Build GRCh38
rs760939609 Chr.11: 66511137-66511138 on Build GRCh38
rs527545476 Chr.11: 66511475-66511475 on Build GRCh38
rs887883469 Chr.11: 66511394-66511394 on Build GRCh38
rs935704561 Chr.11: 66511260-66511260 on Build GRCh38
rs779592725 Chr.11: 66511290-66511290 on Build GRCh38
rs988491731 Chr.11: 66511541-66511541 on Build GRCh38
rs765345749 Chr.11: 66511284-66511284 on Build GRCh38
rs766612419 Chr.11: 66511276-66511276 on Build GRCh38
rs998175544 Chr.11: 66511459-66511459 on Build GRCh38
rs757205522 Chr.11: 66511523-66511523 on Build GRCh38
rs543797671 Chr.11: 66511143-66511143 on Build GRCh38
rs925718222 Chr.11: 66511202-66511202 on Build GRCh38
rs369563296 Chr.11: 66511262-66511262 on Build GRCh38
rs61610137 Chr.11: 66511452-66511452 on Build GRCh38
rs766602837 Chr.11: 66511237-66511237 on Build GRCh38
rs752658686 Chr.11: 66511238-66511238 on Build GRCh38
rs757090408 Chr.11: 66511261-66511261 on Build GRCh38
rs899248150 Chr.11: 66511364-66511364 on Build GRCh38
rs763182339 Chr.11: 66511425-66511425 on Build GRCh38
rs562875897 Chr.11: 66511357-66511357 on Build GRCh38
rs554294689 Chr.11: 66511149-66511149 on Build GRCh38
rs987986409 Chr.11: 66511299-66511299 on Build GRCh38
rs886415093 Chr.11: 66511408-66511408 on Build GRCh38
rs367682811 Chr.11: 66511213-66511213 on Build GRCh38
rs760793954 Chr.11: 66511232-66511232 on Build GRCh38
rs531710732 Chr.11: 66511407-66511407 on Build GRCh38
rs975741601 Chr.11: 66511182-66511182 on Build GRCh38
rs372818503 Chr.11: 66511287-66511287 on Build GRCh38
rs750300870 Chr.11: 66511558-66511558 on Build GRCh38
rs773503074 Chr.11: 66511222-66511222 on Build GRCh38
rs536020758 Chr.11: 66511403-66511403 on Build GRCh38
rs1030618127 Chr.11: 66511463-66511463 on Build GRCh38
rs770554491 Chr.11: 66511132-66511132 on Build GRCh38
rs190639799 Chr.11: 66511191-66511191 on Build GRCh38
rs751245532 Chr.11: 66511254-66511254 on Build GRCh38
rs34286145 Chr.11: 66511456-66511457 on Build GRCh38
rs925052659 Chr.11: 66511269-66511269 on Build GRCh38
rs1015056555 Chr.11: 66511434-66511434 on Build GRCh38
rs749835624 Chr.11: 66511179-66511179 on Build GRCh38
rs367922723 Chr.11: 66511169-66511169 on Build GRCh38
rs745550835 Chr.11: 66511277-66511277 on Build GRCh38
rs922934188 Chr.11: 66511220-66511220 on Build GRCh38
rs370383610 Chr.11: 66511477-66511477 on Build GRCh38
rs1046348073 Chr.11: 66511313-66511313 on Build GRCh38
rs937385193 Chr.11: 66511334-66511334 on Build GRCh38
rs775937558 Chr.11: 66511171-66511171 on Build GRCh38
rs745326262 Chr.11: 66511140-66511140 on Build GRCh38
rs913786444 Chr.11: 66511305-66511305 on Build GRCh38
rs780899721 Chr.11: 66511133-66511133 on Build GRCh38
rs183088220 Chr.11: 66511555-66511555 on Build GRCh38
rs1057516507 Chr.11: 66511241-66511241 on Build GRCh38
rs763839923 Chr.11: 66511448-66511448 on Build GRCh38
rs768991460 Chr.11: 66511188-66511188 on Build GRCh38
rs913179847 Chr.11: 66511309-66511309 on Build GRCh38
rs570240047 Chr.11: 66511308-66511308 on Build GRCh38
rs994866010 Chr.11: 66511368-66511368 on Build GRCh38
rs949984921 Chr.11: 66511310-66511310 on Build GRCh38

View Full Product Details