Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80672684-80673188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
UniGene
Hs.147027
Gene Chromosome Location
Chr.X: 80669488-80809736 on Build GRCh38
Transcripts
NM_153252.4 XM_005262113.3

SNP Detail

SNP ID Chromosome Location
rs932219786 Chr.X: 80672746-80672746 on Build GRCh38
rs1032431930 Chr.X: 80673137-80673137 on Build GRCh38
rs775875353 Chr.X: 80672761-80672761 on Build GRCh38
rs1003167599 Chr.X: 80672942-80672942 on Build GRCh38
rs941706370 Chr.X: 80672832-80672832 on Build GRCh38
rs756983142 Chr.X: 80672892-80672892 on Build GRCh38
rs777667683 Chr.X: 80672713-80672713 on Build GRCh38
rs1052637959 Chr.X: 80672768-80672768 on Build GRCh38
rs904826634 Chr.X: 80672895-80672895 on Build GRCh38
rs776193540 Chr.X: 80672714-80672714 on Build GRCh38
rs1034870540 Chr.X: 80672995-80672995 on Build GRCh38
rs953325451 Chr.X: 80673163-80673163 on Build GRCh38
rs192339199 Chr.X: 80672745-80672745 on Build GRCh38
rs767696195 Chr.X: 80673162-80673162 on Build GRCh38
rs1036569184 Chr.X: 80672835-80672835 on Build GRCh38
rs146779779 Chr.X: 80672822-80672822 on Build GRCh38
rs1016795681 Chr.X: 80673086-80673086 on Build GRCh38
rs958278693 Chr.X: 80673145-80673145 on Build GRCh38
rs951316524 Chr.X: 80673011-80673011 on Build GRCh38
rs879201626 Chr.X: 80673015-80673015 on Build GRCh38
rs762108287 Chr.X: 80673147-80673147 on Build GRCh38
rs893529719 Chr.X: 80672783-80672783 on Build GRCh38
rs764587615 Chr.X: 80673077-80673077 on Build GRCh38
rs781124974 Chr.X: 80673122-80673122 on Build GRCh38
rs1051978671 Chr.X: 80672758-80672758 on Build GRCh38
rs1013689138 Chr.X: 80672897-80672897 on Build GRCh38
rs116559809 Chr.X: 80673126-80673126 on Build GRCh38
rs45627037 Chr.X: 80672759-80672759 on Build GRCh38
rs963786326 Chr.X: 80673123-80673123 on Build GRCh38
rs1010686693 Chr.X: 80672799-80672799 on Build GRCh38

View Full Product Details