Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161766089-161766626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF6
Entrez Gene ID
22926
Gene Name
activating transcription factor 6
Gene Aliases
ACHM7, ATF6A
UniGene
Hs.492740
Gene Chromosome Location
Chr.1: 161766244-161964070 on Build GRCh38
Transcripts
NM_007348.3 XM_006711224.1 XM_011509308.1 XM_011509309.1 XM_011509310.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866227 c.16G>T p.G6W Chr.1: 161766376-161766376 on Build GRCh38
3477342 c.48C>T p.S16S Chr.1: 161766408-161766408 on Build GRCh38
1183877 c.53G>A p.G18E Chr.1: 161766413-161766413 on Build GRCh38
290982 c.2_3insG p.E3fs*14 Chr.1: 161766362-161766363 on Build GRCh38
6059369 c.11C>T p.P4L Chr.1: 161766371-161766371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779891634 Chr.1: 161766372-161766372 on Build GRCh38
rs745683897 Chr.1: 161766317-161766317 on Build GRCh38
rs746651824 Chr.1: 161766373-161766373 on Build GRCh38
rs1048105738 Chr.1: 161766580-161766580 on Build GRCh38
rs750444250 Chr.1: 161766140-161766147 on Build GRCh38
rs772548546 Chr.1: 161766382-161766382 on Build GRCh38
rs375558807 Chr.1: 161766236-161766236 on Build GRCh38
rs985273891 Chr.1: 161766113-161766113 on Build GRCh38
rs182701901 Chr.1: 161766352-161766352 on Build GRCh38
rs777479721 Chr.1: 161766458-161766458 on Build GRCh38
rs532043545 Chr.1: 161766206-161766206 on Build GRCh38
rs904680636 Chr.1: 161766272-161766272 on Build GRCh38
rs371003268 Chr.1: 161766202-161766202 on Build GRCh38
rs944117883 Chr.1: 161766158-161766158 on Build GRCh38
rs760910329 Chr.1: 161766387-161766387 on Build GRCh38
rs774689227 Chr.1: 161766379-161766379 on Build GRCh38
rs867568743 Chr.1: 161766364-161766364 on Build GRCh38
rs764173088 Chr.1: 161766354-161766354 on Build GRCh38
rs763535281 Chr.1: 161766286-161766286 on Build GRCh38
rs759957513 Chr.1: 161766342-161766342 on Build GRCh38
rs60394665 Chr.1: 161766324-161766324 on Build GRCh38
rs763461610 Chr.1: 161766329-161766329 on Build GRCh38
rs757180603 Chr.1: 161766361-161766361 on Build GRCh38
rs900152545 Chr.1: 161766548-161766548 on Build GRCh38
rs976823099 Chr.1: 161766172-161766172 on Build GRCh38
rs765402374 Chr.1: 161766400-161766400 on Build GRCh38
rs540411870 Chr.1: 161766401-161766401 on Build GRCh38
rs200312577 Chr.1: 161766486-161766486 on Build GRCh38
rs768150981 Chr.1: 161766325-161766325 on Build GRCh38
rs563182439 Chr.1: 161766225-161766225 on Build GRCh38
rs797045170 Chr.1: 161766447-161766447 on Build GRCh38
rs985346103 Chr.1: 161766297-161766297 on Build GRCh38
rs574601352 Chr.1: 161766391-161766391 on Build GRCh38
rs774727601 Chr.1: 161766341-161766341 on Build GRCh38
rs370507995 Chr.1: 161766469-161766469 on Build GRCh38
rs111892839 Chr.1: 161766262-161766262 on Build GRCh38
rs186063803 Chr.1: 161766153-161766153 on Build GRCh38
rs79784766 Chr.1: 161766583-161766583 on Build GRCh38
rs2298019 Chr.1: 161766245-161766245 on Build GRCh38
rs776006837 Chr.1: 161766384-161766384 on Build GRCh38
rs1017656039 Chr.1: 161766306-161766306 on Build GRCh38
rs1043108371 Chr.1: 161766435-161766435 on Build GRCh38
rs904733243 Chr.1: 161766460-161766460 on Build GRCh38
rs112863172 Chr.1: 161766407-161766407 on Build GRCh38
rs190591869 Chr.1: 161766268-161766268 on Build GRCh38
rs531147165 Chr.1: 161766520-161766520 on Build GRCh38
rs989482517 Chr.1: 161766331-161766331 on Build GRCh38
rs377663297 Chr.1: 161766318-161766318 on Build GRCh38
rs948250460 Chr.1: 161766237-161766237 on Build GRCh38
rs769182741 Chr.1: 161766438-161766438 on Build GRCh38
rs142912146 Chr.1: 161766189-161766189 on Build GRCh38
rs1012926384 Chr.1: 161766257-161766257 on Build GRCh38
rs111987479 Chr.1: 161766240-161766240 on Build GRCh38
rs776979117 Chr.1: 161766122-161766122 on Build GRCh38
rs548891834 Chr.1: 161766543-161766543 on Build GRCh38
rs896885185 Chr.1: 161766521-161766521 on Build GRCh38
rs778700614 Chr.1: 161766314-161766314 on Build GRCh38
rs747873305 Chr.1: 161766437-161766437 on Build GRCh38
rs935352587 Chr.1: 161766327-161766327 on Build GRCh38
rs764251276 Chr.1: 161766390-161766390 on Build GRCh38
rs775953770 Chr.1: 161766471-161766471 on Build GRCh38
rs546282261 Chr.1: 161766426-161766426 on Build GRCh38
rs761779169 Chr.1: 161766393-161766393 on Build GRCh38
rs1054484132 Chr.1: 161766215-161766215 on Build GRCh38
rs114894622 Chr.1: 161766290-161766290 on Build GRCh38
rs774408877 Chr.1: 161766322-161766323 on Build GRCh38
rs752959048 Chr.1: 161766346-161766346 on Build GRCh38
rs753865001 Chr.1: 161766355-161766355 on Build GRCh38
rs1035413419 Chr.1: 161766277-161766277 on Build GRCh38
rs994619756 Chr.1: 161766579-161766579 on Build GRCh38
rs187071091 Chr.1: 161766597-161766597 on Build GRCh38
rs915211840 Chr.1: 161766363-161766363 on Build GRCh38
rs753909268 Chr.1: 161766392-161766392 on Build GRCh38
rs757385689 Chr.1: 161766231-161766231 on Build GRCh38
rs113060703 Chr.1: 161766482-161766482 on Build GRCh38
rs965173377 Chr.1: 161766313-161766313 on Build GRCh38
rs1009536575 Chr.1: 161766531-161766531 on Build GRCh38
rs894567626 Chr.1: 161766248-161766248 on Build GRCh38
rs758162509 Chr.1: 161766370-161766370 on Build GRCh38
rs768033293 Chr.1: 161766345-161766345 on Build GRCh38
rs778741081 Chr.1: 161766367-161766367 on Build GRCh38
rs370820723 Chr.1: 161766323-161766323 on Build GRCh38
rs768207958 Chr.1: 161766374-161766374 on Build GRCh38
rs935337347 Chr.1: 161766204-161766204 on Build GRCh38
rs761145781 Chr.1: 161766477-161766477 on Build GRCh38
rs760186881 Chr.1: 161766381-161766381 on Build GRCh38
rs149825732 Chr.1: 161766427-161766427 on Build GRCh38
rs35284289 Chr.1: 161766419-161766419 on Build GRCh38
rs751361347 Chr.1: 161766421-161766421 on Build GRCh38
rs559454728 Chr.1: 161766282-161766282 on Build GRCh38
rs575089922 Chr.1: 161766302-161766302 on Build GRCh38
rs111854142 Chr.1: 161766287-161766287 on Build GRCh38
rs200220130 Chr.1: 161766377-161766377 on Build GRCh38
rs757152310 Chr.1: 161766310-161766310 on Build GRCh38
rs977274638 Chr.1: 161766322-161766322 on Build GRCh38
rs1006820128 Chr.1: 161766585-161766585 on Build GRCh38
rs920400176 Chr.1: 161766200-161766200 on Build GRCh38

View Full Product Details