Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 66503855-66504255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH19
Entrez Gene ID
28513
Gene Name
cadherin 19
Gene Aliases
CDH7, CDH7L2
UniGene
Hs.42771
Gene Chromosome Location
Chr.18: 66501187-66604241 on Build GRCh38
Transcripts
NM_001271028.1 NM_021153.3 NR_073130.1 XM_011525931.2 XM_017025711.1

SNP Detail

SNP ID Chromosome Location
rs1030736472 Chr.18: 66504203-66504203 on Build GRCh38
rs967536389 Chr.18: 66503894-66503894 on Build GRCh38
rs1010421149 Chr.18: 66503959-66503959 on Build GRCh38
rs971169073 Chr.18: 66504194-66504194 on Build GRCh38
rs549057311 Chr.18: 66503892-66503892 on Build GRCh38
rs1041428688 Chr.18: 66504010-66504010 on Build GRCh38
rs1014926470 Chr.18: 66504127-66504128 on Build GRCh38
rs951553763 Chr.18: 66504160-66504160 on Build GRCh38
rs568281300 Chr.18: 66504032-66504032 on Build GRCh38
rs892943410 Chr.18: 66503958-66503958 on Build GRCh38
rs573270680 Chr.18: 66504092-66504092 on Build GRCh38
rs777278503 Chr.18: 66503918-66503918 on Build GRCh38
rs751054749 Chr.18: 66504175-66504175 on Build GRCh38
rs191950546 Chr.18: 66504083-66504083 on Build GRCh38
rs375872542 Chr.18: 66504131-66504131 on Build GRCh38
rs927242350 Chr.18: 66504217-66504217 on Build GRCh38
rs1027166855 Chr.18: 66504122-66504122 on Build GRCh38
rs540483950 Chr.18: 66504199-66504199 on Build GRCh38
rs556942079 Chr.18: 66504067-66504067 on Build GRCh38
rs1045631394 Chr.18: 66503915-66503915 on Build GRCh38
rs757205357 Chr.18: 66504187-66504187 on Build GRCh38
rs1030747532 Chr.18: 66503977-66503977 on Build GRCh38
rs937199406 Chr.18: 66503921-66503921 on Build GRCh38
rs995701992 Chr.18: 66504104-66504104 on Build GRCh38
rs1007888471 Chr.18: 66504025-66504025 on Build GRCh38
rs890796254 Chr.18: 66504011-66504011 on Build GRCh38
rs58566192 Chr.18: 66504041-66504041 on Build GRCh38
rs989091848 Chr.18: 66503997-66503997 on Build GRCh38
rs567659037 Chr.18: 66504204-66504209 on Build GRCh38
rs987371610 Chr.18: 66504040-66504040 on Build GRCh38
rs187088877 Chr.18: 66504069-66504069 on Build GRCh38
rs1024551759 Chr.18: 66504174-66504174 on Build GRCh38
rs927204829 Chr.18: 66503917-66503917 on Build GRCh38
rs553118509 Chr.18: 66504000-66504000 on Build GRCh38
rs879224971 Chr.18: 66503951-66503956 on Build GRCh38
rs940534758 Chr.18: 66504058-66504058 on Build GRCh38
rs957723701 Chr.18: 66503981-66503981 on Build GRCh38
rs1028624346 Chr.18: 66504228-66504228 on Build GRCh38
rs781148981 Chr.18: 66504218-66504218 on Build GRCh38
rs931903655 Chr.18: 66504033-66504033 on Build GRCh38
rs899236459 Chr.18: 66504084-66504084 on Build GRCh38
rs72952466 Chr.18: 66504085-66504085 on Build GRCh38
rs1054205439 Chr.18: 66503922-66503922 on Build GRCh38

View Full Product Details