Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46920734-46921308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP35
Entrez Gene ID
2909
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP
UniGene
Hs.509447
Gene Chromosome Location
Chr.19: 46861002-47005077 on Build GRCh38
Transcripts
NM_004491.4 XM_011526873.2 XM_017026714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5493969 c.2071A>G p.N691D Chr.19: 46920746-46920746 on Build GRCh38
4438747 c.2304C>G p.V768V Chr.19: 46920979-46920979 on Build GRCh38
5474193 c.2226C>T p.Y742Y Chr.19: 46920901-46920901 on Build GRCh38
5802076 c.2530delT p.F844fs*43 Chr.19: 46921205-46921205 on Build GRCh38
3536241 c.2072A>C p.N691T Chr.19: 46920747-46920747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565209931 Chr.19: 46920949-46920949 on Build GRCh38
rs2228163 Chr.19: 46920814-46920814 on Build GRCh38
rs777754542 Chr.19: 46921021-46921021 on Build GRCh38
rs749271027 Chr.19: 46921154-46921154 on Build GRCh38
rs369040622 Chr.19: 46920862-46920862 on Build GRCh38
rs764651264 Chr.19: 46920766-46920766 on Build GRCh38
rs529760710 Chr.19: 46921137-46921137 on Build GRCh38
rs764787468 Chr.19: 46920947-46920947 on Build GRCh38
rs764408614 Chr.19: 46920906-46920906 on Build GRCh38
rs748059754 Chr.19: 46921008-46921008 on Build GRCh38
rs542766066 Chr.19: 46921181-46921181 on Build GRCh38
rs188397648 Chr.19: 46921240-46921240 on Build GRCh38
rs759414604 Chr.19: 46920963-46920963 on Build GRCh38
rs991596100 Chr.19: 46921273-46921273 on Build GRCh38
rs996580526 Chr.19: 46921098-46921098 on Build GRCh38
rs780870433 Chr.19: 46921004-46921004 on Build GRCh38
rs1031456574 Chr.19: 46921278-46921278 on Build GRCh38
rs576379485 Chr.19: 46920874-46920874 on Build GRCh38
rs758036557 Chr.19: 46920976-46920976 on Build GRCh38
rs757142299 Chr.19: 46921268-46921268 on Build GRCh38
rs767071078 Chr.19: 46921229-46921229 on Build GRCh38
rs754333661 Chr.19: 46920916-46920916 on Build GRCh38
rs775634128 Chr.19: 46920903-46920903 on Build GRCh38
rs745472938 Chr.19: 46920793-46920793 on Build GRCh38
rs376792949 Chr.19: 46921170-46921170 on Build GRCh38
rs754760484 Chr.19: 46920998-46920998 on Build GRCh38
rs769018982 Chr.19: 46921072-46921072 on Build GRCh38
rs1803672 Chr.19: 46920809-46920809 on Build GRCh38
rs754599800 Chr.19: 46921114-46921114 on Build GRCh38
rs991293926 Chr.19: 46920828-46920828 on Build GRCh38
rs1052665 Chr.19: 46921069-46921069 on Build GRCh38
rs753111353 Chr.19: 46920757-46920757 on Build GRCh38
rs574092629 Chr.19: 46920763-46920763 on Build GRCh38
rs757320618 Chr.19: 46920813-46920814 on Build GRCh38
rs1045689620 Chr.19: 46920911-46920911 on Build GRCh38
rs752386164 Chr.19: 46921237-46921237 on Build GRCh38
rs772278775 Chr.19: 46920901-46920901 on Build GRCh38
rs758124139 Chr.19: 46920800-46920800 on Build GRCh38
rs768383434 Chr.19: 46920819-46920819 on Build GRCh38
rs753731428 Chr.19: 46921260-46921260 on Build GRCh38
rs772190930 Chr.19: 46921032-46921032 on Build GRCh38
rs760842040 Chr.19: 46921058-46921058 on Build GRCh38
rs779731535 Chr.19: 46920990-46920990 on Build GRCh38
rs772088673 Chr.19: 46921155-46921155 on Build GRCh38
rs776876613 Chr.19: 46921078-46921078 on Build GRCh38
rs540715917 Chr.19: 46921027-46921027 on Build GRCh38
rs754354428 Chr.19: 46920781-46920781 on Build GRCh38
rs184959520 Chr.19: 46920962-46920962 on Build GRCh38
rs372676106 Chr.19: 46921099-46921099 on Build GRCh38
rs561026956 Chr.19: 46921096-46921096 on Build GRCh38
rs1054531443 Chr.19: 46921016-46921016 on Build GRCh38
rs780973219 Chr.19: 46921125-46921125 on Build GRCh38
rs763897284 Chr.19: 46921248-46921248 on Build GRCh38
rs751173660 Chr.19: 46921104-46921104 on Build GRCh38
rs562764311 Chr.19: 46921224-46921224 on Build GRCh38
rs765823240 Chr.19: 46921088-46921088 on Build GRCh38
rs773808872 Chr.19: 46921189-46921189 on Build GRCh38
rs775752352 Chr.19: 46921041-46921041 on Build GRCh38
rs755961782 Chr.19: 46921145-46921145 on Build GRCh38
rs375899794 Chr.19: 46920817-46920817 on Build GRCh38
rs773747975 Chr.19: 46921219-46921219 on Build GRCh38
rs755879808 Chr.19: 46921241-46921241 on Build GRCh38
rs761081196 Chr.19: 46920905-46920905 on Build GRCh38
rs751208887 Chr.19: 46920995-46920995 on Build GRCh38
rs765836598 Chr.19: 46921103-46921103 on Build GRCh38
rs765848742 Chr.19: 46921206-46921206 on Build GRCh38
rs545335379 Chr.19: 46920929-46920929 on Build GRCh38
rs763587380 Chr.19: 46921205-46921205 on Build GRCh38
rs151024661 Chr.19: 46920790-46920790 on Build GRCh38
rs762331167 Chr.19: 46921087-46921087 on Build GRCh38
rs769727967 Chr.19: 46921010-46921010 on Build GRCh38
rs774427329 Chr.19: 46920883-46920883 on Build GRCh38
rs780026082 Chr.19: 46921169-46921169 on Build GRCh38
rs542396381 Chr.19: 46921079-46921079 on Build GRCh38
rs780120522 Chr.19: 46921286-46921286 on Build GRCh38
rs748496375 Chr.19: 46921178-46921178 on Build GRCh38
rs779916715 Chr.19: 46920813-46920813 on Build GRCh38
rs776981898 Chr.19: 46921176-46921176 on Build GRCh38
rs756762988 Chr.19: 46920789-46920789 on Build GRCh38
rs748066588 Chr.19: 46920863-46920863 on Build GRCh38
rs759688841 Chr.19: 46920887-46920887 on Build GRCh38

View Full Product Details