Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37951383-37951979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN2
Entrez Gene ID
151393
Gene Name
regulator of microtubule dynamics 2
Gene Aliases
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4
UniGene
Hs.591566
Gene Chromosome Location
Chr.2: 37920789-38068032 on Build GRCh38
Transcripts
NM_001170791.2 NM_001170792.2 NM_001170793.2 NM_001322211.1 NM_001322212.1 NM_144713.4 XM_011532613.2 XM_011532614.2 XM_011532615.2 XM_011532616.2 XM_011532617.2 XM_011532618.2 XM_011532619.2 XM_017003474.1 XM_017003475.1 XM_017003476.1 XM_017003477.1 XM_017003478.1 XM_017003479.1 NR_102712.1

Gene Symbol
RMDN2-AS1
Entrez Gene ID
101410544
Gene Name
RMDN2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.591566
Gene Chromosome Location
Chr.2: 37950334-38036341 on Build GRCh38
Transcripts
NM_001170791.2 NM_001170792.2 NM_001170793.2 NM_001322211.1 NM_001322212.1 NM_144713.4 XM_011532613.2 XM_011532614.2 XM_011532615.2 XM_011532616.2 XM_011532617.2 XM_011532618.2 XM_011532619.2 XM_017003474.1 XM_017003475.1 XM_017003476.1 XM_017003477.1 XM_017003478.1 XM_017003479.1 NR_102712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3581462 c.191T>C p.L64S Chr.2: 37951406-37951406 on Build GRCh38
7224205 c.395T>G p.L132R Chr.2: 37951610-37951610 on Build GRCh38
3581463 c.407C>T p.S136F Chr.2: 37951622-37951622 on Build GRCh38
1020786 c.437C>A p.P146H Chr.2: 37951652-37951652 on Build GRCh38
7164770 c.233C>G p.S78C Chr.2: 37951448-37951448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749362786 Chr.2: 37951553-37951553 on Build GRCh38
rs199847199 Chr.2: 37951854-37951854 on Build GRCh38
rs375054023 Chr.2: 37951750-37951750 on Build GRCh38
rs777954963 Chr.2: 37951613-37951613 on Build GRCh38
rs779968226 Chr.2: 37951634-37951634 on Build GRCh38
rs949329094 Chr.2: 37951515-37951515 on Build GRCh38
rs781418185 Chr.2: 37951428-37951428 on Build GRCh38
rs775026429 Chr.2: 37951571-37951571 on Build GRCh38
rs773267565 Chr.2: 37951805-37951805 on Build GRCh38
rs766619098 Chr.2: 37951812-37951812 on Build GRCh38
rs768531525 Chr.2: 37951714-37951714 on Build GRCh38
rs762391532 Chr.2: 37951802-37951804 on Build GRCh38
rs761444515 Chr.2: 37951659-37951659 on Build GRCh38
rs765502790 Chr.2: 37951799-37951799 on Build GRCh38
rs766060278 Chr.2: 37951847-37951847 on Build GRCh38
rs146307813 Chr.2: 37951573-37951573 on Build GRCh38
rs561874859 Chr.2: 37951460-37951460 on Build GRCh38
rs550643620 Chr.2: 37951754-37951754 on Build GRCh38
rs763128591 Chr.2: 37951871-37951871 on Build GRCh38
rs780608119 Chr.2: 37951410-37951410 on Build GRCh38
rs753115870 Chr.2: 37951688-37951688 on Build GRCh38
rs757260934 Chr.2: 37951619-37951619 on Build GRCh38
rs750531666 Chr.2: 37951761-37951761 on Build GRCh38
rs147729503 Chr.2: 37951721-37951721 on Build GRCh38
rs781017185 Chr.2: 37951720-37951720 on Build GRCh38
rs141587026 Chr.2: 37951530-37951530 on Build GRCh38
rs777057841 Chr.2: 37951853-37951853 on Build GRCh38
rs769302943 Chr.2: 37951664-37951664 on Build GRCh38
rs372320363 Chr.2: 37951942-37951942 on Build GRCh38
rs375825575 Chr.2: 37951945-37951945 on Build GRCh38
rs747397759 Chr.2: 37951413-37951413 on Build GRCh38
rs80125475 Chr.2: 37951795-37951795 on Build GRCh38
rs386645084 Chr.2: 37951914-37951915 on Build GRCh38
rs1036920349 Chr.2: 37951643-37951643 on Build GRCh38
rs368723250 Chr.2: 37951886-37951886 on Build GRCh38
rs778523677 Chr.2: 37951911-37951911 on Build GRCh38
rs1049712699 Chr.2: 37951562-37951562 on Build GRCh38
rs754018418 Chr.2: 37951693-37951693 on Build GRCh38
rs75699143 Chr.2: 37951734-37951734 on Build GRCh38
rs111419892 Chr.2: 37951603-37951603 on Build GRCh38
rs759357021 Chr.2: 37951476-37951476 on Build GRCh38
rs777230222 Chr.2: 37951782-37951782 on Build GRCh38
rs140094166 Chr.2: 37951564-37951564 on Build GRCh38
rs761349993 Chr.2: 37951583-37951583 on Build GRCh38
rs142222765 Chr.2: 37951580-37951580 on Build GRCh38
rs201742730 Chr.2: 37951823-37951823 on Build GRCh38
rs371278815 Chr.2: 37951840-37951840 on Build GRCh38
rs868329463 Chr.2: 37951717-37951717 on Build GRCh38
rs768819143 Chr.2: 37951426-37951426 on Build GRCh38
rs749654593 Chr.2: 37951940-37951940 on Build GRCh38
rs763646716 Chr.2: 37951491-37951491 on Build GRCh38
rs962014959 Chr.2: 37951708-37951708 on Build GRCh38
rs147584543 Chr.2: 37951591-37951591 on Build GRCh38
rs375006700 Chr.2: 37951877-37951877 on Build GRCh38
rs370261611 Chr.2: 37951602-37951602 on Build GRCh38
rs776357587 Chr.2: 37951657-37951657 on Build GRCh38
rs768437569 Chr.2: 37951648-37951648 on Build GRCh38
rs75764221 Chr.2: 37951822-37951822 on Build GRCh38
rs757646117 Chr.2: 37951950-37951950 on Build GRCh38
rs778068513 Chr.2: 37951935-37951935 on Build GRCh38
rs145655316 Chr.2: 37951542-37951542 on Build GRCh38
rs756911962 Chr.2: 37951836-37951836 on Build GRCh38
rs142128542 Chr.2: 37951891-37951891 on Build GRCh38
rs751497436 Chr.2: 37951772-37951772 on Build GRCh38
rs777509798 Chr.2: 37951829-37951829 on Build GRCh38
rs746789696 Chr.2: 37951647-37951647 on Build GRCh38
rs368387990 Chr.2: 37951758-37951758 on Build GRCh38
rs200577012 Chr.2: 37951826-37951826 on Build GRCh38
rs772576983 Chr.2: 37951669-37951669 on Build GRCh38
rs767903168 Chr.2: 37951686-37951686 on Build GRCh38
rs766980027 Chr.2: 37951505-37951505 on Build GRCh38
rs539510009 Chr.2: 37951895-37951899 on Build GRCh38
rs773157930 Chr.2: 37951441-37951441 on Build GRCh38
rs143396477 Chr.2: 37951500-37951500 on Build GRCh38
rs760132190 Chr.2: 37951488-37951488 on Build GRCh38
rs746805856 Chr.2: 37951713-37951713 on Build GRCh38
rs773607549 Chr.2: 37951867-37951867 on Build GRCh38
rs879232653 Chr.2: 37951477-37951477 on Build GRCh38
rs990472565 Chr.2: 37951461-37951461 on Build GRCh38
rs764392789 Chr.2: 37951751-37951751 on Build GRCh38
rs745653440 Chr.2: 37951560-37951560 on Build GRCh38
rs540325773 Chr.2: 37951796-37951796 on Build GRCh38
rs148490802 Chr.2: 37951890-37951890 on Build GRCh38
rs867602558 Chr.2: 37951705-37951708 on Build GRCh38
rs77881371 Chr.2: 37951914-37951914 on Build GRCh38
rs939707723 Chr.2: 37951804-37951804 on Build GRCh38
rs769303162 Chr.2: 37951722-37951722 on Build GRCh38
rs754948686 Chr.2: 37951774-37951774 on Build GRCh38
rs775905089 Chr.2: 37951737-37951737 on Build GRCh38
rs756970764 Chr.2: 37951907-37951907 on Build GRCh38
rs751663535 Chr.2: 37951819-37951819 on Build GRCh38
rs528990994 Chr.2: 37951617-37951617 on Build GRCh38
rs140903543 Chr.2: 37951765-37951765 on Build GRCh38
rs200104184 Chr.2: 37951692-37951692 on Build GRCh38
rs775998944 Chr.2: 37951791-37951791 on Build GRCh38
rs138276197 Chr.2: 37951532-37951532 on Build GRCh38
rs1014624575 Chr.2: 37951844-37951844 on Build GRCh38
rs779799506 Chr.2: 37951712-37951712 on Build GRCh38
rs148126266 Chr.2: 37951950-37951950 on Build GRCh38
rs890818261 Chr.2: 37951609-37951609 on Build GRCh38
rs748653362 Chr.2: 37951925-37951925 on Build GRCh38
rs777868708 Chr.2: 37951544-37951544 on Build GRCh38
rs771880876 Chr.2: 37951633-37951633 on Build GRCh38
rs760132907 Chr.2: 37951674-37951674 on Build GRCh38
rs747954992 Chr.2: 37951778-37951778 on Build GRCh38
rs201632669 Chr.2: 37951865-37951865 on Build GRCh38
rs774634461 Chr.2: 37951885-37951885 on Build GRCh38
rs748185366 Chr.2: 37951433-37951433 on Build GRCh38
rs772731544 Chr.2: 37951736-37951737 on Build GRCh38
rs201710885 Chr.2: 37951797-37951797 on Build GRCh38
rs755850354 Chr.2: 37951780-37951780 on Build GRCh38
rs758420186 Chr.2: 37951711-37951711 on Build GRCh38
rs765884102 Chr.2: 37951481-37951481 on Build GRCh38
rs372732058 Chr.2: 37951794-37951794 on Build GRCh38
rs200509230 Chr.2: 37951762-37951762 on Build GRCh38
rs749269339 Chr.2: 37951448-37951448 on Build GRCh38
rs146089847 Chr.2: 37951538-37951538 on Build GRCh38
rs558576469 Chr.2: 37951623-37951623 on Build GRCh38
rs1051283312 Chr.2: 37951601-37951601 on Build GRCh38
rs751527735 Chr.2: 37951684-37951684 on Build GRCh38
rs554716987 Chr.2: 37951789-37951789 on Build GRCh38
rs371165501 Chr.2: 37951622-37951622 on Build GRCh38
rs767089259 Chr.2: 37951480-37951480 on Build GRCh38
rs140287039 Chr.2: 37951582-37951582 on Build GRCh38
rs138849623 Chr.2: 37951788-37951788 on Build GRCh38
rs779097034 Chr.2: 37951698-37951698 on Build GRCh38
rs761706403 Chr.2: 37951640-37951640 on Build GRCh38
rs745377126 Chr.2: 37951851-37951851 on Build GRCh38
rs370260752 Chr.2: 37951541-37951541 on Build GRCh38
rs921018853 Chr.2: 37951577-37951577 on Build GRCh38
rs532901254 Chr.2: 37951439-37951439 on Build GRCh38
rs538590900 Chr.2: 37951727-37951727 on Build GRCh38
rs116259556 Chr.2: 37951929-37951929 on Build GRCh38
rs575374530 Chr.2: 37951736-37951736 on Build GRCh38
rs548414832 Chr.2: 37951558-37951558 on Build GRCh38
rs756392960 Chr.2: 37951604-37951604 on Build GRCh38
rs551243864 Chr.2: 37951472-37951472 on Build GRCh38
rs371164410 Chr.2: 37951592-37951592 on Build GRCh38
rs267599371 Chr.2: 37951482-37951482 on Build GRCh38
rs78569430 Chr.2: 37951915-37951915 on Build GRCh38
rs766563880 Chr.2: 37951624-37951624 on Build GRCh38
rs781175040 Chr.2: 37951777-37951777 on Build GRCh38
rs958019785 Chr.2: 37951898-37951898 on Build GRCh38
rs202163735 Chr.2: 37951947-37951947 on Build GRCh38
rs765117398 Chr.2: 37951677-37951681 on Build GRCh38
rs770821482 Chr.2: 37951471-37951471 on Build GRCh38
rs138663377 Chr.2: 37951651-37951651 on Build GRCh38
rs1003603915 Chr.2: 37951833-37951833 on Build GRCh38
rs763116818 Chr.2: 37951810-37951810 on Build GRCh38
rs753369400 Chr.2: 37951499-37951499 on Build GRCh38
rs753635611 Chr.2: 37951895-37951895 on Build GRCh38
rs115807310 Chr.2: 37951913-37951913 on Build GRCh38
rs763134386 Chr.2: 37951867-37951870 on Build GRCh38
rs148178880 Chr.2: 37951760-37951760 on Build GRCh38
rs749380869 Chr.2: 37951614-37951614 on Build GRCh38

View Full Product Details