Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111739047-111739322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK4
Entrez Gene ID
9732
Gene Name
dedicator of cytokinesis 4
Gene Aliases
-
UniGene
Hs.654652
Gene Chromosome Location
Chr.7: 111726108-112206407 on Build GRCh38
Transcripts
NM_014705.3 XM_006716188.2 XM_006716189.2 XM_017012819.1 XM_017012820.1 XM_017012821.1 XM_017012822.1 XM_017012823.1 XM_017012824.1 XM_017012825.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5935263 c.1854+6C>T p.? Chr.7: 111739128-111739128 on Build GRCh38
6420128 c.1844A>G p.E615G Chr.7: 111739144-111739144 on Build GRCh38
7137264 c.5195A>T p.E1732V Chr.7: 111739144-111739144 on Build GRCh38
5935262 c.5196+6C>T p.? Chr.7: 111739128-111739128 on Build GRCh38
421641 c.5121C>A p.S1707S Chr.7: 111739209-111739209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778040550 Chr.7: 111739202-111739202 on Build GRCh38
rs199794600 Chr.7: 111739197-111739197 on Build GRCh38
rs750089233 Chr.7: 111739285-111739285 on Build GRCh38
rs886581684 Chr.7: 111739173-111739173 on Build GRCh38
rs200275495 Chr.7: 111739119-111739119 on Build GRCh38
rs772451167 Chr.7: 111739107-111739107 on Build GRCh38
rs942501308 Chr.7: 111739116-111739116 on Build GRCh38
rs771296881 Chr.7: 111739215-111739215 on Build GRCh38
rs370179180 Chr.7: 111739179-111739179 on Build GRCh38
rs1015873212 Chr.7: 111739079-111739079 on Build GRCh38
rs771255631 Chr.7: 111739138-111739138 on Build GRCh38
rs879807345 Chr.7: 111739249-111739249 on Build GRCh38
rs753789003 Chr.7: 111739191-111739191 on Build GRCh38
rs767075249 Chr.7: 111739282-111739282 on Build GRCh38
rs773787936 Chr.7: 111739261-111739261 on Build GRCh38
rs989356663 Chr.7: 111739086-111739086 on Build GRCh38
rs760039214 Chr.7: 111739234-111739234 on Build GRCh38
rs10258414 Chr.7: 111739094-111739094 on Build GRCh38
rs761330263 Chr.7: 111739265-111739265 on Build GRCh38
rs878987083 Chr.7: 111739089-111739089 on Build GRCh38
rs754120556 Chr.7: 111739252-111739254 on Build GRCh38
rs531986343 Chr.7: 111739137-111739137 on Build GRCh38
rs913109761 Chr.7: 111739208-111739208 on Build GRCh38
rs755049992 Chr.7: 111739257-111739258 on Build GRCh38
rs150569245 Chr.7: 111739142-111739142 on Build GRCh38
rs768563732 Chr.7: 111739195-111739195 on Build GRCh38
rs1023601108 Chr.7: 111739254-111739254 on Build GRCh38
rs779750390 Chr.7: 111739189-111739189 on Build GRCh38
rs760291354 Chr.7: 111739289-111739289 on Build GRCh38
rs1031320659 Chr.7: 111739278-111739278 on Build GRCh38
rs777190065 Chr.7: 111739141-111739141 on Build GRCh38
rs750130626 Chr.7: 111739162-111739162 on Build GRCh38
rs776911255 Chr.7: 111739220-111739220 on Build GRCh38
rs772362224 Chr.7: 111739198-111739198 on Build GRCh38
rs139512717 Chr.7: 111739258-111739258 on Build GRCh38
rs774620217 Chr.7: 111739099-111739099 on Build GRCh38
rs747259595 Chr.7: 111739207-111739207 on Build GRCh38
rs766146100 Chr.7: 111739299-111739299 on Build GRCh38
rs778867728 Chr.7: 111739257-111739260 on Build GRCh38
rs746855415 Chr.7: 111739194-111739194 on Build GRCh38
rs1003430622 Chr.7: 111739259-111739259 on Build GRCh38
rs767165415 Chr.7: 111739158-111739158 on Build GRCh38
rs780235664 Chr.7: 111739213-111739218 on Build GRCh38
rs956758475 Chr.7: 111739085-111739085 on Build GRCh38
rs751093222 Chr.7: 111739151-111739151 on Build GRCh38
rs754983694 Chr.7: 111739193-111739193 on Build GRCh38
rs756509239 Chr.7: 111739185-111739187 on Build GRCh38
rs547482522 Chr.7: 111739102-111739102 on Build GRCh38
rs1012345100 Chr.7: 111739235-111739235 on Build GRCh38

View Full Product Details