Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17077629-17078054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1 XM_017013343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097440 c.581T>C p.M194T Chr.8: 17077796-17077796 on Build GRCh38
1097441 c.640G>T p.E214* Chr.8: 17077855-17077855 on Build GRCh38
138671 c.589G>A p.E197K Chr.8: 17077804-17077804 on Build GRCh38
3898474 c.586G>C p.A196P Chr.8: 17077801-17077801 on Build GRCh38
3898475 c.617C>T p.S206L Chr.8: 17077832-17077832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745455177 Chr.8: 17077860-17077860 on Build GRCh38
rs772658345 Chr.8: 17077776-17077776 on Build GRCh38
rs761900190 Chr.8: 17077907-17077907 on Build GRCh38
rs781264663 Chr.8: 17077893-17077893 on Build GRCh38
rs757209670 Chr.8: 17077885-17077886 on Build GRCh38
rs911760500 Chr.8: 17077966-17077966 on Build GRCh38
rs35482790 Chr.8: 17077892-17077892 on Build GRCh38
rs577314553 Chr.8: 17077973-17077973 on Build GRCh38
rs373142084 Chr.8: 17077739-17077739 on Build GRCh38
rs768616883 Chr.8: 17077887-17077887 on Build GRCh38
rs574837028 Chr.8: 17077822-17077822 on Build GRCh38
rs150021056 Chr.8: 17077843-17077843 on Build GRCh38
rs775149413 Chr.8: 17077803-17077803 on Build GRCh38
rs112905373 Chr.8: 17077865-17077865 on Build GRCh38
rs748701997 Chr.8: 17077848-17077848 on Build GRCh38
rs368380392 Chr.8: 17077754-17077754 on Build GRCh38
rs971952623 Chr.8: 17077988-17077988 on Build GRCh38
rs1048416662 Chr.8: 17078026-17078026 on Build GRCh38
rs750645681 Chr.8: 17077830-17077830 on Build GRCh38
rs760599266 Chr.8: 17077805-17077805 on Build GRCh38
rs920526424 Chr.8: 17077996-17077996 on Build GRCh38
rs534901761 Chr.8: 17077726-17077726 on Build GRCh38
rs114950634 Chr.8: 17077986-17077986 on Build GRCh38
rs980460214 Chr.8: 17078022-17078022 on Build GRCh38
rs901373623 Chr.8: 17077920-17077920 on Build GRCh38
rs928475270 Chr.8: 17078027-17078027 on Build GRCh38
rs746121129 Chr.8: 17077759-17077759 on Build GRCh38
rs758768309 Chr.8: 17077762-17077762 on Build GRCh38
rs1010410931 Chr.8: 17077779-17077779 on Build GRCh38
rs776690591 Chr.8: 17077891-17077891 on Build GRCh38
rs117597341 Chr.8: 17078015-17078015 on Build GRCh38
rs780400878 Chr.8: 17077837-17077837 on Build GRCh38
rs760580954 Chr.8: 17077886-17077886 on Build GRCh38
rs377331426 Chr.8: 17077772-17077772 on Build GRCh38
rs764152116 Chr.8: 17077819-17077819 on Build GRCh38
rs537594168 Chr.8: 17077829-17077829 on Build GRCh38
rs1037024191 Chr.8: 17077703-17077703 on Build GRCh38
rs897123759 Chr.8: 17077704-17077704 on Build GRCh38
rs751884530 Chr.8: 17077839-17077839 on Build GRCh38
rs748826347 Chr.8: 17077780-17077780 on Build GRCh38
rs557035254 Chr.8: 17077924-17077924 on Build GRCh38
rs770410801 Chr.8: 17077849-17077849 on Build GRCh38
rs950490659 Chr.8: 17077982-17077982 on Build GRCh38
rs1029147043 Chr.8: 17077943-17077943 on Build GRCh38
rs184515644 Chr.8: 17077800-17077800 on Build GRCh38
rs770467999 Chr.8: 17077798-17077798 on Build GRCh38
rs997264521 Chr.8: 17077960-17077960 on Build GRCh38
rs373857886 Chr.8: 17077823-17077823 on Build GRCh38
rs199805391 Chr.8: 17077856-17077856 on Build GRCh38
rs775308871 Chr.8: 17077884-17077884 on Build GRCh38
rs928496458 Chr.8: 17077723-17077723 on Build GRCh38
rs1007557673 Chr.8: 17077878-17077878 on Build GRCh38
rs969944895 Chr.8: 17078017-17078017 on Build GRCh38
rs758615280 Chr.8: 17077833-17077833 on Build GRCh38
rs747548706 Chr.8: 17077768-17077768 on Build GRCh38
rs771633578 Chr.8: 17077872-17077872 on Build GRCh38
rs769934583 Chr.8: 17077767-17077767 on Build GRCh38
rs953177433 Chr.8: 17077947-17077947 on Build GRCh38
rs10098337 Chr.8: 17077725-17077725 on Build GRCh38
rs753982818 Chr.8: 17077746-17077746 on Build GRCh38
rs767335819 Chr.8: 17077801-17077801 on Build GRCh38
rs752763691 Chr.8: 17077856-17077856 on Build GRCh38
rs565232734 Chr.8: 17077844-17077844 on Build GRCh38
rs765252711 Chr.8: 17077844-17077845 on Build GRCh38
rs1041215986 Chr.8: 17077910-17077910 on Build GRCh38
rs111571635 Chr.8: 17077928-17077928 on Build GRCh38
rs765403122 Chr.8: 17077909-17077909 on Build GRCh38
rs545296363 Chr.8: 17077985-17077985 on Build GRCh38
rs1026507708 Chr.8: 17077961-17077961 on Build GRCh38
rs572404701 Chr.8: 17077994-17077994 on Build GRCh38
rs1002093176 Chr.8: 17077728-17077728 on Build GRCh38

View Full Product Details