Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7668984-7669521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R2
Entrez Gene ID
51700
Gene Name
cytochrome b5 reductase 2
Gene Aliases
B5R.2
UniGene
Hs.414362
Gene Chromosome Location
Chr.11: 7665095-7678568 on Build GRCh38
Transcripts
NM_001302826.1 NM_001302827.1 NM_016229.4 NR_126508.1 XM_005252975.4 XM_006718251.2 XM_011520184.2 XM_011520185.2 XM_017017920.1 XM_017017921.1 XM_017017922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444371 c.367C>T p.R123C Chr.11: 7669226-7669226 on Build GRCh38
6638749 c.344T>C p.I115T Chr.11: 7669249-7669249 on Build GRCh38
4441937 c.311A>C p.Q104P Chr.11: 7669282-7669282 on Build GRCh38
4894672 c.281C>T p.P94L Chr.11: 7669312-7669312 on Build GRCh38
4876941 c.334G>A p.G112R Chr.11: 7669259-7669259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760036370 Chr.11: 7669031-7669031 on Build GRCh38
rs780229893 Chr.11: 7669201-7669201 on Build GRCh38
rs777485658 Chr.11: 7669280-7669283 on Build GRCh38
rs192817196 Chr.11: 7669017-7669017 on Build GRCh38
rs755053589 Chr.11: 7669372-7669372 on Build GRCh38
rs545456393 Chr.11: 7669466-7669466 on Build GRCh38
rs772515234 Chr.11: 7669137-7669137 on Build GRCh38
rs141969366 Chr.11: 7669241-7669241 on Build GRCh38
rs777110593 Chr.11: 7669144-7669144 on Build GRCh38
rs746445836 Chr.11: 7669377-7669378 on Build GRCh38
rs1042385593 Chr.11: 7669418-7669418 on Build GRCh38
rs757072875 Chr.11: 7669244-7669244 on Build GRCh38
rs772568201 Chr.11: 7669032-7669032 on Build GRCh38
rs1045185225 Chr.11: 7669192-7669192 on Build GRCh38
rs933770406 Chr.11: 7669033-7669033 on Build GRCh38
rs752498345 Chr.11: 7669113-7669113 on Build GRCh38
rs760467155 Chr.11: 7669238-7669238 on Build GRCh38
rs182092197 Chr.11: 7669252-7669252 on Build GRCh38
rs1012015492 Chr.11: 7669464-7669464 on Build GRCh38
rs774799960 Chr.11: 7669302-7669302 on Build GRCh38
rs761790193 Chr.11: 7669316-7669316 on Build GRCh38
rs574561525 Chr.11: 7669148-7669148 on Build GRCh38
rs902512918 Chr.11: 7669423-7669423 on Build GRCh38
rs765005001 Chr.11: 7669318-7669318 on Build GRCh38
rs918324006 Chr.11: 7669024-7669024 on Build GRCh38
rs756689273 Chr.11: 7669408-7669408 on Build GRCh38
rs200706480 Chr.11: 7669214-7669214 on Build GRCh38
rs766143055 Chr.11: 7669147-7669147 on Build GRCh38
rs986577492 Chr.11: 7669311-7669311 on Build GRCh38
rs988633958 Chr.11: 7669060-7669060 on Build GRCh38
rs565415073 Chr.11: 7669486-7669486 on Build GRCh38
rs545206907 Chr.11: 7669240-7669240 on Build GRCh38
rs778897640 Chr.11: 7669139-7669139 on Build GRCh38
rs533756199 Chr.11: 7669142-7669142 on Build GRCh38
rs776802534 Chr.11: 7669219-7669219 on Build GRCh38
rs775770139 Chr.11: 7669034-7669034 on Build GRCh38
rs747457495 Chr.11: 7669205-7669205 on Build GRCh38
rs762714248 Chr.11: 7669070-7669070 on Build GRCh38
rs117283049 Chr.11: 7669396-7669396 on Build GRCh38
rs753849233 Chr.11: 7669060-7669061 on Build GRCh38
rs777675618 Chr.11: 7669124-7669124 on Build GRCh38
rs774475677 Chr.11: 7669028-7669028 on Build GRCh38
rs1039244129 Chr.11: 7669389-7669389 on Build GRCh38
rs750630341 Chr.11: 7669108-7669108 on Build GRCh38
rs749567469 Chr.11: 7669006-7669006 on Build GRCh38
rs962632087 Chr.11: 7669259-7669259 on Build GRCh38
rs748545774 Chr.11: 7669221-7669221 on Build GRCh38
rs756749128 Chr.11: 7669379-7669379 on Build GRCh38
rs368476375 Chr.11: 7669260-7669260 on Build GRCh38
rs543700517 Chr.11: 7669463-7669463 on Build GRCh38
rs745906044 Chr.11: 7669181-7669181 on Build GRCh38
rs764935054 Chr.11: 7669439-7669441 on Build GRCh38
rs145877593 Chr.11: 7669223-7669223 on Build GRCh38
rs745507305 Chr.11: 7669290-7669290 on Build GRCh38
rs115981610 Chr.11: 7669059-7669059 on Build GRCh38
rs748031216 Chr.11: 7669092-7669094 on Build GRCh38
rs1016738024 Chr.11: 7669487-7669487 on Build GRCh38
rs1047325547 Chr.11: 7669421-7669421 on Build GRCh38
rs779126801 Chr.11: 7669177-7669177 on Build GRCh38
rs769272217 Chr.11: 7669212-7669212 on Build GRCh38
rs776521770 Chr.11: 7669312-7669312 on Build GRCh38
rs755390101 Chr.11: 7669274-7669274 on Build GRCh38
rs910996309 Chr.11: 7669270-7669270 on Build GRCh38
rs910998956 Chr.11: 7669330-7669330 on Build GRCh38
rs751552393 Chr.11: 7669368-7669368 on Build GRCh38
rs1034970611 Chr.11: 7669218-7669218 on Build GRCh38
rs758426359 Chr.11: 7669153-7669153 on Build GRCh38
rs562842973 Chr.11: 7669276-7669276 on Build GRCh38
rs755837341 Chr.11: 7669121-7669121 on Build GRCh38
rs1006098990 Chr.11: 7669185-7669185 on Build GRCh38
rs972345016 Chr.11: 7669268-7669268 on Build GRCh38
rs1052340316 Chr.11: 7669052-7669052 on Build GRCh38
rs913078012 Chr.11: 7669074-7669074 on Build GRCh38
rs752557121 Chr.11: 7669156-7669156 on Build GRCh38
rs771756683 Chr.11: 7669292-7669292 on Build GRCh38
rs150689769 Chr.11: 7669285-7669285 on Build GRCh38
rs188240040 Chr.11: 7669173-7669173 on Build GRCh38
rs933695948 Chr.11: 7669288-7669288 on Build GRCh38
rs762690495 Chr.11: 7669145-7669145 on Build GRCh38
rs375870163 Chr.11: 7669225-7669225 on Build GRCh38
rs549921306 Chr.11: 7669057-7669057 on Build GRCh38
rs750010780 Chr.11: 7669069-7669069 on Build GRCh38
rs894103755 Chr.11: 7669130-7669130 on Build GRCh38
rs112078510 Chr.11: 7669101-7669101 on Build GRCh38
rs530283483 Chr.11: 7669459-7669459 on Build GRCh38
rs372890518 Chr.11: 7669284-7669284 on Build GRCh38
rs753240895 Chr.11: 7669378-7669378 on Build GRCh38
rs771617201 Chr.11: 7669247-7669248 on Build GRCh38
rs982069659 Chr.11: 7669029-7669029 on Build GRCh38
rs148127253 Chr.11: 7669226-7669226 on Build GRCh38
rs778821591 Chr.11: 7669083-7669086 on Build GRCh38
rs183821192 Chr.11: 7669164-7669164 on Build GRCh38
rs138970017 Chr.11: 7669308-7669308 on Build GRCh38
rs781177307 Chr.11: 7669375-7669375 on Build GRCh38
rs766925880 Chr.11: 7669230-7669230 on Build GRCh38
rs754782223 Chr.11: 7669065-7669067 on Build GRCh38
rs775136382 Chr.11: 7669236-7669236 on Build GRCh38
rs763019913 Chr.11: 7669355-7669355 on Build GRCh38
rs894048147 Chr.11: 7669391-7669391 on Build GRCh38
rs905272059 Chr.11: 7669194-7669194 on Build GRCh38
rs959243247 Chr.11: 7669178-7669178 on Build GRCh38
rs375222567 Chr.11: 7669360-7669360 on Build GRCh38
rs781301373 Chr.11: 7669110-7669110 on Build GRCh38
rs1045638888 Chr.11: 7669460-7669460 on Build GRCh38
rs551444263 Chr.11: 7669107-7669107 on Build GRCh38
rs749461764 Chr.11: 7669133-7669133 on Build GRCh38
rs767861170 Chr.11: 7669064-7669064 on Build GRCh38
rs750623422 Chr.11: 7669255-7669255 on Build GRCh38
rs780082835 Chr.11: 7669020-7669021 on Build GRCh38
rs750286511 Chr.11: 7669337-7669337 on Build GRCh38
rs962579968 Chr.11: 7669488-7669488 on Build GRCh38
rs7131416 Chr.11: 7669136-7669136 on Build GRCh38
rs747117625 Chr.11: 7669264-7669264 on Build GRCh38
rs773767925 Chr.11: 7669286-7669286 on Build GRCh38
rs756138369 Chr.11: 7669157-7669157 on Build GRCh38
rs767178237 Chr.11: 7669154-7669154 on Build GRCh38
rs758439111 Chr.11: 7669187-7669187 on Build GRCh38
rs4631871 Chr.11: 7669454-7669454 on Build GRCh38
rs765974177 Chr.11: 7669087-7669087 on Build GRCh38

View Full Product Details