Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48758773-48759323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Gene Symbol
BEND5
Entrez Gene ID
79656
Gene Name
BEN domain containing 5
Gene Aliases
C1orf165
UniGene
Hs.475348
Gene Chromosome Location
Chr.1: 48727523-48778603 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4454783 c.578A>C p.Q193P Chr.1: 48759067-48759067 on Build GRCh38
5629643 c.364C>A p.P122T Chr.1: 48759281-48759281 on Build GRCh38
6611267 c.690G>A p.E230E Chr.1: 48758955-48758955 on Build GRCh38
1741533 c.491C>T p.S164L Chr.1: 48759154-48759154 on Build GRCh38
6334741 c.231T>A p.L77L Chr.1: 48758907-48758907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367903944 Chr.1: 48759148-48759148 on Build GRCh38
rs773513345 Chr.1: 48759183-48759183 on Build GRCh38
rs1042186251 Chr.1: 48759219-48759219 on Build GRCh38
rs377733697 Chr.1: 48758876-48758876 on Build GRCh38
rs201098567 Chr.1: 48759053-48759053 on Build GRCh38
rs202108114 Chr.1: 48759190-48759190 on Build GRCh38
rs769408885 Chr.1: 48758978-48758978 on Build GRCh38
rs756520704 Chr.1: 48759126-48759126 on Build GRCh38
rs764317457 Chr.1: 48759095-48759095 on Build GRCh38
rs772870853 Chr.1: 48758958-48758958 on Build GRCh38
rs750693064 Chr.1: 48759109-48759109 on Build GRCh38
rs531139897 Chr.1: 48759298-48759298 on Build GRCh38
rs777376627 Chr.1: 48759066-48759066 on Build GRCh38
rs138896175 Chr.1: 48759131-48759131 on Build GRCh38
rs766293637 Chr.1: 48758884-48758884 on Build GRCh38
rs942566888 Chr.1: 48759063-48759063 on Build GRCh38
rs779216654 Chr.1: 48759207-48759207 on Build GRCh38
rs752758809 Chr.1: 48759009-48759009 on Build GRCh38
rs376132991 Chr.1: 48758967-48758967 on Build GRCh38
rs553425088 Chr.1: 48758802-48758802 on Build GRCh38
rs750979533 Chr.1: 48759239-48759239 on Build GRCh38
rs12090576 Chr.1: 48758901-48758901 on Build GRCh38
rs765743070 Chr.1: 48759232-48759232 on Build GRCh38
rs1128934 Chr.1: 48758912-48758912 on Build GRCh38
rs777325753 Chr.1: 48759150-48759150 on Build GRCh38
rs375012612 Chr.1: 48758906-48758906 on Build GRCh38
rs150209705 Chr.1: 48759029-48759029 on Build GRCh38
rs757725667 Chr.1: 48758925-48758925 on Build GRCh38
rs758761416 Chr.1: 48758916-48758916 on Build GRCh38
rs962471023 Chr.1: 48759253-48759253 on Build GRCh38
rs764364656 Chr.1: 48759028-48759028 on Build GRCh38
rs986021311 Chr.1: 48759137-48759137 on Build GRCh38
rs879213736 Chr.1: 48758860-48758860 on Build GRCh38
rs540778259 Chr.1: 48759158-48759158 on Build GRCh38
rs544460557 Chr.1: 48758818-48758818 on Build GRCh38
rs748136013 Chr.1: 48759271-48759271 on Build GRCh38
rs763140197 Chr.1: 48759016-48759016 on Build GRCh38
rs769561530 Chr.1: 48759089-48759089 on Build GRCh38
rs767779765 Chr.1: 48759107-48759107 on Build GRCh38
rs760424879 Chr.1: 48758978-48758978 on Build GRCh38
rs759453573 Chr.1: 48758994-48758994 on Build GRCh38
rs999900027 Chr.1: 48759156-48759156 on Build GRCh38
rs767585722 Chr.1: 48759178-48759178 on Build GRCh38
rs900615772 Chr.1: 48759096-48759096 on Build GRCh38
rs749382544 Chr.1: 48759003-48759003 on Build GRCh38
rs771651589 Chr.1: 48758957-48758957 on Build GRCh38
rs370864690 Chr.1: 48759039-48759039 on Build GRCh38
rs768848202 Chr.1: 48759165-48759165 on Build GRCh38
rs369197717 Chr.1: 48759006-48759006 on Build GRCh38
rs768199056 Chr.1: 48758943-48758943 on Build GRCh38
rs776612748 Chr.1: 48758984-48758984 on Build GRCh38
rs780221438 Chr.1: 48758973-48758973 on Build GRCh38
rs776357116 Chr.1: 48759083-48759083 on Build GRCh38
rs772921005 Chr.1: 48758863-48758863 on Build GRCh38
rs201623671 Chr.1: 48759206-48759206 on Build GRCh38
rs760919716 Chr.1: 48759184-48759184 on Build GRCh38
rs763093466 Chr.1: 48759092-48759092 on Build GRCh38
rs748438575 Chr.1: 48759265-48759265 on Build GRCh38
rs773286114 Chr.1: 48759299-48759299 on Build GRCh38
rs145643434 Chr.1: 48758982-48758982 on Build GRCh38
rs749096188 Chr.1: 48759264-48759264 on Build GRCh38
rs770750910 Chr.1: 48759080-48759080 on Build GRCh38
rs142533057 Chr.1: 48759099-48759099 on Build GRCh38
rs74724453 Chr.1: 48758812-48758812 on Build GRCh38
rs1128934 Chr.1: 48758912-48758912 on Build GRCh38
rs781701119 Chr.1: 48758928-48758928 on Build GRCh38
rs746410856 Chr.1: 48758935-48758935 on Build GRCh38
rs751505805 Chr.1: 48759057-48759057 on Build GRCh38
rs746717048 Chr.1: 48759069-48759069 on Build GRCh38
rs778350481 Chr.1: 48758951-48758951 on Build GRCh38
rs754386085 Chr.1: 48759139-48759139 on Build GRCh38
rs750936942 Chr.1: 48759040-48759040 on Build GRCh38
rs148968303 Chr.1: 48759153-48759153 on Build GRCh38
rs775592004 Chr.1: 48759091-48759091 on Build GRCh38
rs747657670 Chr.1: 48758956-48758956 on Build GRCh38
rs746673782 Chr.1: 48759151-48759151 on Build GRCh38
rs755957106 Chr.1: 48759255-48759255 on Build GRCh38
rs759381131 Chr.1: 48759087-48759087 on Build GRCh38
rs889075951 Chr.1: 48758844-48758844 on Build GRCh38
rs904645045 Chr.1: 48759223-48759223 on Build GRCh38
rs780671877 Chr.1: 48759246-48759246 on Build GRCh38
rs774501741 Chr.1: 48759170-48759170 on Build GRCh38
rs745604168 Chr.1: 48759154-48759154 on Build GRCh38
rs1036126182 Chr.1: 48758796-48758796 on Build GRCh38
rs762007782 Chr.1: 48759106-48759106 on Build GRCh38
rs374150750 Chr.1: 48759164-48759164 on Build GRCh38
rs756507130 Chr.1: 48759052-48759052 on Build GRCh38
rs557411763 Chr.1: 48759252-48759252 on Build GRCh38
rs1023747124 Chr.1: 48759247-48759247 on Build GRCh38
rs973740518 Chr.1: 48758980-48758980 on Build GRCh38
rs771604369 Chr.1: 48759060-48759060 on Build GRCh38
rs958944295 Chr.1: 48759022-48759022 on Build GRCh38
rs780057963 Chr.1: 48759261-48759261 on Build GRCh38
rs756896304 Chr.1: 48759177-48759177 on Build GRCh38
rs369658674 Chr.1: 48758871-48758871 on Build GRCh38
rs753754037 Chr.1: 48758887-48758887 on Build GRCh38
rs144903085 Chr.1: 48758924-48758924 on Build GRCh38

View Full Product Details