Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56368198-56368775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210B
Entrez Gene ID
116151
Gene Name
family with sequence similarity 210 member B
Gene Aliases
5A3, C20orf108, dJ1167H4.1
UniGene
Hs.143736
Gene Chromosome Location
Chr.20: 56358927-56368663 on Build GRCh38
Transcripts
NM_080821.2

SNP Detail

SNP ID Chromosome Location
rs557328998 Chr.20: 56368753-56368753 on Build GRCh38
rs900178997 Chr.20: 56368709-56368709 on Build GRCh38
rs751043653 Chr.20: 56368673-56368673 on Build GRCh38
rs935748882 Chr.20: 56368498-56368498 on Build GRCh38
rs917518746 Chr.20: 56368454-56368454 on Build GRCh38
rs185728074 Chr.20: 56368236-56368236 on Build GRCh38
rs566231831 Chr.20: 56368596-56368596 on Build GRCh38
rs375572161 Chr.20: 56368351-56368351 on Build GRCh38
rs564325052 Chr.20: 56368385-56368385 on Build GRCh38
rs996824215 Chr.20: 56368574-56368574 on Build GRCh38
rs763687596 Chr.20: 56368558-56368558 on Build GRCh38
rs1014380416 Chr.20: 56368661-56368661 on Build GRCh38
rs529473716 Chr.20: 56368532-56368532 on Build GRCh38
rs186921166 Chr.20: 56368615-56368615 on Build GRCh38
rs760901349 Chr.20: 56368362-56368362 on Build GRCh38
rs897307837 Chr.20: 56368655-56368655 on Build GRCh38
rs536864103 Chr.20: 56368529-56368533 on Build GRCh38
rs370338761 Chr.20: 56368460-56368460 on Build GRCh38
rs191069232 Chr.20: 56368337-56368337 on Build GRCh38
rs952914141 Chr.20: 56368273-56368273 on Build GRCh38
rs771378726 Chr.20: 56368391-56368391 on Build GRCh38
rs76886634 Chr.20: 56368329-56368329 on Build GRCh38
rs1005579377 Chr.20: 56368607-56368607 on Build GRCh38
rs201748860 Chr.20: 56368275-56368275 on Build GRCh38
rs1015645487 Chr.20: 56368646-56368646 on Build GRCh38
rs182044580 Chr.20: 56368570-56368570 on Build GRCh38
rs1040289847 Chr.20: 56368541-56368543 on Build GRCh38
rs6024835 Chr.20: 56368531-56368531 on Build GRCh38
rs1033071289 Chr.20: 56368726-56368726 on Build GRCh38
rs533151176 Chr.20: 56368420-56368420 on Build GRCh38
rs1052877540 Chr.20: 56368520-56368520 on Build GRCh38
rs925677246 Chr.20: 56368494-56368494 on Build GRCh38
rs575435487 Chr.20: 56368697-56368697 on Build GRCh38
rs554206065 Chr.20: 56368285-56368285 on Build GRCh38
rs767511161 Chr.20: 56368730-56368730 on Build GRCh38
rs534514491 Chr.20: 56368272-56368272 on Build GRCh38
rs1050184245 Chr.20: 56368577-56368577 on Build GRCh38
rs144298554 Chr.20: 56368363-56368363 on Build GRCh38
rs993011726 Chr.20: 56368429-56368429 on Build GRCh38
rs571503492 Chr.20: 56368263-56368263 on Build GRCh38
rs1028367951 Chr.20: 56368231-56368231 on Build GRCh38
rs12150 Chr.20: 56368476-56368476 on Build GRCh38
rs148519468 Chr.20: 56368336-56368337 on Build GRCh38
rs559469117 Chr.20: 56368335-56368335 on Build GRCh38
rs41274724 Chr.20: 56368683-56368683 on Build GRCh38
rs762738436 Chr.20: 56368526-56368526 on Build GRCh38
rs191755543 Chr.20: 56368706-56368706 on Build GRCh38

View Full Product Details