Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 6444715-6445292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBS
Entrez Gene ID
875
Gene Name
cystathionine-beta-synthase
Gene Aliases
HIP4
UniGene
Hs.533013
Gene Chromosome Location
Chr.21: 43053190-43076868 on Build GRCh38
Transcripts
NM_000071.2 NM_001178008.2 NM_001178009.2 NM_001320298.1 NM_001321072.1 XM_011529773.2 XM_011529774.2 XM_011529775.2 XM_011529776.2 XM_011529777.1 XM_011529781.1 XM_011529782.1 XM_011529783.2 XM_017028489.1 XM_017028490.1 XM_017028491.1 XM_017028492.1 XM_017028493.1 NM_001321073.1 XM_011546094.1 XM_011546095.2 XM_011546097.2 XM_011546098.1 XM_011546099.1 XM_017028209.1 XM_017028210.1 XM_017028211.1 XM_017028212.1 XM_017028213.1 XM_017028214.1 XM_017028215.1 XM_017028216.1 XM_017028217.1 XM_017028218.1 XM_017028222.1 XM_017028224.1

Gene Symbol
CBSL
Entrez Gene ID
102724560
Gene Name
cystathionine-beta-synthase like
Gene Aliases
-
UniGene
Hs.533013
Gene Chromosome Location
Chr.21: 6444868-6468606 on Build GRCh38
Transcripts
NM_000071.2 NM_001178008.2 NM_001178009.2 NM_001320298.1 NM_001321072.1 XM_011529773.2 XM_011529774.2 XM_011529775.2 XM_011529776.2 XM_011529777.1 XM_011529781.1 XM_011529782.1 XM_011529783.2 XM_017028489.1 XM_017028490.1 XM_017028491.1 XM_017028492.1 XM_017028493.1 NM_001321073.1 XM_011546094.1 XM_011546095.2 XM_011546097.2 XM_011546098.1 XM_011546099.1 XM_017028209.1 XM_017028210.1 XM_017028211.1 XM_017028212.1 XM_017028213.1 XM_017028214.1 XM_017028215.1 XM_017028216.1 XM_017028217.1 XM_017028218.1 XM_017028222.1 XM_017028224.1

Gene Symbol
LOC102724843
Entrez Gene ID
102724843
Gene Name
uncharacterized LOC102724843
Gene Aliases
-
UniGene
Hs.743984
Gene Chromosome Location
Chr.21: 6318410-7087230 on Build GRCh38
Transcripts
NM_000071.2 NM_001178008.2 NM_001178009.2 NM_001320298.1 NM_001321072.1 XM_011529773.2 XM_011529774.2 XM_011529775.2 XM_011529776.2 XM_011529777.1 XM_011529781.1 XM_011529782.1 XM_011529783.2 XM_017028489.1 XM_017028490.1 XM_017028491.1 XM_017028492.1 XM_017028493.1 NM_001321073.1 XM_011546094.1 XM_011546095.2 XM_011546097.2 XM_011546098.1 XM_011546099.1 XM_017028209.1 XM_017028210.1 XM_017028211.1 XM_017028212.1 XM_017028213.1 XM_017028214.1 XM_017028215.1 XM_017028216.1 XM_017028217.1 XM_017028218.1 XM_017028222.1 XM_017028224.1

SNP Detail

SNP ID Chromosome Location
rs192889818 Chr.21: 43053582-43053582 on Build GRCh38
rs989199646 Chr.21: 43053262-43053262 on Build GRCh38
rs901065865 Chr.21: 43053409-43053409 on Build GRCh38
rs536375183 Chr.21: 43053546-43053546 on Build GRCh38
rs866356348 Chr.21: 43053288-43053288 on Build GRCh38
rs4987122 Chr.21: 43053584-43053584 on Build GRCh38
rs12626371 Chr.21: 43053337-43053337 on Build GRCh38
rs888430576 Chr.21: 43053365-43053365 on Build GRCh38
rs145080355 Chr.21: 43053093-43053093 on Build GRCh38
rs888512644 Chr.21: 43053555-43053555 on Build GRCh38
rs998021241 Chr.21: 43053594-43053594 on Build GRCh38
rs938925773 Chr.21: 43053504-43053504 on Build GRCh38
rs546027250 Chr.21: 43053350-43053350 on Build GRCh38
rs111413325 Chr.21: 43053199-43053199 on Build GRCh38
rs564495957 Chr.21: 43053435-43053435 on Build GRCh38
rs114892713 Chr.21: 43053130-43053130 on Build GRCh38
rs706208 Chr.21: 43053336-43053336 on Build GRCh38
rs953792933 Chr.21: 43053254-43053254 on Build GRCh38
rs367548583 Chr.21: 43053493-43053493 on Build GRCh38
rs1004093810 Chr.21: 43053180-43053180 on Build GRCh38
rs73372351 Chr.21: 43053136-43053136 on Build GRCh38
rs976975618 Chr.21: 43053230-43053230 on Build GRCh38
rs546587617 Chr.21: 43053459-43053459 on Build GRCh38
rs945057271 Chr.21: 43053291-43053291 on Build GRCh38
rs1057350399 Chr.21: 43053131-43053131 on Build GRCh38
rs914872117 Chr.21: 43053279-43053279 on Build GRCh38
rs1026978286 Chr.21: 43053168-43053168 on Build GRCh38
rs184082001 Chr.21: 43053113-43053113 on Build GRCh38
rs568843318 Chr.21: 43053111-43053111 on Build GRCh38
rs940141053 Chr.21: 43053573-43053573 on Build GRCh38
rs952830789 Chr.21: 43053169-43053169 on Build GRCh38
rs978142879 Chr.21: 43053499-43053499 on Build GRCh38
rs948884406 Chr.21: 43053550-43053550 on Build GRCh38
rs563366034 Chr.21: 43053277-43053277 on Build GRCh38
rs998096478 Chr.21: 43053424-43053424 on Build GRCh38
rs1015621641 Chr.21: 43053398-43053398 on Build GRCh38
rs927691463 Chr.21: 43053501-43053501 on Build GRCh38
rs1048480409 Chr.21: 43053554-43053555 on Build GRCh38
rs1013046093 Chr.21: 43053147-43053147 on Build GRCh38
rs1022016517 Chr.21: 43053495-43053495 on Build GRCh38
rs1041946509 Chr.21: 43053292-43053292 on Build GRCh38
rs375873791 Chr.21: 43053485-43053485 on Build GRCh38
rs532204095 Chr.21: 43053494-43053494 on Build GRCh38
rs187340100 Chr.21: 43053536-43053536 on Build GRCh38
rs921118061 Chr.21: 43053253-43053253 on Build GRCh38
rs706209 Chr.21: 43053315-43053315 on Build GRCh38
rs12613 Chr.21: 43053581-43053581 on Build GRCh38
rs894681582 Chr.21: 43053327-43053327 on Build GRCh38
rs372286324 Chr.21: 43053496-43053496 on Build GRCh38
rs1042366409 Chr.21: 43053112-43053112 on Build GRCh38
rs924844460 Chr.21: 43053296-43053296 on Build GRCh38
rs1028081090 Chr.21: 43053462-43053462 on Build GRCh38
rs1015945040 Chr.21: 43053194-43053194 on Build GRCh38
rs886057096 Chr.21: 43053408-43053408 on Build GRCh38
rs73372352 Chr.21: 43053497-43053497 on Build GRCh38
rs914944120 Chr.21: 43053106-43053106 on Build GRCh38
rs571374721 Chr.21: 43053471-43053471 on Build GRCh38
rs111969522 Chr.21: 43053314-43053314 on Build GRCh38
rs545040087 Chr.21: 43053264-43053264 on Build GRCh38
rs111386779 Chr.21: 43053340-43053340 on Build GRCh38
rs916041440 Chr.21: 43053537-43053537 on Build GRCh38
rs1004633896 Chr.21: 43053397-43053397 on Build GRCh38

View Full Product Details