Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38321936-38322365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2268613 c.216G>T p.L72F Chr.15: 38322249-38322249 on Build GRCh38
3500691 c.240C>T p.L80L Chr.15: 38322273-38322273 on Build GRCh38
4387951 c.276G>A p.W92* Chr.15: 38322309-38322309 on Build GRCh38
4054215 c.305C>T p.T102M Chr.15: 38322338-38322338 on Build GRCh38
5980706 c.287A>G p.D96G Chr.15: 38322320-38322320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025878665 Chr.15: 38322138-38322138 on Build GRCh38
rs369150309 Chr.15: 38322244-38322244 on Build GRCh38
rs727504170 Chr.15: 38322262-38322262 on Build GRCh38
rs556636985 Chr.15: 38322323-38322323 on Build GRCh38
rs751006467 Chr.15: 38322059-38322059 on Build GRCh38
rs763749955 Chr.15: 38322261-38322261 on Build GRCh38
rs200529523 Chr.15: 38322198-38322198 on Build GRCh38
rs534638060 Chr.15: 38322186-38322186 on Build GRCh38
rs567106024 Chr.15: 38322155-38322155 on Build GRCh38
rs773943895 Chr.15: 38322303-38322303 on Build GRCh38
rs140151387 Chr.15: 38322056-38322056 on Build GRCh38
rs553227041 Chr.15: 38322187-38322187 on Build GRCh38
rs928301935 Chr.15: 38322236-38322236 on Build GRCh38
rs768883825 Chr.15: 38322196-38322196 on Build GRCh38
rs183031739 Chr.15: 38322204-38322204 on Build GRCh38
rs746787097 Chr.15: 38322195-38322195 on Build GRCh38
rs898633486 Chr.15: 38322132-38322132 on Build GRCh38
rs957744615 Chr.15: 38322001-38322001 on Build GRCh38
rs780800297 Chr.15: 38322202-38322203 on Build GRCh38
rs1001618889 Chr.15: 38322028-38322028 on Build GRCh38
rs946042558 Chr.15: 38321962-38321962 on Build GRCh38
rs749950643 Chr.15: 38322313-38322313 on Build GRCh38
rs754706111 Chr.15: 38322338-38322338 on Build GRCh38
rs773490200 Chr.15: 38322207-38322207 on Build GRCh38
rs951699960 Chr.15: 38322150-38322150 on Build GRCh38
rs577606048 Chr.15: 38322188-38322188 on Build GRCh38
rs776741198 Chr.15: 38322202-38322202 on Build GRCh38
rs769963508 Chr.15: 38322206-38322206 on Build GRCh38
rs756975029 Chr.15: 38322305-38322305 on Build GRCh38
rs763539012 Chr.15: 38322210-38322210 on Build GRCh38
rs537122708 Chr.15: 38322122-38322122 on Build GRCh38
rs1034521094 Chr.15: 38322060-38322060 on Build GRCh38
rs372827979 Chr.15: 38322154-38322154 on Build GRCh38
rs767004300 Chr.15: 38322212-38322212 on Build GRCh38
rs569822005 Chr.15: 38322105-38322105 on Build GRCh38
rs372791883 Chr.15: 38322339-38322339 on Build GRCh38
rs995774365 Chr.15: 38322134-38322134 on Build GRCh38
rs751879511 Chr.15: 38322230-38322230 on Build GRCh38
rs778384985 Chr.15: 38322312-38322312 on Build GRCh38
rs1031787425 Chr.15: 38321966-38321966 on Build GRCh38
rs907313173 Chr.15: 38322009-38322009 on Build GRCh38
rs761473300 Chr.15: 38322314-38322314 on Build GRCh38
rs7182445 Chr.15: 38322324-38322324 on Build GRCh38
rs1046124723 Chr.15: 38321970-38321970 on Build GRCh38
rs398122371 Chr.15: 38322223-38322223 on Build GRCh38

View Full Product Details