Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28003343-28003814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
UniGene
Hs.507590
Gene Chromosome Location
Chr.13: 28003274-28100592 on Build GRCh38
Transcripts
NM_004119.2 NR_130706.1 XM_011535015.2 XM_011535017.2 XM_011535018.2 XM_017020486.1 XM_017020487.1 XM_017020488.1 XM_017020489.1

SNP Detail

SNP ID Chromosome Location
rs185137504 Chr.13: 28003507-28003507 on Build GRCh38
rs181954154 Chr.13: 28003484-28003484 on Build GRCh38
rs527893229 Chr.13: 28003515-28003515 on Build GRCh38
rs771151828 Chr.13: 28003612-28003612 on Build GRCh38
rs753098986 Chr.13: 28003618-28003618 on Build GRCh38
rs138434705 Chr.13: 28003502-28003502 on Build GRCh38
rs59251752 Chr.13: 28003536-28003536 on Build GRCh38
rs998482418 Chr.13: 28003691-28003691 on Build GRCh38
rs569666291 Chr.13: 28003609-28003609 on Build GRCh38
rs979322517 Chr.13: 28003659-28003659 on Build GRCh38
rs554326978 Chr.13: 28003762-28003762 on Build GRCh38
rs955346534 Chr.13: 28003617-28003617 on Build GRCh38
rs939990128 Chr.13: 28003637-28003637 on Build GRCh38
rs541180659 Chr.13: 28003375-28003375 on Build GRCh38
rs746975737 Chr.13: 28003733-28003737 on Build GRCh38
rs190904178 Chr.13: 28003388-28003388 on Build GRCh38
rs887402589 Chr.13: 28003781-28003781 on Build GRCh38
rs888547361 Chr.13: 28003621-28003621 on Build GRCh38
rs1048967868 Chr.13: 28003376-28003376 on Build GRCh38
rs917601175 Chr.13: 28003723-28003723 on Build GRCh38
rs755723136 Chr.13: 28003622-28003622 on Build GRCh38
rs968366659 Chr.13: 28003649-28003649 on Build GRCh38
rs953241389 Chr.13: 28003490-28003490 on Build GRCh38
rs896133612 Chr.13: 28003515-28003515 on Build GRCh38
rs773235846 Chr.13: 28003455-28003456 on Build GRCh38
rs144851128 Chr.13: 28003415-28003416 on Build GRCh38
rs1037058903 Chr.13: 28003640-28003640 on Build GRCh38
rs556238637 Chr.13: 28003573-28003573 on Build GRCh38
rs536932359 Chr.13: 28003560-28003560 on Build GRCh38
rs3829382 Chr.13: 28003551-28003551 on Build GRCh38
rs35613052 Chr.13: 28003417-28003417 on Build GRCh38
rs1029637760 Chr.13: 28003603-28003603 on Build GRCh38
rs913738420 Chr.13: 28003393-28003393 on Build GRCh38
rs1042106 Chr.13: 28003792-28003792 on Build GRCh38
rs1048452456 Chr.13: 28003576-28003576 on Build GRCh38
rs1047279357 Chr.13: 28003549-28003549 on Build GRCh38
rs1047848278 Chr.13: 28003742-28003742 on Build GRCh38
rs111552438 Chr.13: 28003677-28003677 on Build GRCh38
rs562069146 Chr.13: 28003554-28003554 on Build GRCh38
rs529915508 Chr.13: 28003396-28003396 on Build GRCh38
rs189844359 Chr.13: 28003702-28003702 on Build GRCh38
rs889920755 Chr.13: 28003714-28003714 on Build GRCh38
rs73436930 Chr.13: 28003445-28003445 on Build GRCh38
rs540895062 Chr.13: 28003732-28003732 on Build GRCh38
rs1004237198 Chr.13: 28003397-28003397 on Build GRCh38
rs1028531428 Chr.13: 28003470-28003470 on Build GRCh38
rs766372068 Chr.13: 28003541-28003541 on Build GRCh38
rs538582324 Chr.13: 28003631-28003631 on Build GRCh38
rs1046654813 Chr.13: 28003555-28003555 on Build GRCh38
rs985863339 Chr.13: 28003501-28003501 on Build GRCh38
rs1056169287 Chr.13: 28003514-28003514 on Build GRCh38
rs998743295 Chr.13: 28003459-28003459 on Build GRCh38
rs34517967 Chr.13: 28003475-28003476 on Build GRCh38

View Full Product Details