Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 65351289-65351646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA5
Entrez Gene ID
2044
Gene Name
EPH receptor A5
Gene Aliases
CEK7, EHK-1, EHK1, EK7, HEK7, TYRO4
UniGene
Hs.654492
Gene Chromosome Location
Chr.4: 65319563-65670838 on Build GRCh38
Transcripts
NM_001281765.2 NM_001281766.2 NM_001281767.2 NM_001318761.1 NM_004439.7 NM_182472.4 XM_005265653.3 XM_011531735.2 XM_017007878.1 XM_017007879.1 XM_017007880.1 XM_017007881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6930726 c.2321T>A p.V774E Chr.4: 65351576-65351576 on Build GRCh38
4536503 c.2305G>A p.D769N Chr.4: 65351592-65351592 on Build GRCh38
5698466 c.2336G>T p.G779V Chr.4: 65351561-65351561 on Build GRCh38
6976282 c.2450C>T p.S817F Chr.4: 65351447-65351447 on Build GRCh38
6100618 c.2357C>A p.A786E Chr.4: 65351540-65351540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202005571 Chr.4: 65351372-65351372 on Build GRCh38
rs747385140 Chr.4: 65351430-65351430 on Build GRCh38
rs758578574 Chr.4: 65351426-65351426 on Build GRCh38
rs771424766 Chr.4: 65351432-65351432 on Build GRCh38
rs748209436 Chr.4: 65351603-65351603 on Build GRCh38
rs777926897 Chr.4: 65351591-65351591 on Build GRCh38
rs774897926 Chr.4: 65351464-65351464 on Build GRCh38
rs757668101 Chr.4: 65351603-65351603 on Build GRCh38
rs530933021 Chr.4: 65351573-65351573 on Build GRCh38
rs755145140 Chr.4: 65351411-65351411 on Build GRCh38
rs767617774 Chr.4: 65351409-65351409 on Build GRCh38
rs374985302 Chr.4: 65351414-65351414 on Build GRCh38
rs932027161 Chr.4: 65351319-65351319 on Build GRCh38
rs1056327058 Chr.4: 65351606-65351606 on Build GRCh38
rs13435365 Chr.4: 65351360-65351360 on Build GRCh38
rs369412311 Chr.4: 65351508-65351508 on Build GRCh38
rs746338965 Chr.4: 65351613-65351613 on Build GRCh38
rs548263764 Chr.4: 65351427-65351427 on Build GRCh38
rs906814320 Chr.4: 65351414-65351436 on Build GRCh38
rs781559724 Chr.4: 65351612-65351612 on Build GRCh38
rs781694367 Chr.4: 65351446-65351446 on Build GRCh38
rs751891319 Chr.4: 65351593-65351593 on Build GRCh38
rs371973459 Chr.4: 65351469-65351469 on Build GRCh38
rs371238908 Chr.4: 65351395-65351395 on Build GRCh38
rs774180811 Chr.4: 65351521-65351521 on Build GRCh38
rs768209868 Chr.4: 65351516-65351516 on Build GRCh38
rs199801676 Chr.4: 65351549-65351549 on Build GRCh38
rs907768294 Chr.4: 65351512-65351512 on Build GRCh38
rs760710515 Chr.4: 65351559-65351559 on Build GRCh38
rs762539388 Chr.4: 65351384-65351384 on Build GRCh38
rs759116583 Chr.4: 65351375-65351375 on Build GRCh38
rs549914252 Chr.4: 65351586-65351586 on Build GRCh38
rs770210947 Chr.4: 65351619-65351619 on Build GRCh38
rs775265862 Chr.4: 65351382-65351382 on Build GRCh38
rs896125276 Chr.4: 65351620-65351620 on Build GRCh38
rs145959986 Chr.4: 65351398-65351398 on Build GRCh38
rs765320417 Chr.4: 65351578-65351578 on Build GRCh38
rs764842332 Chr.4: 65351377-65351377 on Build GRCh38
rs139853168 Chr.4: 65351410-65351410 on Build GRCh38
rs1050334812 Chr.4: 65351333-65351333 on Build GRCh38
rs747507614 Chr.4: 65351370-65351370 on Build GRCh38
rs763912590 Chr.4: 65351390-65351390 on Build GRCh38
rs758586357 Chr.4: 65351587-65351587 on Build GRCh38
rs761764875 Chr.4: 65351527-65351527 on Build GRCh38
rs745360988 Chr.4: 65351450-65351450 on Build GRCh38
rs568570258 Chr.4: 65351456-65351456 on Build GRCh38

View Full Product Details