Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105125188-105125711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BVES
Entrez Gene ID
11149
Gene Name
blood vessel epicardial substance
Gene Aliases
HBVES, LGMD2X, POP1, POPDC1
UniGene
Hs.221660
Gene Chromosome Location
Chr.6: 105096822-105137174 on Build GRCh38
Transcripts
NM_001199563.1 NM_007073.4 NM_147147.3 XM_011535398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305189 c.366G>T p.K122N Chr.6: 105125564-105125564 on Build GRCh38
3410517 c.389G>A p.R130Q Chr.6: 105125541-105125541 on Build GRCh38
450257 c.397G>A p.E133K Chr.6: 105125533-105125533 on Build GRCh38
4495849 c.461C>T p.T154I Chr.6: 105125469-105125469 on Build GRCh38
7192968 c.373A>G p.S125G Chr.6: 105125557-105125557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373393976 Chr.6: 105125351-105125351 on Build GRCh38
rs369142492 Chr.6: 105125460-105125460 on Build GRCh38
rs781412943 Chr.6: 105125372-105125373 on Build GRCh38
rs970755941 Chr.6: 105125272-105125272 on Build GRCh38
rs771907922 Chr.6: 105125427-105125427 on Build GRCh38
rs749739338 Chr.6: 105125299-105125299 on Build GRCh38
rs959109245 Chr.6: 105125337-105125337 on Build GRCh38
rs755391039 Chr.6: 105125371-105125371 on Build GRCh38
rs376927875 Chr.6: 105125373-105125373 on Build GRCh38
rs530742250 Chr.6: 105125415-105125415 on Build GRCh38
rs765386566 Chr.6: 105125459-105125459 on Build GRCh38
rs756497204 Chr.6: 105125607-105125607 on Build GRCh38
rs766483699 Chr.6: 105125577-105125577 on Build GRCh38
rs781667333 Chr.6: 105125495-105125495 on Build GRCh38
rs747600337 Chr.6: 105125645-105125645 on Build GRCh38
rs527773132 Chr.6: 105125265-105125265 on Build GRCh38
rs748912585 Chr.6: 105125408-105125408 on Build GRCh38
rs893975395 Chr.6: 105125660-105125660 on Build GRCh38
rs759858875 Chr.6: 105125586-105125586 on Build GRCh38
rs775286797 Chr.6: 105125447-105125447 on Build GRCh38
rs770535404 Chr.6: 105125409-105125409 on Build GRCh38
rs146676450 Chr.6: 105125487-105125487 on Build GRCh38
rs1012909808 Chr.6: 105125664-105125664 on Build GRCh38
rs9486039 Chr.6: 105125549-105125549 on Build GRCh38
rs777319338 Chr.6: 105125407-105125407 on Build GRCh38
rs984443041 Chr.6: 105125311-105125311 on Build GRCh38
rs750662888 Chr.6: 105125347-105125347 on Build GRCh38
rs764531451 Chr.6: 105125614-105125614 on Build GRCh38
rs761920317 Chr.6: 105125340-105125340 on Build GRCh38
rs748691293 Chr.6: 105125389-105125389 on Build GRCh38
rs374576734 Chr.6: 105125552-105125552 on Build GRCh38
rs746912580 Chr.6: 105125550-105125550 on Build GRCh38
rs763068609 Chr.6: 105125568-105125568 on Build GRCh38
rs754349687 Chr.6: 105125621-105125621 on Build GRCh38
rs2275289 Chr.6: 105125545-105125545 on Build GRCh38
rs756719192 Chr.6: 105125624-105125624 on Build GRCh38
rs935105532 Chr.6: 105125605-105125605 on Build GRCh38
rs370640207 Chr.6: 105125479-105125479 on Build GRCh38
rs1017387832 Chr.6: 105125231-105125231 on Build GRCh38
rs778545273 Chr.6: 105125627-105125627 on Build GRCh38
rs140271160 Chr.6: 105125502-105125502 on Build GRCh38
rs751830675 Chr.6: 105125369-105125369 on Build GRCh38
rs1006920414 Chr.6: 105125574-105125575 on Build GRCh38
rs756658925 Chr.6: 105125504-105125504 on Build GRCh38
rs997377011 Chr.6: 105125675-105125675 on Build GRCh38
rs757686585 Chr.6: 105125349-105125349 on Build GRCh38
rs150910551 Chr.6: 105125561-105125561 on Build GRCh38
rs148128379 Chr.6: 105125544-105125544 on Build GRCh38
rs753095706 Chr.6: 105125603-105125603 on Build GRCh38
rs764520256 Chr.6: 105125521-105125521 on Build GRCh38
rs748751226 Chr.6: 105125524-105125524 on Build GRCh38
rs141154991 Chr.6: 105125416-105125416 on Build GRCh38
rs780534799 Chr.6: 105125364-105125364 on Build GRCh38
rs889853104 Chr.6: 105125218-105125218 on Build GRCh38
rs1012506603 Chr.6: 105125244-105125244 on Build GRCh38
rs760754660 Chr.6: 105125453-105125453 on Build GRCh38
rs1023599508 Chr.6: 105125259-105125259 on Build GRCh38
rs1043012012 Chr.6: 105125436-105125436 on Build GRCh38
rs565045818 Chr.6: 105125324-105125324 on Build GRCh38
rs754945530 Chr.6: 105125561-105125561 on Build GRCh38
rs570929978 Chr.6: 105125645-105125646 on Build GRCh38
rs17524448 Chr.6: 105125229-105125229 on Build GRCh38
rs143158984 Chr.6: 105125471-105125471 on Build GRCh38
rs566763596 Chr.6: 105125589-105125589 on Build GRCh38
rs539276661 Chr.6: 105125687-105125687 on Build GRCh38
rs192533910 Chr.6: 105125211-105125211 on Build GRCh38
rs769286149 Chr.6: 105125406-105125406 on Build GRCh38
rs746236429 Chr.6: 105125403-105125404 on Build GRCh38
rs982183064 Chr.6: 105125273-105125273 on Build GRCh38
rs768248477 Chr.6: 105125411-105125412 on Build GRCh38
rs756776885 Chr.6: 105125543-105125543 on Build GRCh38
rs143510978 Chr.6: 105125523-105125523 on Build GRCh38
rs779809004 Chr.6: 105125516-105125516 on Build GRCh38

View Full Product Details