Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35713909-35714508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf216
Entrez Gene ID
127703
Gene Name
chromosome 1 open reading frame 216
Gene Aliases
-
UniGene
Hs.112023
Gene Chromosome Location
Chr.1: 35713876-35719189 on Build GRCh38
Transcripts
NM_152374.1

SNP Detail

SNP ID Chromosome Location
rs964192003 Chr.1: 35714454-35714454 on Build GRCh38
rs1035305069 Chr.1: 35713929-35713929 on Build GRCh38
rs968423907 Chr.1: 35714047-35714047 on Build GRCh38
rs1056978534 Chr.1: 35714157-35714157 on Build GRCh38
rs908282368 Chr.1: 35714214-35714214 on Build GRCh38
rs895699400 Chr.1: 35714197-35714205 on Build GRCh38
rs1032347491 Chr.1: 35714234-35714234 on Build GRCh38
rs1019796902 Chr.1: 35714294-35714294 on Build GRCh38
rs1031081050 Chr.1: 35714354-35714354 on Build GRCh38
rs1031028518 Chr.1: 35714094-35714094 on Build GRCh38
rs945907328 Chr.1: 35714451-35714451 on Build GRCh38
rs1020189884 Chr.1: 35714209-35714209 on Build GRCh38
rs769171465 Chr.1: 35714426-35714426 on Build GRCh38
rs975702994 Chr.1: 35714168-35714168 on Build GRCh38
rs963997253 Chr.1: 35714156-35714156 on Build GRCh38
rs954161886 Chr.1: 35714249-35714249 on Build GRCh38
rs1053755 Chr.1: 35713992-35713992 on Build GRCh38
rs571648568 Chr.1: 35714237-35714237 on Build GRCh38
rs574951191 Chr.1: 35714062-35714062 on Build GRCh38
rs1001263191 Chr.1: 35714344-35714344 on Build GRCh38
rs142991468 Chr.1: 35714213-35714213 on Build GRCh38
rs956785934 Chr.1: 35714370-35714370 on Build GRCh38
rs895638988 Chr.1: 35714449-35714449 on Build GRCh38
rs940876373 Chr.1: 35714248-35714248 on Build GRCh38
rs1012511911 Chr.1: 35714275-35714275 on Build GRCh38
rs965952781 Chr.1: 35714296-35714296 on Build GRCh38
rs1008227826 Chr.1: 35714392-35714392 on Build GRCh38
rs1045702394 Chr.1: 35714311-35714311 on Build GRCh38
rs748633707 Chr.1: 35714258-35714258 on Build GRCh38
rs944313661 Chr.1: 35713977-35713977 on Build GRCh38
rs772547416 Chr.1: 35714212-35714212 on Build GRCh38
rs930954136 Chr.1: 35714203-35714203 on Build GRCh38
rs927001857 Chr.1: 35714357-35714357 on Build GRCh38
rs1012872401 Chr.1: 35713975-35713975 on Build GRCh38
rs555132517 Chr.1: 35714021-35714021 on Build GRCh38
rs896620816 Chr.1: 35714267-35714267 on Build GRCh38
rs905614439 Chr.1: 35714095-35714095 on Build GRCh38
rs530424457 Chr.1: 35713983-35713983 on Build GRCh38
rs912518768 Chr.1: 35714145-35714145 on Build GRCh38
rs975378721 Chr.1: 35714481-35714481 on Build GRCh38
rs374780803 Chr.1: 35714039-35714039 on Build GRCh38
rs903878908 Chr.1: 35714312-35714312 on Build GRCh38
rs1023643105 Chr.1: 35713982-35713982 on Build GRCh38
rs985647568 Chr.1: 35713931-35713931 on Build GRCh38
rs1057308735 Chr.1: 35714430-35714430 on Build GRCh38
rs139355518 Chr.1: 35714107-35714107 on Build GRCh38
rs1019415307 Chr.1: 35714395-35714395 on Build GRCh38
rs918238146 Chr.1: 35714053-35714053 on Build GRCh38
rs747454281 Chr.1: 35714298-35714298 on Build GRCh38
rs769391823 Chr.1: 35713986-35713986 on Build GRCh38
rs930962892 Chr.1: 35714321-35714321 on Build GRCh38
rs919620410 Chr.1: 35714172-35714172 on Build GRCh38
rs78434711 Chr.1: 35713936-35713936 on Build GRCh38
rs532380274 Chr.1: 35714420-35714420 on Build GRCh38
rs986588302 Chr.1: 35714144-35714144 on Build GRCh38

View Full Product Details