Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38350739-38351333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
80574 c.738C>G p.V246V Chr.15: 38351067-38351067 on Build GRCh38
78569 c.821A>C p.D274A Chr.15: 38351150-38351150 on Build GRCh38
1227426 c.863A>C p.K288T Chr.15: 38351192-38351192 on Build GRCh38
1227425 c.686T>C p.V229A Chr.15: 38351015-38351015 on Build GRCh38
7429387 c.869C>T p.S290L Chr.15: 38351198-38351198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946203764 Chr.15: 38351147-38351147 on Build GRCh38
rs535687467 Chr.15: 38351208-38351208 on Build GRCh38
rs886051105 Chr.15: 38351210-38351210 on Build GRCh38
rs886041428 Chr.15: 38351136-38351136 on Build GRCh38
rs569947862 Chr.15: 38350845-38350845 on Build GRCh38
rs750890621 Chr.15: 38351306-38351306 on Build GRCh38
rs140644874 Chr.15: 38351268-38351268 on Build GRCh38
rs543503614 Chr.15: 38350824-38350824 on Build GRCh38
rs761001792 Chr.15: 38351221-38351221 on Build GRCh38
rs551134910 Chr.15: 38350960-38350960 on Build GRCh38
rs375700452 Chr.15: 38351260-38351260 on Build GRCh38
rs115440602 Chr.15: 38351273-38351273 on Build GRCh38
rs913339445 Chr.15: 38351146-38351146 on Build GRCh38
rs1043239392 Chr.15: 38351158-38351158 on Build GRCh38
rs369834749 Chr.15: 38350999-38350999 on Build GRCh38
rs1025445663 Chr.15: 38350948-38350948 on Build GRCh38
rs529060512 Chr.15: 38350855-38350855 on Build GRCh38
rs755557783 Chr.15: 38351170-38351170 on Build GRCh38
rs12914722 Chr.15: 38350830-38350830 on Build GRCh38
rs992984515 Chr.15: 38350839-38350839 on Build GRCh38
rs766814966 Chr.15: 38351267-38351267 on Build GRCh38
rs547532432 Chr.15: 38350896-38350896 on Build GRCh38
rs778423017 Chr.15: 38351195-38351203 on Build GRCh38
rs755109864 Chr.15: 38350949-38350949 on Build GRCh38
rs759197561 Chr.15: 38351183-38351183 on Build GRCh38
rs987974431 Chr.15: 38351038-38351038 on Build GRCh38
rs1016860501 Chr.15: 38350990-38350990 on Build GRCh38
rs769730984 Chr.15: 38350975-38350975 on Build GRCh38
rs765684707 Chr.15: 38351094-38351094 on Build GRCh38
rs970314232 Chr.15: 38350908-38350908 on Build GRCh38
rs371620589 Chr.15: 38351007-38351007 on Build GRCh38
rs864622410 Chr.15: 38351125-38351126 on Build GRCh38
rs121434317 Chr.15: 38351113-38351113 on Build GRCh38
rs576178445 Chr.15: 38350822-38350822 on Build GRCh38
rs752201824 Chr.15: 38351164-38351164 on Build GRCh38
rs760690837 Chr.15: 38351006-38351006 on Build GRCh38
rs777379558 Chr.15: 38351193-38351193 on Build GRCh38
rs780737673 Chr.15: 38351152-38351152 on Build GRCh38
rs1036559955 Chr.15: 38350961-38350961 on Build GRCh38
rs1028970466 Chr.15: 38351307-38351307 on Build GRCh38
rs200974459 Chr.15: 38351216-38351216 on Build GRCh38
rs770678427 Chr.15: 38350991-38350991 on Build GRCh38
rs76128643 Chr.15: 38351013-38351013 on Build GRCh38
rs886051104 Chr.15: 38351036-38351036 on Build GRCh38
rs1022058333 Chr.15: 38350807-38350807 on Build GRCh38
rs1057518683 Chr.15: 38351302-38351302 on Build GRCh38
rs775556490 Chr.15: 38351217-38351217 on Build GRCh38
rs899712805 Chr.15: 38351280-38351280 on Build GRCh38
rs773221725 Chr.15: 38350966-38350966 on Build GRCh38
rs1043269912 Chr.15: 38350787-38350787 on Build GRCh38
rs749268930 Chr.15: 38350985-38350985 on Build GRCh38
rs963992906 Chr.15: 38351005-38351005 on Build GRCh38
rs750832911 Chr.15: 38351099-38351099 on Build GRCh38
rs765841849 Chr.15: 38351251-38351251 on Build GRCh38
rs904821832 Chr.15: 38350791-38350791 on Build GRCh38
rs749692794 Chr.15: 38351292-38351297 on Build GRCh38
rs762478369 Chr.15: 38351041-38351041 on Build GRCh38
rs369711772 Chr.15: 38351292-38351292 on Build GRCh38
rs752191960 Chr.15: 38351257-38351257 on Build GRCh38
rs554327738 Chr.15: 38351215-38351215 on Build GRCh38
rs566051898 Chr.15: 38350998-38350999 on Build GRCh38
rs1014436638 Chr.15: 38350931-38350931 on Build GRCh38
rs764868020 Chr.15: 38351249-38351249 on Build GRCh38
rs111363743 Chr.15: 38351150-38351150 on Build GRCh38
rs201933663 Chr.15: 38351204-38351204 on Build GRCh38
rs747615028 Chr.15: 38350974-38350974 on Build GRCh38
rs116667267 Chr.15: 38351124-38351124 on Build GRCh38
rs960230909 Chr.15: 38350832-38350832 on Build GRCh38
rs540124243 Chr.15: 38351228-38351228 on Build GRCh38
rs372665535 Chr.15: 38351195-38351195 on Build GRCh38
rs766152186 Chr.15: 38351060-38351060 on Build GRCh38
rs776909752 Chr.15: 38351235-38351235 on Build GRCh38
rs904563789 Chr.15: 38351165-38351165 on Build GRCh38
rs762047423 Chr.15: 38350897-38350897 on Build GRCh38
rs143030909 Chr.15: 38350943-38350943 on Build GRCh38
rs1014323023 Chr.15: 38351243-38351243 on Build GRCh38
rs138553244 Chr.15: 38351031-38351031 on Build GRCh38
rs1005383827 Chr.15: 38351270-38351270 on Build GRCh38
rs12913526 Chr.15: 38350808-38350808 on Build GRCh38
rs559419292 Chr.15: 38350911-38350911 on Build GRCh38
rs764287445 Chr.15: 38351025-38351025 on Build GRCh38
rs774196007 Chr.15: 38350995-38350995 on Build GRCh38
rs201837340 Chr.15: 38351014-38351014 on Build GRCh38
rs1007409378 Chr.15: 38350781-38350781 on Build GRCh38
rs916188514 Chr.15: 38350847-38350847 on Build GRCh38
rs1042940854 Chr.15: 38351082-38351082 on Build GRCh38
rs777593473 Chr.15: 38350981-38350981 on Build GRCh38
rs114636635 Chr.15: 38351255-38351255 on Build GRCh38
rs753357510 Chr.15: 38351283-38351283 on Build GRCh38
rs866765311 Chr.15: 38350815-38350815 on Build GRCh38
rs946292526 Chr.15: 38350771-38350771 on Build GRCh38
rs12913505 Chr.15: 38350770-38350770 on Build GRCh38
rs376226762 Chr.15: 38351016-38351016 on Build GRCh38
rs908876401 Chr.15: 38350914-38350914 on Build GRCh38

View Full Product Details