Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31327962-31328489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1
Entrez Gene ID
1828
Gene Name
desmoglein 1
Gene Aliases
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1
UniGene
Hs.2633
Gene Chromosome Location
Chr.18: 31318089-31359246 on Build GRCh38
Transcripts
NM_001942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303660 c.266G>C p.G89A Chr.18: 31328238-31328238 on Build GRCh38
6202141 c.269T>A p.V90E Chr.18: 31328241-31328241 on Build GRCh38
376387 c.331A>G p.I111V Chr.18: 31328303-31328303 on Build GRCh38
987523 c.352G>T p.E118* Chr.18: 31328324-31328324 on Build GRCh38
987522 c.233C>A p.A78D Chr.18: 31328205-31328205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762635618 Chr.18: 31328268-31328268 on Build GRCh38
rs752467784 Chr.18: 31328265-31328265 on Build GRCh38
rs949820079 Chr.18: 31328005-31328005 on Build GRCh38
rs940139013 Chr.18: 31328129-31328129 on Build GRCh38
rs980280280 Chr.18: 31328342-31328342 on Build GRCh38
rs543163097 Chr.18: 31328259-31328259 on Build GRCh38
rs532831057 Chr.18: 31328429-31328429 on Build GRCh38
rs1033378368 Chr.18: 31328309-31328309 on Build GRCh38
rs150168711 Chr.18: 31328318-31328318 on Build GRCh38
rs138672131 Chr.18: 31328341-31328341 on Build GRCh38
rs1006958346 Chr.18: 31328447-31328447 on Build GRCh38
rs941602848 Chr.18: 31328061-31328061 on Build GRCh38
rs780552898 Chr.18: 31328206-31328206 on Build GRCh38
rs751040177 Chr.18: 31328184-31328184 on Build GRCh38
rs902446057 Chr.18: 31328273-31328273 on Build GRCh38
rs941559029 Chr.18: 31328371-31328371 on Build GRCh38
rs755271644 Chr.18: 31328321-31328321 on Build GRCh38
rs376000128 Chr.18: 31328335-31328335 on Build GRCh38
rs761748106 Chr.18: 31328228-31328228 on Build GRCh38
rs750949228 Chr.18: 31328290-31328290 on Build GRCh38
rs1051065211 Chr.18: 31328440-31328440 on Build GRCh38
rs573281660 Chr.18: 31328289-31328289 on Build GRCh38
rs970413489 Chr.18: 31328424-31328424 on Build GRCh38
rs773940417 Chr.18: 31328379-31328379 on Build GRCh38
rs554216628 Chr.18: 31328000-31328000 on Build GRCh38
rs372730921 Chr.18: 31328139-31328139 on Build GRCh38
rs535958672 Chr.18: 31327999-31327999 on Build GRCh38
rs367991715 Chr.18: 31328137-31328137 on Build GRCh38
rs556306911 Chr.18: 31328177-31328177 on Build GRCh38
rs759197744 Chr.18: 31328146-31328146 on Build GRCh38
rs781492654 Chr.18: 31328191-31328191 on Build GRCh38
rs756663231 Chr.18: 31328310-31328310 on Build GRCh38
rs959115979 Chr.18: 31328016-31328016 on Build GRCh38
rs891161855 Chr.18: 31328444-31328444 on Build GRCh38
rs2303465 Chr.18: 31328127-31328127 on Build GRCh38
rs752417138 Chr.18: 31328173-31328173 on Build GRCh38
rs149439494 Chr.18: 31328193-31328193 on Build GRCh38
rs932765440 Chr.18: 31328161-31328161 on Build GRCh38
rs774601712 Chr.18: 31328222-31328222 on Build GRCh38
rs1038542694 Chr.18: 31328376-31328376 on Build GRCh38
rs1011983162 Chr.18: 31328365-31328365 on Build GRCh38
rs745843187 Chr.18: 31328343-31328343 on Build GRCh38
rs748602947 Chr.18: 31328099-31328099 on Build GRCh38
rs561317580 Chr.18: 31328260-31328260 on Build GRCh38
rs201339427 Chr.18: 31328181-31328181 on Build GRCh38
rs780602124 Chr.18: 31328311-31328311 on Build GRCh38
rs199753914 Chr.18: 31328387-31328387 on Build GRCh38
rs906053807 Chr.18: 31328039-31328039 on Build GRCh38
rs74368609 Chr.18: 31328215-31328215 on Build GRCh38
rs917602553 Chr.18: 31328053-31328053 on Build GRCh38
rs563120284 Chr.18: 31328437-31328437 on Build GRCh38
rs76275716 Chr.18: 31328068-31328068 on Build GRCh38
rs763738346 Chr.18: 31328370-31328370 on Build GRCh38
rs779222304 Chr.18: 31328323-31328323 on Build GRCh38
rs763697283 Chr.18: 31328269-31328269 on Build GRCh38
rs138930324 Chr.18: 31328190-31328190 on Build GRCh38
rs768964686 Chr.18: 31328220-31328220 on Build GRCh38
rs550961079 Chr.18: 31328415-31328415 on Build GRCh38
rs775296123 Chr.18: 31328360-31328360 on Build GRCh38
rs990865508 Chr.18: 31328051-31328051 on Build GRCh38
rs988398208 Chr.18: 31328322-31328322 on Build GRCh38
rs766184127 Chr.18: 31328140-31328140 on Build GRCh38
rs748267927 Chr.18: 31328330-31328330 on Build GRCh38
rs756653522 Chr.18: 31328288-31328288 on Build GRCh38
rs994133731 Chr.18: 31328124-31328124 on Build GRCh38
rs377621121 Chr.18: 31328075-31328075 on Build GRCh38
rs762689151 Chr.18: 31328364-31328364 on Build GRCh38
rs770916767 Chr.18: 31328229-31328229 on Build GRCh38
rs141709185 Chr.18: 31328130-31328130 on Build GRCh38
rs753691947 Chr.18: 31328141-31328141 on Build GRCh38
rs770122916 Chr.18: 31328204-31328204 on Build GRCh38
rs751104288 Chr.18: 31328272-31328272 on Build GRCh38
rs776369313 Chr.18: 31328236-31328236 on Build GRCh38
rs1044540712 Chr.18: 31328023-31328023 on Build GRCh38
rs769689615 Chr.18: 31328344-31328344 on Build GRCh38
rs764877346 Chr.18: 31328152-31328152 on Build GRCh38
rs373204718 Chr.18: 31328359-31328359 on Build GRCh38
rs1008682996 Chr.18: 31328211-31328211 on Build GRCh38

View Full Product Details