Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11758501-11759096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA4
Entrez Gene ID
2626
Gene Name
GATA binding protein 4
Gene Aliases
ASD2, TACHD, TOF, VSD1
UniGene
Hs.243987
Gene Chromosome Location
Chr.8: 11676919-11760002 on Build GRCh38
Transcripts
NM_001308093.1 NM_001308094.1 NM_002052.4 XM_005272385.4 XM_005272386.1 XM_006716248.1 XM_011543817.2 XM_011543818.2 XM_017013312.1

SNP Detail

SNP ID Chromosome Location
rs983919063 Chr.8: 11758566-11758566 on Build GRCh38
rs1053543079 Chr.8: 11758841-11758841 on Build GRCh38
rs1023296405 Chr.8: 11758693-11758693 on Build GRCh38
rs928795540 Chr.8: 11758528-11758528 on Build GRCh38
rs867858 Chr.8: 11758829-11758829 on Build GRCh38
rs386722304 Chr.8: 11759061-11759062 on Build GRCh38
rs956736692 Chr.8: 11759050-11759050 on Build GRCh38
rs552028139 Chr.8: 11758753-11758753 on Build GRCh38
rs538035276 Chr.8: 11758787-11758787 on Build GRCh38
rs539917769 Chr.8: 11758913-11758913 on Build GRCh38
rs980091937 Chr.8: 11758702-11758702 on Build GRCh38
rs551166697 Chr.8: 11758560-11758560 on Build GRCh38
rs549563637 Chr.8: 11758924-11758924 on Build GRCh38
rs181138914 Chr.8: 11758956-11758956 on Build GRCh38
rs983566366 Chr.8: 11758756-11758756 on Build GRCh38
rs1013671588 Chr.8: 11758814-11758814 on Build GRCh38
rs879397137 Chr.8: 11758639-11758639 on Build GRCh38
rs1012286366 Chr.8: 11758674-11758674 on Build GRCh38
rs570426044 Chr.8: 11758786-11758786 on Build GRCh38
rs770477079 Chr.8: 11758708-11758708 on Build GRCh38
rs902327569 Chr.8: 11758650-11758650 on Build GRCh38
rs1027951123 Chr.8: 11758774-11758774 on Build GRCh38
rs367895649 Chr.8: 11758843-11758843 on Build GRCh38
rs933246114 Chr.8: 11758828-11758828 on Build GRCh38
rs896319694 Chr.8: 11758988-11758988 on Build GRCh38
rs763243694 Chr.8: 11758957-11758957 on Build GRCh38
rs376521625 Chr.8: 11758842-11758842 on Build GRCh38
rs1052569594 Chr.8: 11758710-11758710 on Build GRCh38
rs1051624585 Chr.8: 11758632-11758632 on Build GRCh38
rs371473910 Chr.8: 11758622-11758622 on Build GRCh38
rs893479515 Chr.8: 11758667-11758667 on Build GRCh38
rs951931198 Chr.8: 11758574-11758574 on Build GRCh38
rs185327204 Chr.8: 11759061-11759061 on Build GRCh38
rs947820454 Chr.8: 11758846-11758846 on Build GRCh38
rs988379865 Chr.8: 11758864-11758864 on Build GRCh38
rs950858179 Chr.8: 11758735-11758735 on Build GRCh38
rs923780493 Chr.8: 11758833-11758833 on Build GRCh38
rs983292508 Chr.8: 11758584-11758584 on Build GRCh38
rs887714639 Chr.8: 11758783-11758783 on Build GRCh38
rs1038955437 Chr.8: 11758585-11758585 on Build GRCh38
rs963493166 Chr.8: 11758765-11758765 on Build GRCh38
rs530610803 Chr.8: 11758706-11758706 on Build GRCh38
rs956931765 Chr.8: 11758860-11758860 on Build GRCh38
rs779125034 Chr.8: 11758637-11758637 on Build GRCh38
rs762862929 Chr.8: 11758905-11758905 on Build GRCh38
rs941995168 Chr.8: 11758582-11758582 on Build GRCh38
rs1062221 Chr.8: 11759020-11759020 on Build GRCh38
rs804291 Chr.8: 11759062-11759062 on Build GRCh38
rs992268352 Chr.8: 11758616-11758616 on Build GRCh38
rs1053918468 Chr.8: 11758656-11758656 on Build GRCh38
rs931612238 Chr.8: 11758891-11758891 on Build GRCh38
rs759536597 Chr.8: 11758764-11758764 on Build GRCh38
rs1001379532 Chr.8: 11759034-11759034 on Build GRCh38
rs975185092 Chr.8: 11758784-11758784 on Build GRCh38
rs916557946 Chr.8: 11758651-11758651 on Build GRCh38
rs3729857 Chr.8: 11758613-11758613 on Build GRCh38
rs575549926 Chr.8: 11758731-11758731 on Build GRCh38
rs1036315797 Chr.8: 11758939-11758939 on Build GRCh38
rs12825 Chr.8: 11759038-11759038 on Build GRCh38
rs935471061 Chr.8: 11758709-11758709 on Build GRCh38
rs1062219 Chr.8: 11758901-11758901 on Build GRCh38
rs746560516 Chr.8: 11758692-11758692 on Build GRCh38
rs935111814 Chr.8: 11758653-11758653 on Build GRCh38
rs775514138 Chr.8: 11758904-11758904 on Build GRCh38
rs921886321 Chr.8: 11758586-11758586 on Build GRCh38
rs188654804 Chr.8: 11758873-11758873 on Build GRCh38
rs1003062573 Chr.8: 11758716-11758716 on Build GRCh38
rs969515867 Chr.8: 11759013-11759013 on Build GRCh38
rs909228246 Chr.8: 11758575-11758575 on Build GRCh38
rs912721646 Chr.8: 11758866-11758866 on Build GRCh38
rs1014989090 Chr.8: 11758587-11758587 on Build GRCh38
rs7459940 Chr.8: 11758655-11758655 on Build GRCh38
rs988348653 Chr.8: 11759055-11759055 on Build GRCh38
rs7008652 Chr.8: 11758889-11758889 on Build GRCh38
rs904018 Chr.8: 11759007-11759007 on Build GRCh38
rs772690686 Chr.8: 11758550-11758550 on Build GRCh38
rs778346114 Chr.8: 11758524-11758524 on Build GRCh38
rs370567650 Chr.8: 11758525-11758525 on Build GRCh38
rs78334561 Chr.8: 11758840-11758840 on Build GRCh38
rs884662 Chr.8: 11758992-11758992 on Build GRCh38

View Full Product Details