Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42507856-42508427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM1
Entrez Gene ID
634
Gene Name
carcinoembryonic antigen related cell adhesion molecule 1
Gene Aliases
BGP, BGP1, BGPI
UniGene
Hs.512682
Gene Chromosome Location
Chr.19: 42507306-42528509 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 NR_073179.1 NR_073180.1

Gene Symbol
LIPE-AS1
Entrez Gene ID
100996307
Gene Name
LIPE antisense RNA 1
Gene Aliases
-
UniGene
Hs.466840
Gene Chromosome Location
Chr.19: 42397148-42652355 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 NR_073179.1 NR_073180.1

SNP Detail

SNP ID Chromosome Location
rs952684723 Chr.19: 42508326-42508326 on Build GRCh38
rs759270789 Chr.19: 42508078-42508078 on Build GRCh38
rs952540714 Chr.19: 42507920-42507920 on Build GRCh38
rs1026847300 Chr.19: 42508294-42508294 on Build GRCh38
rs943242012 Chr.19: 42508026-42508026 on Build GRCh38
rs531538714 Chr.19: 42508244-42508244 on Build GRCh38
rs934564903 Chr.19: 42508113-42508113 on Build GRCh38
rs1041910206 Chr.19: 42508127-42508127 on Build GRCh38
rs1044503064 Chr.19: 42508333-42508333 on Build GRCh38
rs769229241 Chr.19: 42507944-42507944 on Build GRCh38
rs1019202264 Chr.19: 42508230-42508230 on Build GRCh38
rs918918395 Chr.19: 42507892-42507892 on Build GRCh38
rs184689347 Chr.19: 42508184-42508184 on Build GRCh38
rs910263466 Chr.19: 42508016-42508016 on Build GRCh38
rs982997867 Chr.19: 42508361-42508361 on Build GRCh38
rs982665618 Chr.19: 42507921-42507921 on Build GRCh38
rs907932236 Chr.19: 42507938-42507938 on Build GRCh38
rs1007988157 Chr.19: 42508190-42508190 on Build GRCh38
rs553598371 Chr.19: 42508103-42508103 on Build GRCh38
rs1005747096 Chr.19: 42508376-42508376 on Build GRCh38
rs548807046 Chr.19: 42508140-42508140 on Build GRCh38
rs559869918 Chr.19: 42507918-42507918 on Build GRCh38
rs551715375 Chr.19: 42508253-42508253 on Build GRCh38
rs1046116395 Chr.19: 42507950-42507950 on Build GRCh38
rs987187384 Chr.19: 42507952-42507952 on Build GRCh38
rs904476367 Chr.19: 42508094-42508094 on Build GRCh38
rs996503458 Chr.19: 42508285-42508285 on Build GRCh38
rs937535247 Chr.19: 42508033-42508033 on Build GRCh38
rs893425281 Chr.19: 42508101-42508101 on Build GRCh38
rs1056052765 Chr.19: 42508097-42508097 on Build GRCh38
rs529052748 Chr.19: 42508048-42508048 on Build GRCh38
rs919883143 Chr.19: 42507904-42507904 on Build GRCh38
rs571558604 Chr.19: 42508379-42508379 on Build GRCh38
rs1043270689 Chr.19: 42508078-42508078 on Build GRCh38
rs928626907 Chr.19: 42507954-42507954 on Build GRCh38

View Full Product Details