Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34709232-34709750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC6
Entrez Gene ID
54102
Gene Name
chloride intracellular channel 6
Gene Aliases
CLIC1L
UniGene
Hs.473695
Gene Chromosome Location
Chr.21: 34668953-34718227 on Build GRCh38
Transcripts
NM_001317009.1 NM_053277.2 XM_017028406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841853 c.1800G>A p.L600L Chr.21: 34709385-34709385 on Build GRCh38
4295976 c.1942C>T p.H648Y Chr.21: 34709527-34709527 on Build GRCh38
5392863 c.1884G>A p.R628R Chr.21: 34709469-34709469 on Build GRCh38
3550502 c.1830G>A p.R610R Chr.21: 34709469-34709469 on Build GRCh38
4672803 c.1773C>T p.S591S Chr.21: 34709412-34709412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968103661 Chr.21: 34709577-34709577 on Build GRCh38
rs771094545 Chr.21: 34709579-34709579 on Build GRCh38
rs868050110 Chr.21: 34709484-34709484 on Build GRCh38
rs752451900 Chr.21: 34709457-34709457 on Build GRCh38
rs948437953 Chr.21: 34709719-34709719 on Build GRCh38
rs201204252 Chr.21: 34709578-34709578 on Build GRCh38
rs779768925 Chr.21: 34709347-34709347 on Build GRCh38
rs773294906 Chr.21: 34709502-34709502 on Build GRCh38
rs776019390 Chr.21: 34709646-34709646 on Build GRCh38
rs956755043 Chr.21: 34709267-34709267 on Build GRCh38
rs766141950 Chr.21: 34709442-34709442 on Build GRCh38
rs772477580 Chr.21: 34709416-34709416 on Build GRCh38
rs770373227 Chr.21: 34709559-34709559 on Build GRCh38
rs771067357 Chr.21: 34709328-34709328 on Build GRCh38
rs780800620 Chr.21: 34709387-34709387 on Build GRCh38
rs773946128 Chr.21: 34709700-34709700 on Build GRCh38
rs762898725 Chr.21: 34709511-34709511 on Build GRCh38
rs747240450 Chr.21: 34709418-34709418 on Build GRCh38
rs968154458 Chr.21: 34709327-34709327 on Build GRCh38
rs142329053 Chr.21: 34709361-34709361 on Build GRCh38
rs776650681 Chr.21: 34709421-34709421 on Build GRCh38
rs758895188 Chr.21: 34709565-34709565 on Build GRCh38
rs771624637 Chr.21: 34709718-34709718 on Build GRCh38
rs761849510 Chr.21: 34709422-34709422 on Build GRCh38
rs774144719 Chr.21: 34709586-34709586 on Build GRCh38
rs562026572 Chr.21: 34709604-34709604 on Build GRCh38
rs766259690 Chr.21: 34709538-34709538 on Build GRCh38
rs1002826953 Chr.21: 34709254-34709254 on Build GRCh38
rs3819041 Chr.21: 34709328-34709328 on Build GRCh38
rs775770648 Chr.21: 34709308-34709308 on Build GRCh38
rs141470374 Chr.21: 34709495-34709495 on Build GRCh38
rs112154158 Chr.21: 34709294-34709294 on Build GRCh38
rs1035419002 Chr.21: 34709256-34709256 on Build GRCh38
rs764520669 Chr.21: 34709457-34709458 on Build GRCh38
rs746424107 Chr.21: 34709523-34709523 on Build GRCh38
rs867764212 Chr.21: 34709462-34709462 on Build GRCh38
rs751559847 Chr.21: 34709539-34709539 on Build GRCh38
rs146402009 Chr.21: 34709448-34709448 on Build GRCh38
rs765189005 Chr.21: 34709323-34709323 on Build GRCh38
rs780964138 Chr.21: 34709456-34709456 on Build GRCh38
rs373654576 Chr.21: 34709575-34709575 on Build GRCh38
rs575486849 Chr.21: 34709445-34709445 on Build GRCh38
rs376492520 Chr.21: 34709560-34709560 on Build GRCh38
rs764164937 Chr.21: 34709313-34709313 on Build GRCh38
rs774979904 Chr.21: 34709403-34709403 on Build GRCh38
rs557555993 Chr.21: 34709392-34709392 on Build GRCh38
rs61748623 Chr.21: 34709487-34709487 on Build GRCh38
rs746316307 Chr.21: 34709488-34709488 on Build GRCh38
rs751931428 Chr.21: 34709584-34709588 on Build GRCh38
rs919624782 Chr.21: 34709716-34709716 on Build GRCh38
rs753923490 Chr.21: 34709319-34709319 on Build GRCh38
rs764240977 Chr.21: 34709419-34709419 on Build GRCh38
rs768364697 Chr.21: 34709674-34709674 on Build GRCh38
rs762805266 Chr.21: 34709439-34709439 on Build GRCh38
rs760870660 Chr.21: 34709311-34709311 on Build GRCh38
rs2275969 Chr.21: 34709714-34709714 on Build GRCh38
rs146983202 Chr.21: 34709496-34709496 on Build GRCh38
rs768993623 Chr.21: 34709570-34709570 on Build GRCh38
rs987428155 Chr.21: 34709615-34709615 on Build GRCh38
rs1003184359 Chr.21: 34709528-34709528 on Build GRCh38
rs141654550 Chr.21: 34709673-34709673 on Build GRCh38
rs372559724 Chr.21: 34709555-34709555 on Build GRCh38
rs988608576 Chr.21: 34709275-34709275 on Build GRCh38
rs747599807 Chr.21: 34709567-34709567 on Build GRCh38
rs762078426 Chr.21: 34709498-34709498 on Build GRCh38
rs761498364 Chr.21: 34709715-34709715 on Build GRCh38
rs913508420 Chr.21: 34709661-34709661 on Build GRCh38
rs376511276 Chr.21: 34709425-34709425 on Build GRCh38
rs754621091 Chr.21: 34709453-34709453 on Build GRCh38
rs952636981 Chr.21: 34709530-34709530 on Build GRCh38
rs572899754 Chr.21: 34709554-34709554 on Build GRCh38
rs750345784 Chr.21: 34709430-34709430 on Build GRCh38
rs769858861 Chr.21: 34709500-34709500 on Build GRCh38
rs780393925 Chr.21: 34709566-34709566 on Build GRCh38
rs780413051 Chr.21: 34709490-34709490 on Build GRCh38
rs369275068 Chr.21: 34709343-34709343 on Build GRCh38
rs973730196 Chr.21: 34709701-34709701 on Build GRCh38
rs755674686 Chr.21: 34709481-34709481 on Build GRCh38
rs1052074449 Chr.21: 34709622-34709623 on Build GRCh38
rs759635739 Chr.21: 34709454-34709454 on Build GRCh38
rs1038090438 Chr.21: 34709433-34709433 on Build GRCh38
rs781602568 Chr.21: 34709574-34709574 on Build GRCh38
rs868597004 Chr.21: 34709540-34709540 on Build GRCh38
rs267606118 Chr.21: 34709388-34709388 on Build GRCh38
rs3171439 Chr.21: 34709480-34709480 on Build GRCh38
rs373486130 Chr.21: 34709383-34709383 on Build GRCh38
rs555164083 Chr.21: 34709268-34709268 on Build GRCh38
rs368821974 Chr.21: 34709489-34709489 on Build GRCh38
rs746438234 Chr.21: 34709408-34709408 on Build GRCh38

View Full Product Details