Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30667178-30667692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
UniGene
Hs.247186
Gene Chromosome Location
Chr.16: 30658142-30670810 on Build GRCh38
Transcripts
NM_001105079.2 XM_011545916.2 XM_011545917.2 XM_011545919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5706109 c.52-1G>A p.? Chr.16: 30667319-30667319 on Build GRCh38
3817815 c.220G>T p.G74* Chr.16: 30667592-30667592 on Build GRCh38
3957474 c.2004C>T p.H668H Chr.16: 30667552-30667552 on Build GRCh38
1721221 c.2055G>A p.L685L Chr.16: 30667603-30667603 on Build GRCh38
6002197 c.2013C>G p.A671A Chr.16: 30667561-30667561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971482438 Chr.16: 30667592-30667592 on Build GRCh38
rs773308863 Chr.16: 30667344-30667344 on Build GRCh38
rs552475434 Chr.16: 30667370-30667370 on Build GRCh38
rs370733333 Chr.16: 30667291-30667291 on Build GRCh38
rs539429225 Chr.16: 30667552-30667552 on Build GRCh38
rs1000037008 Chr.16: 30667556-30667556 on Build GRCh38
rs756901081 Chr.16: 30667605-30667605 on Build GRCh38
rs774653148 Chr.16: 30667575-30667575 on Build GRCh38
rs765347895 Chr.16: 30667343-30667343 on Build GRCh38
rs747347751 Chr.16: 30667391-30667391 on Build GRCh38
rs754944022 Chr.16: 30667369-30667369 on Build GRCh38
rs113847395 Chr.16: 30667216-30667216 on Build GRCh38
rs201854581 Chr.16: 30667414-30667414 on Build GRCh38
rs553091194 Chr.16: 30667354-30667354 on Build GRCh38
rs769264994 Chr.16: 30667314-30667314 on Build GRCh38
rs780332655 Chr.16: 30667388-30667388 on Build GRCh38
rs200584237 Chr.16: 30667479-30667479 on Build GRCh38
rs1020260028 Chr.16: 30667397-30667397 on Build GRCh38
rs774077258 Chr.16: 30667514-30667514 on Build GRCh38
rs34738501 Chr.16: 30667473-30667474 on Build GRCh38
rs1023999980 Chr.16: 30667583-30667583 on Build GRCh38
rs1039377165 Chr.16: 30667486-30667486 on Build GRCh38
rs1047379431 Chr.16: 30667267-30667267 on Build GRCh38
rs761971365 Chr.16: 30667327-30667327 on Build GRCh38
rs200777036 Chr.16: 30667471-30667471 on Build GRCh38
rs900314629 Chr.16: 30667495-30667495 on Build GRCh38
rs373092234 Chr.16: 30667385-30667385 on Build GRCh38
rs766588108 Chr.16: 30667481-30667481 on Build GRCh38
rs202186794 Chr.16: 30667591-30667591 on Build GRCh38
rs1009938957 Chr.16: 30667328-30667328 on Build GRCh38
rs944361668 Chr.16: 30667578-30667578 on Build GRCh38
rs143966902 Chr.16: 30667374-30667374 on Build GRCh38
rs568219271 Chr.16: 30667458-30667458 on Build GRCh38
rs878874812 Chr.16: 30667427-30667427 on Build GRCh38
rs973418432 Chr.16: 30667389-30667389 on Build GRCh38
rs200530945 Chr.16: 30667317-30667317 on Build GRCh38
rs1021621601 Chr.16: 30667333-30667333 on Build GRCh38
rs748596768 Chr.16: 30667432-30667432 on Build GRCh38
rs374614845 Chr.16: 30667373-30667373 on Build GRCh38
rs375914693 Chr.16: 30667379-30667379 on Build GRCh38
rs755398536 Chr.16: 30667662-30667662 on Build GRCh38
rs756887548 Chr.16: 30667386-30667386 on Build GRCh38
rs376987425 Chr.16: 30667571-30667571 on Build GRCh38
rs560051740 Chr.16: 30667301-30667301 on Build GRCh38
rs891590521 Chr.16: 30667572-30667572 on Build GRCh38
rs374123540 Chr.16: 30667553-30667553 on Build GRCh38
rs996412054 Chr.16: 30667620-30667620 on Build GRCh38
rs748925396 Chr.16: 30667383-30667383 on Build GRCh38
rs763535633 Chr.16: 30667580-30667580 on Build GRCh38
rs750037492 Chr.16: 30667641-30667641 on Build GRCh38
rs1010299815 Chr.16: 30667582-30667582 on Build GRCh38
rs778612975 Chr.16: 30667634-30667634 on Build GRCh38
rs372227421 Chr.16: 30667464-30667464 on Build GRCh38
rs1054524393 Chr.16: 30667637-30667637 on Build GRCh38
rs760470990 Chr.16: 30667372-30667372 on Build GRCh38
rs3747482 Chr.16: 30667638-30667638 on Build GRCh38
rs927858027 Chr.16: 30667429-30667429 on Build GRCh38
rs762127111 Chr.16: 30667643-30667643 on Build GRCh38
rs929303162 Chr.16: 30667421-30667421 on Build GRCh38
rs751343122 Chr.16: 30667365-30667365 on Build GRCh38
rs376786476 Chr.16: 30667468-30667468 on Build GRCh38
rs1033894121 Chr.16: 30667608-30667608 on Build GRCh38
rs753368876 Chr.16: 30667602-30667602 on Build GRCh38
rs566035880 Chr.16: 30667547-30667547 on Build GRCh38
rs1009110102 Chr.16: 30667297-30667297 on Build GRCh38
rs1032781500 Chr.16: 30667242-30667242 on Build GRCh38
rs549940654 Chr.16: 30667330-30667330 on Build GRCh38
rs529792238 Chr.16: 30667241-30667241 on Build GRCh38
rs367601169 Chr.16: 30667415-30667415 on Build GRCh38
rs971808080 Chr.16: 30667410-30667410 on Build GRCh38
rs930405669 Chr.16: 30667217-30667217 on Build GRCh38
rs1041033448 Chr.16: 30667299-30667299 on Build GRCh38
rs776917556 Chr.16: 30667325-30667325 on Build GRCh38
rs541460713 Chr.16: 30667261-30667261 on Build GRCh38
rs34604074 Chr.16: 30667343-30667344 on Build GRCh38

View Full Product Details