Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17069233-17069740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1 XM_017013343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411701 c.545C>T p.T182I Chr.8: 17069697-17069697 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs114137374 Chr.8: 17069492-17069492 on Build GRCh38
rs768291985 Chr.8: 17069480-17069480 on Build GRCh38
rs867600533 Chr.8: 17069691-17069691 on Build GRCh38
rs754678282 Chr.8: 17069349-17069349 on Build GRCh38
rs369080878 Chr.8: 17069644-17069644 on Build GRCh38
rs1025014519 Chr.8: 17069571-17069572 on Build GRCh38
rs112061001 Chr.8: 17069317-17069317 on Build GRCh38
rs563035345 Chr.8: 17069357-17069357 on Build GRCh38
rs17686780 Chr.8: 17069510-17069510 on Build GRCh38
rs965115203 Chr.8: 17069466-17069466 on Build GRCh38
rs35402310 Chr.8: 17069374-17069375 on Build GRCh38
rs1038011272 Chr.8: 17069306-17069306 on Build GRCh38
rs960561876 Chr.8: 17069375-17069375 on Build GRCh38
rs199953621 Chr.8: 17069631-17069631 on Build GRCh38
rs117689011 Chr.8: 17069446-17069446 on Build GRCh38
rs777609948 Chr.8: 17069507-17069507 on Build GRCh38
rs540244215 Chr.8: 17069287-17069287 on Build GRCh38
rs751369104 Chr.8: 17069649-17069649 on Build GRCh38
rs779365540 Chr.8: 17069707-17069707 on Build GRCh38
rs1020762722 Chr.8: 17069414-17069414 on Build GRCh38
rs139785699 Chr.8: 17069594-17069594 on Build GRCh38
rs1005809705 Chr.8: 17069501-17069501 on Build GRCh38
rs531954053 Chr.8: 17069359-17069359 on Build GRCh38
rs555259881 Chr.8: 17069517-17069517 on Build GRCh38
rs879403978 Chr.8: 17069652-17069652 on Build GRCh38
rs146875492 Chr.8: 17069515-17069515 on Build GRCh38
rs752616162 Chr.8: 17069659-17069659 on Build GRCh38
rs917966071 Chr.8: 17069315-17069315 on Build GRCh38
rs774032453 Chr.8: 17069483-17069483 on Build GRCh38
rs184905691 Chr.8: 17069543-17069543 on Build GRCh38
rs979494522 Chr.8: 17069445-17069445 on Build GRCh38
rs781072337 Chr.8: 17069416-17069416 on Build GRCh38
rs754822698 Chr.8: 17069655-17069655 on Build GRCh38
rs936902056 Chr.8: 17069409-17069409 on Build GRCh38
rs755186716 Chr.8: 17069678-17069682 on Build GRCh38
rs551537618 Chr.8: 17069431-17069431 on Build GRCh38
rs568934346 Chr.8: 17069539-17069539 on Build GRCh38
rs373529901 Chr.8: 17069451-17069451 on Build GRCh38
rs115831455 Chr.8: 17069255-17069255 on Build GRCh38
rs527822440 Chr.8: 17069469-17069469 on Build GRCh38
rs1002310700 Chr.8: 17069546-17069546 on Build GRCh38
rs529588933 Chr.8: 17069311-17069311 on Build GRCh38
rs756193526 Chr.8: 17069666-17069666 on Build GRCh38
rs1054073425 Chr.8: 17069418-17069418 on Build GRCh38
rs975016371 Chr.8: 17069470-17069470 on Build GRCh38
rs762414388 Chr.8: 17069304-17069304 on Build GRCh38
rs1020710412 Chr.8: 17069608-17069608 on Build GRCh38
rs577661992 Chr.8: 17069624-17069624 on Build GRCh38
rs149841074 Chr.8: 17069266-17069266 on Build GRCh38
rs540355495 Chr.8: 17069634-17069634 on Build GRCh38
rs879545740 Chr.8: 17069402-17069402 on Build GRCh38
rs1034052121 Chr.8: 17069358-17069358 on Build GRCh38
rs886879321 Chr.8: 17069290-17069290 on Build GRCh38
rs79880009 Chr.8: 17069585-17069585 on Build GRCh38
rs978247889 Chr.8: 17069328-17069328 on Build GRCh38
rs372444696 Chr.8: 17069658-17069658 on Build GRCh38
rs1055037478 Chr.8: 17069576-17069576 on Build GRCh38
rs188665033 Chr.8: 17069296-17069296 on Build GRCh38
rs757554095 Chr.8: 17069704-17069704 on Build GRCh38
rs924092197 Chr.8: 17069621-17069621 on Build GRCh38
rs778053188 Chr.8: 17069677-17069677 on Build GRCh38
rs749515238 Chr.8: 17069701-17069701 on Build GRCh38

View Full Product Details