Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168056364-168056856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD1
Entrez Gene ID
79981
Gene Name
FERM domain containing 1
Gene Aliases
bA164L23.1
UniGene
Hs.266746
Gene Chromosome Location
Chr.6: 168055745-168093007 on Build GRCh38
Transcripts
NM_001122841.2 NM_024919.4 NR_110312.1 XM_011536137.1 XM_011536138.1 XM_011536140.1 XM_011536141.1 XM_011536142.2 XM_011536143.1 XM_011536145.1 XM_011536146.2

SNP Detail

SNP ID Chromosome Location
rs918228953 Chr.6: 168056695-168056695 on Build GRCh38
rs985219756 Chr.6: 168056656-168056656 on Build GRCh38
rs1043018255 Chr.6: 168056403-168056403 on Build GRCh38
rs1057492606 Chr.6: 168056445-168056445 on Build GRCh38
rs146383340 Chr.6: 168056636-168056637 on Build GRCh38
rs937409727 Chr.6: 168056415-168056415 on Build GRCh38
rs779411720 Chr.6: 168056735-168056735 on Build GRCh38
rs1002347022 Chr.6: 168056489-168056489 on Build GRCh38
rs1026611665 Chr.6: 168056492-168056494 on Build GRCh38
rs79234806 Chr.6: 168056637-168056637 on Build GRCh38
rs1005162803 Chr.6: 168056471-168056471 on Build GRCh38
rs868492016 Chr.6: 168056548-168056548 on Build GRCh38
rs912486132 Chr.6: 168056657-168056657 on Build GRCh38
rs201906854 Chr.6: 168056632-168056633 on Build GRCh38
rs887930196 Chr.6: 168056487-168056487 on Build GRCh38
rs972413516 Chr.6: 168056691-168056691 on Build GRCh38
rs1055266454 Chr.6: 168056779-168056779 on Build GRCh38
rs576650912 Chr.6: 168056751-168056751 on Build GRCh38
rs560304576 Chr.6: 168056649-168056649 on Build GRCh38
rs905947482 Chr.6: 168056414-168056414 on Build GRCh38
rs1047398148 Chr.6: 168056501-168056501 on Build GRCh38
rs927310238 Chr.6: 168056672-168056672 on Build GRCh38
rs567843479 Chr.6: 168056715-168056715 on Build GRCh38
rs1007073473 Chr.6: 168056793-168056793 on Build GRCh38
rs3832455 Chr.6: 168056638-168056639 on Build GRCh38
rs1032398912 Chr.6: 168056604-168056604 on Build GRCh38
rs965258760 Chr.6: 168056685-168056685 on Build GRCh38
rs1006632831 Chr.6: 168056511-168056511 on Build GRCh38
rs148747421 Chr.6: 168056541-168056541 on Build GRCh38
rs896778047 Chr.6: 168056784-168056784 on Build GRCh38
rs899386829 Chr.6: 168056481-168056481 on Build GRCh38
rs534153093 Chr.6: 168056438-168056438 on Build GRCh38
rs528660794 Chr.6: 168056824-168056824 on Build GRCh38
rs192739754 Chr.6: 168056831-168056831 on Build GRCh38
rs759715985 Chr.6: 168056389-168056389 on Build GRCh38
rs542435642 Chr.6: 168056757-168056757 on Build GRCh38
rs1048567844 Chr.6: 168056758-168056758 on Build GRCh38
rs1049253578 Chr.6: 168056483-168056483 on Build GRCh38
rs940853253 Chr.6: 168056744-168056744 on Build GRCh38
rs780460065 Chr.6: 168056565-168056565 on Build GRCh38
rs937463857 Chr.6: 168056687-168056687 on Build GRCh38
rs931054984 Chr.6: 168056426-168056426 on Build GRCh38
rs6930196 Chr.6: 168056703-168056703 on Build GRCh38
rs761987445 Chr.6: 168056640-168056641 on Build GRCh38
rs889455079 Chr.6: 168056792-168056792 on Build GRCh38
rs1048633899 Chr.6: 168056745-168056745 on Build GRCh38
rs966841191 Chr.6: 168056585-168056585 on Build GRCh38
rs577713525 Chr.6: 168056586-168056586 on Build GRCh38
rs144641811 Chr.6: 168056637-168056638 on Build GRCh38
rs978742309 Chr.6: 168056653-168056653 on Build GRCh38
rs540785038 Chr.6: 168056814-168056814 on Build GRCh38
rs555880290 Chr.6: 168056572-168056572 on Build GRCh38
rs3734897 Chr.6: 168056599-168056599 on Build GRCh38
rs917434587 Chr.6: 168056708-168056708 on Build GRCh38
rs574233467 Chr.6: 168056517-168056517 on Build GRCh38
rs1036604242 Chr.6: 168056473-168056473 on Build GRCh38
rs555958314 Chr.6: 168056453-168056453 on Build GRCh38
rs869278819 Chr.6: 168056639-168056640 on Build GRCh38
rs755780229 Chr.6: 168056498-168056498 on Build GRCh38
rs928955355 Chr.6: 168056491-168056491 on Build GRCh38
rs968569960 Chr.6: 168056650-168056650 on Build GRCh38

View Full Product Details