Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 70139696-70140196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN8OS
Entrez Gene ID
6315
Gene Name
ATXN8 opposite strand (non-protein coding)
Gene Aliases
KLHL1AS, NCRNA00003, SCA8
UniGene
Hs.676453
Gene Chromosome Location
Chr.13: 70107213-70139753 on Build GRCh38
Transcripts
NR_002717.2

SNP Detail

SNP ID Chromosome Location
rs1052462623 Chr.13: 70139775-70139775 on Build GRCh38
rs183190581 Chr.13: 70139730-70139730 on Build GRCh38
rs565510968 Chr.13: 70139880-70139880 on Build GRCh38
rs899178646 Chr.13: 70140045-70140045 on Build GRCh38
rs190513896 Chr.13: 70139894-70139894 on Build GRCh38
rs940690871 Chr.13: 70139743-70139743 on Build GRCh38
rs148696111 Chr.13: 70139867-70139868 on Build GRCh38
rs911801967 Chr.13: 70139971-70139971 on Build GRCh38
rs987747235 Chr.13: 70139962-70139962 on Build GRCh38
rs972152255 Chr.13: 70139961-70139961 on Build GRCh38
rs890409884 Chr.13: 70139732-70139732 on Build GRCh38
rs933495246 Chr.13: 70139979-70139979 on Build GRCh38
rs1000448475 Chr.13: 70140056-70140056 on Build GRCh38
rs765729430 Chr.13: 70139838-70139838 on Build GRCh38
rs1036289554 Chr.13: 70140034-70140034 on Build GRCh38
rs530177763 Chr.13: 70139744-70139744 on Build GRCh38
rs866020274 Chr.13: 70140062-70140062 on Build GRCh38
rs1039393323 Chr.13: 70139828-70139828 on Build GRCh38
rs940637476 Chr.13: 70140003-70140003 on Build GRCh38
rs185792331 Chr.13: 70139885-70139885 on Build GRCh38
rs930687340 Chr.13: 70140068-70140068 on Build GRCh38
rs919462236 Chr.13: 70139754-70139754 on Build GRCh38
rs892698132 Chr.13: 70140154-70140154 on Build GRCh38
rs1045150489 Chr.13: 70140080-70140080 on Build GRCh38
rs981078693 Chr.13: 70139922-70139922 on Build GRCh38
rs903525942 Chr.13: 70139830-70139830 on Build GRCh38
rs969362620 Chr.13: 70139881-70139882 on Build GRCh38
rs1003701195 Chr.13: 70140126-70140126 on Build GRCh38
rs61964572 Chr.13: 70139745-70139745 on Build GRCh38
rs943672241 Chr.13: 70140049-70140049 on Build GRCh38
rs1043281167 Chr.13: 70140050-70140050 on Build GRCh38
rs1013741552 Chr.13: 70140105-70140105 on Build GRCh38
rs749709812 Chr.13: 70140059-70140059 on Build GRCh38
rs903536028 Chr.13: 70140055-70140055 on Build GRCh38
rs302017 Chr.13: 70139802-70139802 on Build GRCh38
rs933472668 Chr.13: 70139758-70139758 on Build GRCh38
rs1004156861 Chr.13: 70139833-70139833 on Build GRCh38
rs113131589 Chr.13: 70139958-70139958 on Build GRCh38
rs1025325057 Chr.13: 70139875-70139875 on Build GRCh38
rs909201205 Chr.13: 70139724-70139724 on Build GRCh38
rs978516013 Chr.13: 70139902-70139902 on Build GRCh38
rs899239349 Chr.13: 70139759-70139759 on Build GRCh38
rs1024834083 Chr.13: 70140113-70140113 on Build GRCh38
rs1057056578 Chr.13: 70140143-70140143 on Build GRCh38
rs750895955 Chr.13: 70139883-70139883 on Build GRCh38
rs956541707 Chr.13: 70139857-70139857 on Build GRCh38
rs905309508 Chr.13: 70140109-70140109 on Build GRCh38
rs1009836747 Chr.13: 70140156-70140156 on Build GRCh38
rs547057225 Chr.13: 70140020-70140020 on Build GRCh38
rs921722401 Chr.13: 70139929-70139929 on Build GRCh38
rs919305961 Chr.13: 70139963-70139963 on Build GRCh38
rs75410142 Chr.13: 70139982-70139982 on Build GRCh38
rs559289884 Chr.13: 70139807-70139807 on Build GRCh38
rs564982331 Chr.13: 70139884-70139884 on Build GRCh38
rs905361087 Chr.13: 70139823-70139823 on Build GRCh38
rs889342847 Chr.13: 70139788-70139788 on Build GRCh38

View Full Product Details