Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 121570819-121571368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMBP1
Entrez Gene ID
647121
Gene Name
embigin pseudogene 1
Gene Aliases
-
UniGene
Hs.697682
Gene Chromosome Location
Chr.1: 121519112-121571888 on Build GRCh38
Transcripts
NR_003955.1

SNP Detail

SNP ID Chromosome Location
rs587651333 Chr.1: 121571062-121571062 on Build GRCh38
rs138515587 Chr.1: 121571253-121571253 on Build GRCh38
rs184309970 Chr.1: 121571180-121571180 on Build GRCh38
rs376762221 Chr.1: 121571097-121571097 on Build GRCh38
rs782215266 Chr.1: 121571000-121571000 on Build GRCh38
rs587605867 Chr.1: 121571034-121571034 on Build GRCh38
rs782234156 Chr.1: 121571120-121571121 on Build GRCh38
rs191760539 Chr.1: 121570888-121570888 on Build GRCh38
rs17842570 Chr.1: 121570889-121570889 on Build GRCh38
rs768442383 Chr.1: 121570875-121570875 on Build GRCh38
rs587688736 Chr.1: 121571139-121571139 on Build GRCh38
rs112805592 Chr.1: 121571113-121571113 on Build GRCh38
rs587766560 Chr.1: 121571148-121571148 on Build GRCh38
rs587629770 Chr.1: 121571106-121571106 on Build GRCh38
rs587728133 Chr.1: 121571345-121571345 on Build GRCh38
rs767727976 Chr.1: 121571250-121571250 on Build GRCh38
rs782530219 Chr.1: 121570913-121570913 on Build GRCh38
rs781799886 Chr.1: 121571164-121571164 on Build GRCh38
rs188325708 Chr.1: 121571322-121571322 on Build GRCh38
rs377764137 Chr.1: 121570911-121570911 on Build GRCh38
rs587696112 Chr.1: 121570861-121570861 on Build GRCh38
rs782659083 Chr.1: 121570919-121570919 on Build GRCh38
rs587735798 Chr.1: 121571079-121571079 on Build GRCh38
rs587699374 Chr.1: 121571026-121571026 on Build GRCh38
rs781907467 Chr.1: 121570953-121570953 on Build GRCh38
rs111312348 Chr.1: 121571283-121571283 on Build GRCh38

View Full Product Details