Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5241843-5242392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBBP1
Entrez Gene ID
3044
Gene Name
hemoglobin subunit beta pseudogene 1
Gene Aliases
HBH1, HBHP
UniGene
Hs.20205
Gene Chromosome Location
Chr.11: 5241954-5243592 on Build GRCh38
Transcripts
NR_001589.1

SNP Detail

SNP ID Chromosome Location
rs559268551 Chr.11: 5242022-5242022 on Build GRCh38
rs996915793 Chr.11: 5241919-5241919 on Build GRCh38
rs1037462348 Chr.11: 5241883-5241883 on Build GRCh38
rs764145541 Chr.11: 5242203-5242203 on Build GRCh38
rs539283587 Chr.11: 5242230-5242230 on Build GRCh38
rs775412388 Chr.11: 5242200-5242200 on Build GRCh38
rs10128555 Chr.11: 5242347-5242347 on Build GRCh38
rs547059693 Chr.11: 5242069-5242090 on Build GRCh38
rs148999369 Chr.11: 5242368-5242368 on Build GRCh38
rs951850794 Chr.11: 5242104-5242104 on Build GRCh38
rs973187021 Chr.11: 5242358-5242358 on Build GRCh38
rs975027267 Chr.11: 5242081-5242081 on Build GRCh38
rs112863595 Chr.11: 5242105-5242105 on Build GRCh38
rs984608285 Chr.11: 5242322-5242322 on Build GRCh38
rs368092467 Chr.11: 5242262-5242262 on Build GRCh38
rs909949092 Chr.11: 5242125-5242125 on Build GRCh38
rs1022042410 Chr.11: 5241907-5241907 on Build GRCh38
rs71480748 Chr.11: 5241945-5241945 on Build GRCh38
rs951964800 Chr.11: 5242315-5242315 on Build GRCh38
rs1029249947 Chr.11: 5241943-5241943 on Build GRCh38
rs150292763 Chr.11: 5242202-5242202 on Build GRCh38
rs765399666 Chr.11: 5242227-5242227 on Build GRCh38
rs368425028 Chr.11: 5241899-5241899 on Build GRCh38
rs774428496 Chr.11: 5242020-5242020 on Build GRCh38
rs895276905 Chr.11: 5241926-5241926 on Build GRCh38
rs61729466 Chr.11: 5242219-5242219 on Build GRCh38
rs543635565 Chr.11: 5241968-5241968 on Build GRCh38
rs1010686767 Chr.11: 5241901-5241901 on Build GRCh38
rs1011066734 Chr.11: 5241928-5241928 on Build GRCh38
rs576981726 Chr.11: 5242331-5242331 on Build GRCh38
rs57730466 Chr.11: 5242306-5242306 on Build GRCh38
rs939234207 Chr.11: 5242115-5242115 on Build GRCh38
rs866957866 Chr.11: 5242088-5242088 on Build GRCh38
rs980808062 Chr.11: 5242340-5242340 on Build GRCh38
rs529634190 Chr.11: 5242050-5242050 on Build GRCh38
rs762919249 Chr.11: 5241872-5241872 on Build GRCh38
rs555081294 Chr.11: 5241898-5241898 on Build GRCh38
rs750716854 Chr.11: 5242095-5242095 on Build GRCh38
rs73402614 Chr.11: 5242073-5242073 on Build GRCh38
rs969842055 Chr.11: 5242337-5242337 on Build GRCh38
rs566393640 Chr.11: 5242271-5242271 on Build GRCh38
rs946869863 Chr.11: 5242250-5242250 on Build GRCh38
rs973537954 Chr.11: 5242300-5242300 on Build GRCh38
rs796497268 Chr.11: 5242285-5242285 on Build GRCh38
rs973487201 Chr.11: 5242063-5242063 on Build GRCh38
rs1052335537 Chr.11: 5242151-5242151 on Build GRCh38
rs372023319 Chr.11: 5242176-5242176 on Build GRCh38
rs565124414 Chr.11: 5241992-5241992 on Build GRCh38
rs755604678 Chr.11: 5242291-5242291 on Build GRCh38
rs902703739 Chr.11: 5242276-5242276 on Build GRCh38
rs940127210 Chr.11: 5242148-5242148 on Build GRCh38
rs543117467 Chr.11: 5241896-5241896 on Build GRCh38
rs564147471 Chr.11: 5242209-5242209 on Build GRCh38
rs766847759 Chr.11: 5242263-5242263 on Build GRCh38
rs955095761 Chr.11: 5241948-5241948 on Build GRCh38
rs762292835 Chr.11: 5242025-5242025 on Build GRCh38
rs1055779701 Chr.11: 5242207-5242207 on Build GRCh38
rs187692483 Chr.11: 5242162-5242162 on Build GRCh38
rs901471755 Chr.11: 5242294-5242294 on Build GRCh38
rs750758180 Chr.11: 5242235-5242235 on Build GRCh38
rs537545908 Chr.11: 5242324-5242324 on Build GRCh38
rs928076886 Chr.11: 5242366-5242366 on Build GRCh38
rs144906987 Chr.11: 5242281-5242281 on Build GRCh38
rs962217723 Chr.11: 5242298-5242298 on Build GRCh38
rs987613439 Chr.11: 5241990-5241990 on Build GRCh38
rs140236872 Chr.11: 5242003-5242003 on Build GRCh38
rs772008175 Chr.11: 5242188-5242188 on Build GRCh38
rs757015006 Chr.11: 5241976-5241980 on Build GRCh38
rs540976995 Chr.11: 5242012-5242012 on Build GRCh38
rs536635210 Chr.11: 5242278-5242278 on Build GRCh38
rs948117016 Chr.11: 5242084-5242084 on Build GRCh38
rs939323236 Chr.11: 5242348-5242348 on Build GRCh38
rs757820140 Chr.11: 5242350-5242350 on Build GRCh38
rs558678517 Chr.11: 5242325-5242325 on Build GRCh38
rs113355107 Chr.11: 5242091-5242112 on Build GRCh38
rs963781744 Chr.11: 5241917-5241917 on Build GRCh38

View Full Product Details