Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5243352-5243916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBBP1
Entrez Gene ID
3044
Gene Name
hemoglobin subunit beta pseudogene 1
Gene Aliases
HBH1, HBHP
UniGene
Hs.20205
Gene Chromosome Location
Chr.11: 5241954-5243592 on Build GRCh38
Transcripts
NR_001589.1

SNP Detail

SNP ID Chromosome Location
rs531282235 Chr.11: 5243867-5243867 on Build GRCh38
rs750282920 Chr.11: 5243672-5243672 on Build GRCh38
rs938358843 Chr.11: 5243615-5243616 on Build GRCh38
rs570834457 Chr.11: 5243879-5243879 on Build GRCh38
rs545474276 Chr.11: 5243537-5243537 on Build GRCh38
rs74910558 Chr.11: 5243692-5243692 on Build GRCh38
rs1026798510 Chr.11: 5243451-5243451 on Build GRCh38
rs763572149 Chr.11: 5243750-5243750 on Build GRCh38
rs1030583269 Chr.11: 5243535-5243535 on Build GRCh38
rs888321185 Chr.11: 5243752-5243752 on Build GRCh38
rs113974489 Chr.11: 5243453-5243453 on Build GRCh38
rs1052521053 Chr.11: 5243708-5243708 on Build GRCh38
rs948538284 Chr.11: 5243423-5243423 on Build GRCh38
rs552848035 Chr.11: 5243434-5243434 on Build GRCh38
rs540332487 Chr.11: 5243585-5243585 on Build GRCh38
rs535122104 Chr.11: 5243384-5243384 on Build GRCh38
rs1016535906 Chr.11: 5243828-5243828 on Build GRCh38
rs112359560 Chr.11: 5243890-5243890 on Build GRCh38
rs535334842 Chr.11: 5243455-5243455 on Build GRCh38
rs560715511 Chr.11: 5243798-5243798 on Build GRCh38
rs763527386 Chr.11: 5243476-5243476 on Build GRCh38
rs12417960 Chr.11: 5243876-5243876 on Build GRCh38
rs751017106 Chr.11: 5243610-5243610 on Build GRCh38
rs944051901 Chr.11: 5243682-5243682 on Build GRCh38
rs867974979 Chr.11: 5243414-5243414 on Build GRCh38
rs925496948 Chr.11: 5243561-5243561 on Build GRCh38
rs141484342 Chr.11: 5243857-5243857 on Build GRCh38
rs1051738830 Chr.11: 5243769-5243769 on Build GRCh38
rs572752236 Chr.11: 5243659-5243659 on Build GRCh38
rs113474492 Chr.11: 5243479-5243479 on Build GRCh38
rs557784327 Chr.11: 5243539-5243539 on Build GRCh38
rs7934275 Chr.11: 5243699-5243699 on Build GRCh38
rs1057041081 Chr.11: 5243395-5243395 on Build GRCh38
rs749192921 Chr.11: 5243791-5243791 on Build GRCh38
rs992927696 Chr.11: 5243636-5243636 on Build GRCh38
rs925749005 Chr.11: 5243697-5243697 on Build GRCh38
rs1007776956 Chr.11: 5243784-5243784 on Build GRCh38
rs992454513 Chr.11: 5243493-5243493 on Build GRCh38
rs188968413 Chr.11: 5243540-5243540 on Build GRCh38
rs918180902 Chr.11: 5243536-5243536 on Build GRCh38
rs936819946 Chr.11: 5243562-5243562 on Build GRCh38
rs889819802 Chr.11: 5243404-5243404 on Build GRCh38
rs963571976 Chr.11: 5243851-5243851 on Build GRCh38
rs948609865 Chr.11: 5243661-5243661 on Build GRCh38
rs911291400 Chr.11: 5243651-5243651 on Build GRCh38
rs1040945790 Chr.11: 5243424-5243424 on Build GRCh38
rs959743015 Chr.11: 5243810-5243810 on Build GRCh38
rs575189932 Chr.11: 5243496-5243496 on Build GRCh38
rs1049868808 Chr.11: 5243615-5243615 on Build GRCh38
rs889141221 Chr.11: 5243773-5243773 on Build GRCh38
rs1048776391 Chr.11: 5243740-5243740 on Build GRCh38
rs994069341 Chr.11: 5243723-5243723 on Build GRCh38
rs897046572 Chr.11: 5243433-5243433 on Build GRCh38
rs1019938702 Chr.11: 5243842-5243842 on Build GRCh38
rs1013942759 Chr.11: 5243816-5243816 on Build GRCh38
rs756880377 Chr.11: 5243655-5243655 on Build GRCh38
rs542471116 Chr.11: 5243809-5243809 on Build GRCh38
rs915814587 Chr.11: 5243648-5243648 on Build GRCh38
rs978365790 Chr.11: 5243873-5243873 on Build GRCh38
rs767255679 Chr.11: 5243660-5243660 on Build GRCh38
rs193070288 Chr.11: 5243548-5243548 on Build GRCh38
rs374630136 Chr.11: 5243688-5243688 on Build GRCh38
rs959774000 Chr.11: 5243487-5243487 on Build GRCh38
rs963544624 Chr.11: 5243543-5243543 on Build GRCh38

View Full Product Details