Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46530786-46531357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
-
UniGene
Hs.97414
Gene Chromosome Location
Chr.16: 46469337-46569097 on Build GRCh38
Transcripts
NR_026556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299249 c.797+2T>C p.? Chr.16: 46531102-46531102 on Build GRCh38
5955630 c.772G>C p.E258Q Chr.16: 46531129-46531129 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886421249 Chr.16: 46530888-46530888 on Build GRCh38
rs989059400 Chr.16: 46530994-46530994 on Build GRCh38
rs573824910 Chr.16: 46530939-46530939 on Build GRCh38
rs767889131 Chr.16: 46530956-46530956 on Build GRCh38
rs1047375899 Chr.16: 46531311-46531311 on Build GRCh38
rs1011882761 Chr.16: 46530895-46530898 on Build GRCh38
rs763231520 Chr.16: 46530869-46530869 on Build GRCh38
rs753167109 Chr.16: 46531100-46531100 on Build GRCh38
rs909760422 Chr.16: 46531316-46531316 on Build GRCh38
rs902649716 Chr.16: 46531086-46531086 on Build GRCh38
rs947066488 Chr.16: 46531000-46531000 on Build GRCh38
rs556151513 Chr.16: 46531228-46531228 on Build GRCh38
rs887474096 Chr.16: 46531215-46531215 on Build GRCh38
rs531648745 Chr.16: 46530950-46530950 on Build GRCh38
rs978365881 Chr.16: 46531249-46531249 on Build GRCh38
rs766443647 Chr.16: 46530885-46530885 on Build GRCh38
rs929817593 Chr.16: 46531292-46531292 on Build GRCh38
rs1034735935 Chr.16: 46530832-46530832 on Build GRCh38
rs992516316 Chr.16: 46531272-46531272 on Build GRCh38
rs576305194 Chr.16: 46531312-46531312 on Build GRCh38
rs951978108 Chr.16: 46530907-46530907 on Build GRCh38
rs985150662 Chr.16: 46530911-46530911 on Build GRCh38
rs954888144 Chr.16: 46531181-46531181 on Build GRCh38
rs772507024 Chr.16: 46530984-46530984 on Build GRCh38
rs926977995 Chr.16: 46531250-46531250 on Build GRCh38
rs139414629 Chr.16: 46530862-46530862 on Build GRCh38
rs1056867022 Chr.16: 46531032-46531032 on Build GRCh38
rs937174860 Chr.16: 46530831-46530831 on Build GRCh38
rs996274958 Chr.16: 46531153-46531153 on Build GRCh38
rs781684743 Chr.16: 46531090-46531090 on Build GRCh38
rs532336584 Chr.16: 46531318-46531319 on Build GRCh38
rs946983255 Chr.16: 46531066-46531066 on Build GRCh38
rs1056897905 Chr.16: 46530835-46530835 on Build GRCh38
rs1030195320 Chr.16: 46531290-46531290 on Build GRCh38
rs895672987 Chr.16: 46531037-46531037 on Build GRCh38
rs978438023 Chr.16: 46531007-46531007 on Build GRCh38
rs1020533123 Chr.16: 46531235-46531235 on Build GRCh38
rs745310526 Chr.16: 46531248-46531248 on Build GRCh38
rs146490332 Chr.16: 46531329-46531329 on Build GRCh38
rs933948874 Chr.16: 46531028-46531028 on Build GRCh38
rs577088526 Chr.16: 46531213-46531213 on Build GRCh38
rs985461036 Chr.16: 46531031-46531031 on Build GRCh38
rs955099089 Chr.16: 46530976-46530976 on Build GRCh38
rs771772888 Chr.16: 46531271-46531271 on Build GRCh38
rs575969440 Chr.16: 46530897-46530897 on Build GRCh38
rs911254163 Chr.16: 46531064-46531064 on Build GRCh38
rs1436436 Chr.16: 46531107-46531107 on Build GRCh38
rs1010856291 Chr.16: 46531191-46531191 on Build GRCh38
rs1016347910 Chr.16: 46530924-46530924 on Build GRCh38
rs778485834 Chr.16: 46531170-46531170 on Build GRCh38
rs961878437 Chr.16: 46530952-46530952 on Build GRCh38
rs190812744 Chr.16: 46531269-46531269 on Build GRCh38
rs913502959 Chr.16: 46530999-46530999 on Build GRCh38
rs1006100475 Chr.16: 46531218-46531218 on Build GRCh38
rs371823018 Chr.16: 46531125-46531125 on Build GRCh38
rs557941054 Chr.16: 46531282-46531282 on Build GRCh38
rs937101266 Chr.16: 46531030-46531030 on Build GRCh38
rs564350261 Chr.16: 46530881-46530881 on Build GRCh38
rs941282402 Chr.16: 46531322-46531322 on Build GRCh38
rs374529002 Chr.16: 46531075-46531075 on Build GRCh38
rs927008024 Chr.16: 46531013-46531013 on Build GRCh38
rs888395013 Chr.16: 46531099-46531099 on Build GRCh38
rs929820571 Chr.16: 46531068-46531068 on Build GRCh38
rs539734870 Chr.16: 46531279-46531279 on Build GRCh38
rs999197701 Chr.16: 46530815-46530815 on Build GRCh38
rs549859836 Chr.16: 46530953-46530953 on Build GRCh38
rs964828622 Chr.16: 46530946-46530946 on Build GRCh38
rs552462122 Chr.16: 46530863-46530863 on Build GRCh38
rs765337050 Chr.16: 46530812-46530812 on Build GRCh38

View Full Product Details