Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62829842-62830113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP1
Entrez Gene ID
23239
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
PHLPP, PLEKHE1, PPM3A, SCOP
UniGene
Hs.465337
Gene Chromosome Location
Chr.18: 62715439-62980443 on Build GRCh38
Transcripts
NM_194449.3 XM_011525886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5819376 c.1603G>T p.E535* Chr.18: 62830061-62830061 on Build GRCh38
5520265 c.1577-10T>A p.? Chr.18: 62830025-62830025 on Build GRCh38
5819377 c.64G>T p.E22* Chr.18: 62830061-62830061 on Build GRCh38
5520266 c.28T>A p.F10I Chr.18: 62830025-62830025 on Build GRCh38
5343147 c.1637G>A p.R546H Chr.18: 62830095-62830095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371701925 Chr.18: 62830056-62830056 on Build GRCh38
rs908016814 Chr.18: 62829915-62829915 on Build GRCh38
rs368316175 Chr.18: 62830029-62830029 on Build GRCh38
rs764143063 Chr.18: 62830032-62830032 on Build GRCh38
rs373755664 Chr.18: 62830059-62830059 on Build GRCh38
rs957519255 Chr.18: 62830003-62830003 on Build GRCh38
rs928862111 Chr.18: 62829946-62829948 on Build GRCh38
rs946408873 Chr.18: 62829975-62829975 on Build GRCh38
rs367782479 Chr.18: 62830088-62830088 on Build GRCh38
rs79067482 Chr.18: 62830051-62830051 on Build GRCh38
rs1055170420 Chr.18: 62829960-62829960 on Build GRCh38
rs370875793 Chr.18: 62830045-62830045 on Build GRCh38
rs940854823 Chr.18: 62829916-62829916 on Build GRCh38
rs996883947 Chr.18: 62829930-62829930 on Build GRCh38
rs369896541 Chr.18: 62830064-62830064 on Build GRCh38
rs764541037 Chr.18: 62829956-62829956 on Build GRCh38
rs779610862 Chr.18: 62830065-62830065 on Build GRCh38
rs1031455069 Chr.18: 62829982-62829982 on Build GRCh38
rs138214330 Chr.18: 62829871-62829871 on Build GRCh38
rs576009064 Chr.18: 62829929-62829929 on Build GRCh38
rs555274400 Chr.18: 62829972-62829972 on Build GRCh38
rs145321935 Chr.18: 62829996-62829997 on Build GRCh38
rs201614913 Chr.18: 62830050-62830050 on Build GRCh38
rs890848234 Chr.18: 62829964-62829964 on Build GRCh38
rs984844955 Chr.18: 62829901-62829901 on Build GRCh38
rs752943874 Chr.18: 62829991-62829991 on Build GRCh38
rs187964040 Chr.18: 62829990-62829990 on Build GRCh38

View Full Product Details