Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56236905-56237166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A5
Entrez Gene ID
283375
Gene Name
solute carrier family 39 member 5
Gene Aliases
LZT-Hs7, MYP24, ZIP5
UniGene
Hs.591018
Gene Chromosome Location
Chr.12: 56230036-56237846 on Build GRCh38
Transcripts
NM_001135195.1 NM_173596.2 XM_005268803.1 XM_011538198.1 XM_011538199.1 XM_011538200.1 XM_011538201.1 XM_017019185.1 XM_017019186.1 XM_017019187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6782177 c.1282G>A p.E428K Chr.12: 56237005-56237005 on Build GRCh38
4854299 c.1295T>C p.F432S Chr.12: 56237156-56237156 on Build GRCh38
4043565 c.1210G>A p.A404T Chr.12: 56236933-56236933 on Build GRCh38
1747150 c.1251C>T p.T417T Chr.12: 56236974-56236974 on Build GRCh38
1363075 c.1293C>A p.D431E Chr.12: 56237154-56237154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754485689 Chr.12: 56237020-56237020 on Build GRCh38
rs771372400 Chr.12: 56236960-56236960 on Build GRCh38
rs578116860 Chr.12: 56236979-56236979 on Build GRCh38
rs370112874 Chr.12: 56236927-56236927 on Build GRCh38
rs376349643 Chr.12: 56237059-56237059 on Build GRCh38
rs143449387 Chr.12: 56237004-56237004 on Build GRCh38
rs531957758 Chr.12: 56237144-56237144 on Build GRCh38
rs753590060 Chr.12: 56237129-56237129 on Build GRCh38
rs750169649 Chr.12: 56237005-56237005 on Build GRCh38
rs755802300 Chr.12: 56237007-56237007 on Build GRCh38
rs549345170 Chr.12: 56237089-56237089 on Build GRCh38
rs748747916 Chr.12: 56236952-56236952 on Build GRCh38
rs770088607 Chr.12: 56237102-56237102 on Build GRCh38
rs764042659 Chr.12: 56236981-56236981 on Build GRCh38
rs371513876 Chr.12: 56237034-56237034 on Build GRCh38
rs754643129 Chr.12: 56237140-56237140 on Build GRCh38
rs370427401 Chr.12: 56237106-56237106 on Build GRCh38
rs376657405 Chr.12: 56236959-56236959 on Build GRCh38
rs925951973 Chr.12: 56237009-56237009 on Build GRCh38
rs556197547 Chr.12: 56237048-56237048 on Build GRCh38
rs768764585 Chr.12: 56237111-56237111 on Build GRCh38
rs944592747 Chr.12: 56236973-56236973 on Build GRCh38
rs368608134 Chr.12: 56237056-56237056 on Build GRCh38
rs1005254235 Chr.12: 56237134-56237134 on Build GRCh38
rs750130241 Chr.12: 56236931-56236931 on Build GRCh38
rs17118409 Chr.12: 56236980-56236980 on Build GRCh38
rs544593155 Chr.12: 56236938-56236938 on Build GRCh38
rs778539711 Chr.12: 56237029-56237029 on Build GRCh38
rs753426688 Chr.12: 56237016-56237016 on Build GRCh38
rs776859931 Chr.12: 56236961-56236961 on Build GRCh38
rs761708318 Chr.12: 56236986-56236986 on Build GRCh38
rs373287583 Chr.12: 56236934-56236934 on Build GRCh38
rs1054109565 Chr.12: 56237088-56237088 on Build GRCh38
rs527862211 Chr.12: 56237012-56237012 on Build GRCh38
rs539450197 Chr.12: 56237031-56237031 on Build GRCh38
rs910366213 Chr.12: 56236940-56236940 on Build GRCh38
rs984781825 Chr.12: 56236965-56236965 on Build GRCh38
rs368515048 Chr.12: 56237105-56237105 on Build GRCh38
rs967915711 Chr.12: 56236933-56236933 on Build GRCh38
rs201975982 Chr.12: 56236926-56236926 on Build GRCh38
rs759863433 Chr.12: 56236969-56236969 on Build GRCh38
rs374778265 Chr.12: 56237114-56237114 on Build GRCh38
rs892416039 Chr.12: 56237095-56237095 on Build GRCh38
rs780685715 Chr.12: 56237065-56237065 on Build GRCh38
rs200448390 Chr.12: 56236923-56236923 on Build GRCh38
rs368085387 Chr.12: 56237118-56237118 on Build GRCh38
rs774410618 Chr.12: 56237112-56237112 on Build GRCh38
rs766148953 Chr.12: 56237122-56237122 on Build GRCh38
rs747589203 Chr.12: 56237043-56237043 on Build GRCh38
rs749351237 Chr.12: 56237120-56237120 on Build GRCh38
rs751548975 Chr.12: 56236984-56236984 on Build GRCh38

View Full Product Details