Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165595003-165595276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP3
Entrez Gene ID
80034
Gene Name
cysteine and serine rich nuclear protein 3
Gene Aliases
FAM130A2, PPP1R73, TAIP-2, TAIP2
UniGene
Hs.470479
Gene Chromosome Location
Chr.2: 165469647-165689407 on Build GRCh38
Transcripts
NM_001172173.1 NM_024969.3 XM_005246865.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1235999 c.144T>A p.F48L Chr.2: 165595209-165595209 on Build GRCh38
1008656 c.148C>T p.P50S Chr.2: 165595213-165595213 on Build GRCh38
6857185 c.33A>C p.E11D Chr.2: 165595098-165595098 on Build GRCh38
717085 c.49C>A p.P17T Chr.2: 165595114-165595114 on Build GRCh38
6446509 c.31G>C p.E11Q Chr.2: 165595096-165595096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908678127 Chr.2: 165595114-165595114 on Build GRCh38
rs754892916 Chr.2: 165595063-165595063 on Build GRCh38
rs138994502 Chr.2: 165595144-165595144 on Build GRCh38
rs916998154 Chr.2: 165595228-165595228 on Build GRCh38
rs142366433 Chr.2: 165595115-165595115 on Build GRCh38
rs777098791 Chr.2: 165595164-165595164 on Build GRCh38
rs141741469 Chr.2: 165595170-165595170 on Build GRCh38
rs372562167 Chr.2: 165595061-165595061 on Build GRCh38
rs779314149 Chr.2: 165595091-165595091 on Build GRCh38
rs1028574050 Chr.2: 165595049-165595049 on Build GRCh38
rs758157548 Chr.2: 165595047-165595047 on Build GRCh38
rs865867186 Chr.2: 165595132-165595132 on Build GRCh38
rs533790698 Chr.2: 165595104-165595104 on Build GRCh38
rs764686470 Chr.2: 165595183-165595183 on Build GRCh38
rs561761060 Chr.2: 165595241-165595241 on Build GRCh38
rs867931644 Chr.2: 165595204-165595204 on Build GRCh38
rs769210504 Chr.2: 165595160-165595160 on Build GRCh38
rs764917749 Chr.2: 165595040-165595040 on Build GRCh38
rs761297751 Chr.2: 165595033-165595033 on Build GRCh38
rs766090032 Chr.2: 165595216-165595216 on Build GRCh38
rs553866853 Chr.2: 165595135-165595135 on Build GRCh38
rs200907525 Chr.2: 165595130-165595130 on Build GRCh38
rs766145732 Chr.2: 165595048-165595048 on Build GRCh38
rs751460443 Chr.2: 165595054-165595054 on Build GRCh38
rs763720126 Chr.2: 165595039-165595039 on Build GRCh38
rs201041137 Chr.2: 165595129-165595129 on Build GRCh38
rs75788285 Chr.2: 165595234-165595234 on Build GRCh38
rs991131183 Chr.2: 165595196-165595196 on Build GRCh38
rs369124260 Chr.2: 165595064-165595064 on Build GRCh38
rs750059661 Chr.2: 165595043-165595043 on Build GRCh38
rs557202355 Chr.2: 165595213-165595213 on Build GRCh38
rs757383748 Chr.2: 165595088-165595088 on Build GRCh38
rs73972107 Chr.2: 165595065-165595065 on Build GRCh38
rs754800837 Chr.2: 165595235-165595235 on Build GRCh38
rs199893751 Chr.2: 165595105-165595105 on Build GRCh38
rs940203900 Chr.2: 165595126-165595126 on Build GRCh38
rs772758656 Chr.2: 165595191-165595191 on Build GRCh38

View Full Product Details