Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58496468-58496733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3779293 c.803C>T p.S268F Chr.8: 58496709-58496709 on Build GRCh38
2719726 c.791A>G p.N264S Chr.8: 58496721-58496721 on Build GRCh38
5599372 c.819G>A p.L273L Chr.8: 58496693-58496693 on Build GRCh38
6639500 c.878C>T p.A293V Chr.8: 58496634-58496634 on Build GRCh38
2719724 c.858G>A p.S286S Chr.8: 58496654-58496654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569910870 Chr.8: 58496595-58496595 on Build GRCh38
rs1040725233 Chr.8: 58496511-58496511 on Build GRCh38
rs760487952 Chr.8: 58496628-58496628 on Build GRCh38
rs756548694 Chr.8: 58496686-58496686 on Build GRCh38
rs779616560 Chr.8: 58496553-58496553 on Build GRCh38
rs902658512 Chr.8: 58496572-58496572 on Build GRCh38
rs535428616 Chr.8: 58496624-58496624 on Build GRCh38
rs766417104 Chr.8: 58496655-58496655 on Build GRCh38
rs778199686 Chr.8: 58496694-58496694 on Build GRCh38
rs753362618 Chr.8: 58496704-58496705 on Build GRCh38
rs751787597 Chr.8: 58496580-58496580 on Build GRCh38
rs565488904 Chr.8: 58496706-58496706 on Build GRCh38
rs528732489 Chr.8: 58496661-58496661 on Build GRCh38
rs922273358 Chr.8: 58496513-58496513 on Build GRCh38
rs149183844 Chr.8: 58496533-58496538 on Build GRCh38
rs1003369754 Chr.8: 58496621-58496621 on Build GRCh38
rs142476981 Chr.8: 58496604-58496604 on Build GRCh38
rs201581713 Chr.8: 58496572-58496573 on Build GRCh38
rs768382295 Chr.8: 58496634-58496634 on Build GRCh38
rs745559202 Chr.8: 58496610-58496610 on Build GRCh38
rs1024942782 Chr.8: 58496697-58496697 on Build GRCh38
rs771684358 Chr.8: 58496623-58496623 on Build GRCh38
rs776499055 Chr.8: 58496641-58496641 on Build GRCh38
rs1041461475 Chr.8: 58496569-58496569 on Build GRCh38
rs942372487 Chr.8: 58496501-58496501 on Build GRCh38
rs373514835 Chr.8: 58496598-58496598 on Build GRCh38
rs945764471 Chr.8: 58496563-58496563 on Build GRCh38
rs895423346 Chr.8: 58496654-58496654 on Build GRCh38
rs386352295 Chr.8: 58496605-58496605 on Build GRCh38
rs545609682 Chr.8: 58496672-58496672 on Build GRCh38
rs560268189 Chr.8: 58496588-58496588 on Build GRCh38
rs753140939 Chr.8: 58496587-58496587 on Build GRCh38
rs531164077 Chr.8: 58496499-58496499 on Build GRCh38
rs761796743 Chr.8: 58496645-58496645 on Build GRCh38
rs755226965 Chr.8: 58496581-58496581 on Build GRCh38
rs767871705 Chr.8: 58496665-58496665 on Build GRCh38

View Full Product Details