Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25171479-25171704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER2
Entrez Gene ID
219287
Gene Name
APC membrane recruitment protein 2
Gene Aliases
FAM123A
UniGene
Hs.528335
Gene Chromosome Location
Chr.13: 25161679-25172456 on Build GRCh38
Transcripts
NM_152704.3 NM_199138.2 XM_005266279.3 XM_005266280.3 XM_017020415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362112 c.19C>T p.R7C Chr.13: 25171601-25171601 on Build GRCh38
6599151 c.120G>A p.A40A Chr.13: 25171500-25171500 on Build GRCh38
5951399 c.43G>A p.E15K Chr.13: 25171577-25171577 on Build GRCh38
5953076 c.24C>T p.G8G Chr.13: 25171596-25171596 on Build GRCh38
5806246 c.120G>C p.A40A Chr.13: 25171500-25171500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760756687 Chr.13: 25171541-25171541 on Build GRCh38
rs760624250 Chr.13: 25171604-25171604 on Build GRCh38
rs746237286 Chr.13: 25171578-25171578 on Build GRCh38
rs919685899 Chr.13: 25171617-25171617 on Build GRCh38
rs761781237 Chr.13: 25171503-25171503 on Build GRCh38
rs77396502 Chr.13: 25171545-25171545 on Build GRCh38
rs779764803 Chr.13: 25171509-25171509 on Build GRCh38
rs753516298 Chr.13: 25171511-25171511 on Build GRCh38
rs750366641 Chr.13: 25171539-25171539 on Build GRCh38
rs368035363 Chr.13: 25171531-25171531 on Build GRCh38
rs774275570 Chr.13: 25171505-25171506 on Build GRCh38
rs752386259 Chr.13: 25171566-25171566 on Build GRCh38
rs747625357 Chr.13: 25171516-25171516 on Build GRCh38
rs969047023 Chr.13: 25171592-25171592 on Build GRCh38
rs765128528 Chr.13: 25171627-25171627 on Build GRCh38
rs747510016 Chr.13: 25171569-25171569 on Build GRCh38
rs749574835 Chr.13: 25171598-25171598 on Build GRCh38
rs772527302 Chr.13: 25171518-25171518 on Build GRCh38
rs770015071 Chr.13: 25171589-25171589 on Build GRCh38
rs775715674 Chr.13: 25171560-25171560 on Build GRCh38
rs757730969 Chr.13: 25171663-25171663 on Build GRCh38
rs747561489 Chr.13: 25171523-25171523 on Build GRCh38
rs757705946 Chr.13: 25171574-25171574 on Build GRCh38
rs765004677 Chr.13: 25171650-25171650 on Build GRCh38
rs778453019 Chr.13: 25171560-25171560 on Build GRCh38
rs377030859 Chr.13: 25171602-25171602 on Build GRCh38
rs763524108 Chr.13: 25171661-25171661 on Build GRCh38
rs772077267 Chr.13: 25171547-25171547 on Build GRCh38
rs759614388 Chr.13: 25171506-25171511 on Build GRCh38
rs566825438 Chr.13: 25171526-25171526 on Build GRCh38
rs535609841 Chr.13: 25171527-25171527 on Build GRCh38
rs970084439 Chr.13: 25171664-25171664 on Build GRCh38
rs753581586 Chr.13: 25171553-25171553 on Build GRCh38
rs901661430 Chr.13: 25171588-25171588 on Build GRCh38
rs112746394 Chr.13: 25171524-25171524 on Build GRCh38
rs757882312 Chr.13: 25171567-25171567 on Build GRCh38
rs201715479 Chr.13: 25171501-25171501 on Build GRCh38
rs759220184 Chr.13: 25171618-25171618 on Build GRCh38
rs754601093 Chr.13: 25171554-25171554 on Build GRCh38
rs766193562 Chr.13: 25171552-25171552 on Build GRCh38
rs776438191 Chr.13: 25171611-25171611 on Build GRCh38
rs766241390 Chr.13: 25171605-25171605 on Build GRCh38
rs775586742 Chr.13: 25171535-25171535 on Build GRCh38
rs200277642 Chr.13: 25171507-25171507 on Build GRCh38
rs372290519 Chr.13: 25171532-25171532 on Build GRCh38
rs752331057 Chr.13: 25171646-25171646 on Build GRCh38
rs758111613 Chr.13: 25171655-25171655 on Build GRCh38
rs1039997001 Chr.13: 25171680-25171680 on Build GRCh38
rs777321292 Chr.13: 25171568-25171568 on Build GRCh38
rs781559385 Chr.13: 25171577-25171577 on Build GRCh38
rs752722281 Chr.13: 25171502-25171502 on Build GRCh38
rs199742395 Chr.13: 25171520-25171520 on Build GRCh38
rs778420590 Chr.13: 25171514-25171514 on Build GRCh38
rs558386017 Chr.13: 25171588-25171590 on Build GRCh38
rs930121890 Chr.13: 25171619-25171619 on Build GRCh38
rs930289558 Chr.13: 25171512-25171512 on Build GRCh38
rs781578513 Chr.13: 25171667-25171667 on Build GRCh38
rs768846115 Chr.13: 25171600-25171600 on Build GRCh38
rs944268781 Chr.13: 25171679-25171679 on Build GRCh38
rs1022936962 Chr.13: 25171647-25171647 on Build GRCh38
rs759732982 Chr.13: 25171500-25171500 on Build GRCh38

View Full Product Details